Type:
Whole Allergen
Whole Allergen
Whole Allergen
Acarus siro
d70
Acaridae
Acarus siro
Inhalation (1)
Acarus siro
Flour mite, grain mite
Acarus siro is one of the Acaridae family of storage mites, known commonly as the ‘flour’ or ‘grain’ mite. Traditionally associated with occupational exposure in rural workers, storage mites have been more recently linked with non-occupational sensitization in urban environments, causing contact dermatitis, rhinoconjunctivitis and asthma. A. siro is the most common species of mite contaminating food and is primarily found in stored cereal products, but has also been reported in other foodstuffs such as meat, pulses, dried fruit and cheese. Importantly, A. siro is also found in the home, in soft furnishings, mattresses and pet beds, where house dust mites (HDM) are characteristically found. Allergenic cross-reactivity between storage mites and HDMs is well documented.
Acarus siro is mainly found in stored cereal-based food products, such as grain and flour (1), but has also been found in other food products, including cheese (2) (3), meat, pulses, dried fruit, tea and spices (4).
Of 571 UK shop-bought cereal-based food products, 21% were contaminated with storage mites. After home storage for 6 weeks, 38% of 421 food samples contained storage mites (1).
Storage mites are also found in the home; mattresses, soft furnishings, pets and pet bedding are all sources of storage mites (1).
Taxonomic tree of Acarus siro (5) |
|
Domain |
Eukaryota |
Kingdom |
Metazoa |
Phylum |
Arthropoda |
Subphylum |
Chelicerata |
Class |
Arachnida |
Subclass |
Acari |
Order |
Astigmata |
Family |
Acaridae |
Genus |
Acarus |
Mite feces are the most relevant source of allergens (6).
Sensitization to storage mites, including Acarus siro, is an occupational hazard for farm workers (7) and bakers (1). However, approximately 25% of patients sensitized to storage mites have no relationship with farms or bakeries (1).
Acarus siro is more common in temperate regions, as these meet its preferred physical conditions, but can be found worldwide (4).
Inhalation (1).
Contact dermatitis, commonly known as (‘grocer’s itch’) (2).
Mites are the sources of potent allergens that sensitize and induce IgE-mediated allergic reactions (8). Sensitization to storage mites causes rhinitis, asthma and conjunctivitis (8) as well as human acarine dermatitis (2).
Among 200 patients assessed for rhinitis and/or asthma in an outpatient setting, 123 (61.5%) had sensitization to at least one storage mite (9). Patients with storage mite sensitization had the following disease characteristics: rhinitis (96.7%), asthma (46.3%), rhinitis and asthma (43.1%), conjunctivitis (25.2%) and atopic dermatitis (9.8%) (9).
When compared with non-allergic participants without asthma, sensitization to A. siro is associated with increased odds of having nasal allergies without asthma, and of having asthma with nasal allergies, but is not associated with having asthma without nasal allergies (1).
Table adapted from Allergome.org (10)
Allergen |
Type |
Mass (kDa) |
Aca s 1 |
Cysteine protease |
37.3 |
Aca s 10 |
Tropomyosin |
33 |
Aca s 13 |
Fatty acid binding protein |
14.7 |
Aca s 2 |
NPC2 family |
14.6 |
Aca s 3 |
Serine protease |
27.8 |
Aca s 4 |
Amylase |
58 |
Aca s 7 |
Unknown |
23 |
Aca s 8 |
Glutathione S-transferase |
27.4 |
Allergens in storage mites are proteins with a variety of functions, including fatty acid-binding proteins, tropomyosin and paramyosin homologues, apoliphorine-like proteins, alfa tubulines and other allergens, such as group 2, 5 and 7 allergens. In particular, the allergen Aca s 13 (a fatty acid binding protein) reacted with serum from 23% of patients sensitized to A. siro (11).
Allergenic cross-reactivity between storage mites and HDMs is well documented (1); in a European Community Respiratory Health Survey, 8% of people were found to be sensitized to HDMs and 10% to storage mites. Among those patients with storage mite sensitization, 44% were also sensitized to HDM (1).
Another study reported that 84% (n/N=274/310) of patients sensitive to house dust mites were also sensitive to storage mites; 73% (n/N=227/310) of patients were sensitized to all three species of storage mite studied (Lepidoglyphus destructor, Tyrophagus putrescentiae and A. siro). In this study, none of the patients tested positive for A. siro alone. Approximately 70% of patients undergoing skin prick tests (SPTs) had negative skin reactions to A. siro; additionally, the positive SPT observed measured between 3-12 mm, with a very small percentage measuring >12 mm. When A. siro-specific IgE were measured in the participating patients, 25% had a negative result (12).
Aca s 13 (a fatty acid binding protein) may help identify HDM sensitization as 23% of patients tested were sensitized to A. siro (11).
Author: RubyDuke Communications
Reviewer: Dr. Christian Fischer
Last reviewed:May 2022