Table of Contents
Component
f439 Cor a 14
f439 Cor a 14 Scientific Information
Type:
Component
Name; WHO/IUIS:
Cor a 14
Biological function:
2S albumin
Allergen code:
f439
Molecular Weight:
15-16 kDa
Source Material:
Seed storage proteins Native sourced from the seed extracts of Corylus avellana
Summary
Cor a 14 is a water-soluble, thermoresistant, 2S albumin storage protein found in hazelnuts (Corylus avellana). It is a 147 amino-acid protein with a molecular weight of 15-16 kDa. Cor a 14 sensitization among hazelnut-sensitized individuals has been found in Central, Southern and North-western Europe as well as in the United States. Also, age-related variations in prevalence, with greater IgE-reactivity towards Cor a 14 in children than in adults, are also reported. Cor a 14 possesses two important features: its ability to induce direct (primary) sensitization to hazelnut, as opposed to pollen-related sensitization, and its association with more severe clinical reactions (skin, respiratory, gastrointestinal, or even anaphylaxis). Determination of specific immunoglobulin E (sIgE) for Cor a 14 can serve as a predictive marker for clinical allergy to hazelnut with moderate-to-severe symptoms and could differentiate between sensitization and clinical allergy. However, the predictive ability of Cor a 14-sIgE could be enhanced by combining it with Cor a 9-sIgE and thus alleviate the risk of misclassifying patients. Moreover, Cor a 14-sIgE determination could predict the oral food challenge (OFC) outcomes better than other allergens (Cor a 1, Cor a 8 and Cor a 9) and could thus reduce the number of OFC to be conducted. Furthermore, even though clinical cross-reactivity among 2S albumins is variable, significant sequence homology and identity of Cor a 14 has been reported with Jug r 1 (walnut), Car i 1 (pecan) and Ana o 3 (cashew nut).
Epidemiology
Worldwide distribution
Hazelnut allergy (Corylus avellana) is the most common tree nut allergy in Europe (1, 2). According to a systematic review, this allergy accounts for about 17-100% of the total tree nut allergies in Europe (3). Further, hazelnut allergy as well as sensitization is also frequent outside Europe, in countries such as India, Russia, Israel, Australia, Mexico and South Africa (4-9).
Cor a 14 is a storage protein of the 2S albumin family (10). In the EuroPrevall study conducted from 2005 to 2010 among 731 hazelnut-sensitized individuals in 12 European centers (central, eastern, north-western and southern countries), sensitization to Cor a 14 exhibited an overall prevalence of 5.9%. The highest prevalence was noted (18.2%-27.3%) in some cities of Central Europe (Prague [Czech Republic], Utrecht [The Netherlands], and Manchester [United Kingdom], Southern Europe (Madrid [Spain]) and North-western Europe (Reykjavik [Iceland]), while the prevalence in other cities of Central and Southern Europe was found to be <7% (11).
In the Netherlands, sensitization to Cor a 14 was found to be 34% (55 out of 161) in hazelnut-sensitized patients (81 children and 80 adults) (12), while in Sweden among 220 hazelnut-allergic and asthmatic children (10-≤18 years) and young adults (>18-35 years), sensitization to Cor a 14 was 10.4% (23 out of 220) (13). On the other hand, a study conducted in Turkey reported 66.7% prevalence of Cor a 14 sensitization, among hazelnut-sensitized children having a median age of 3.4 years (34 out of 51) (14).
Similarly, the prevalence of Cor a 14 among hazelnut-sensitized patients ranged from 26% to 32% in different regions of United States (US). It was further noted that the prevalence rates decreased with an increasing age (15).
Age-related variations in prevalence, with greater IgE-reactivity towards Cor a 14 in children than in adults, was reported in a few studies. In Belgium, sensitization to Cor a 14 was reported to be 90% in preschool children, 80% in school-age children and 29% in adults. Furthermore, 21% of infants with atopic dermatitis were found to be sensitized to Cor a 14 (16). A similar pattern was reported in a study conducted in Dutch children and adults, where sensitization to Cor a 14 was reported to be higher in children (70%) than in adults (39%) with objective symptoms (allergic symptoms seen on food challenge) or convincing history of hazelnut allergy (17).
Environmental Characteristics
Source and tissue
Cor a 14 is extracted from hazelnut, the seed of hazel tree (Corylus avellana). It is mainly a water-soluble, thermoresistant, 2S albumin (18).
Gel filtration and ion-exchange chromatography were used for the purification of natural Cor a 14 from defatted hazelnut extract (19, 20). It was further characterized by mass spectrometry, followed by circular dichroism or HPLC-HRMS (high-performance liquid chromatography-high resolution mass spectrometry) (19, 20). The molecule can also be generated by recombinant systems (19, 20).
Clinical Relevance
Disease severity
Similar to other nut storage proteins, Cor a 14 possesses two important features: its ability to induce direct (primary) sensitization to hazelnut, as opposed to pollen-related sensitization, and its association with more severe clinical reactions (skin, respiratory, gastrointestinal [GI], or even anaphylaxis) (18, 21, 22). As an example, the Europrevall study conducted among 731 hazelnut-sensitized individuals demonstrated a statistically significant association of Cor a 14 sensitization (p=0.006) with the prevalence of moderate-to-severe clinical presentations. In this study, Cor a 14 sensitization was present in 15.2% of patients with severe symptoms, 5.6% with moderate symptoms, and 3.7% with OAS (21).  Similar data were collected in a study conducted in Belgium among 119 hazelnut-sensitized patients, with Cor a 14 sensitization significantly more prevalent in patients with generalized reactions (erythema, urticaria, angioedema, GI symptoms, and respiratory symptoms) as compared to patients with OAS (76% vs. 5%, p<0.05) (16). Also, in another study involving 220 hazelnut-allergic asthmatic patients, individuals displaying positive specific immunoglobulin E (sIgE) to Cor a 14 or Cor a 9 were found to report more generalized reactions, like GI symptoms or anaphylaxis as compared to ones without Cor a 14 or Cor a 9 sensitization (13).
Further, sensitization to Cor a 14 was found to be associated with the presence of clinical allergy to hazelnut. This could be seen from two studies conducted in Belgium (n=15) and the Netherlands (n= 38), which showed no sensitization to Cor a 14 in each of the hazelnut-tolerant individuals (12, 16).
Cross-reactive molecules
Clinical cross-reactivity among 2S albumins is variable. However, co-sensitization of Cor a 14 with Jug r 1 (walnut) or Ana o 3 (cashew nut) may be present as observed in a study conducted among 10,503 hazelnut-sensitized individuals in the US. Strong co-sensitization was reported between Cor a 14 and Jug r 1 of walnut (correlation coefficient=0.7), with the co-sensitization being comparatively lower between Cor a 14 and Ana o 3 of cashew nut (correlation coefficient=0.43) (15).
A study conducted on 13 walnut and/or hazelnut-allergic patients sensitized to both nuts revealed strong cross-inhibition between the extracts and Cor a 14 or Jug r 1, indicating strong biological cross-reactivity and suggesting a potential risk of clinically relevant cross-reactivity (23).
Cor a 14 and peanut 2S albumin Ara h 2 are not found to be cross-reactive, as demonstrated in a study investigating peanut and hazelnut molecular profiles in 114 hazelnut-allergic patients, of whom 48% were also confirmed as peanut-allergic. Co-sensitization between Cor a 14 and Ara h 2 was virtually absent in hazelnut-allergic patients without peanut allergy, however reached up to 20% in peanut-allergic patients, with no hazelnut allergy, and to even 40% in patients with both hazelnut- and peanut-allergy (12). 
Molecular Aspects
Biochemistry
Cor a 14, a 147 amino-acid protein with a molecular weight of 15-16 kDa (10 kDa under reducing conditions), belongs to the 2S albumin family of storage proteins, a member of the prolamin superfamily of proteins (18, 19). 2S albumins are small, water soluble, globular heterodimers. Their subunits (8-10 kDa and 3-4 kDa) are connected by four disulfide bridges (18, 19). They are highly abundant in the seed cells and helpful during germination due to their ability to donate nitrogen and sulfur (18, 19).
The allergen has demonstrated high heat resistance (90℃) and stability to severe conditions of GI tract (18, 20). The allergenicity of Cor a 14 was found stable by roasting (hot air or infrared), till a temperature of 140℃, however was lost by roasting at 170℃ after infra-red processing (24).
Isoforms, epitopes, antibodies
As of 17th February 2021, only one isoallergen of Cor a 14 i.e., Cor a 14.0101 has been identified and officially published by the World Health Organization (WHO) and International Union of Immunological Societies (IUIS) Allergen Nomenclature (10, 18).
Cross-reactivity
As a rule, cross-reactivity among 2S albumins is variable; however, Cor a 14 shares significant sequence homology, ≥ 60%, with Jug r 1 (63%) and Car i 1 (62%), the walnut and pecan 2S albumins, allowing clinical cross-reactivity (18, 25, 26).  
Furthermore, the sequence identity of Cor a 14 with the 2S albumins of cashew nut (Ana o 3) was reported to be 43% (27) and with 2S albumin of peanut (Ara h 2) was reported to be 29%  (12).
Diagnostic Relevance
Disease Severity
Determination of sIgE for Cor a 14 can serve as a prediction marker for clinical allergy to hazelnut with moderate-to-severe symptoms (27) which holds true even in regions with less birch pollen-sensitization as reported by a study conducted on 82 individuals in Finland (28).
This predictive ability was best observed at a cut-off value of 0.35 kUA/L as reported in a meta-analysis involving 8 studies. However, it was suggested that rather than an independent test, the predictive ability of Cor a 14-sIgE could be enhanced by combining it with Cor a 9-sIgE and thus alleviate the risk of misclassifying patients (29). Furthermore, the predictive capacity of Cor a 14 determination by both sIgE antibodies and basophil activation, was confirmed by double-blind, placebo-controlled, food challenge (DBPCFC) test conducted on 8 children. The specificity of these tests (IgE antibodies and basophil activation) was found to be >97% and >94%, respectively with the sensitivity being 100%. Hence, Cor a 14 determination could differentiate between sensitization and clinical allergy (30). This was also shown in two studies conducted in Italy (n=36) and Denmark (n=155), wherein Cor a 14-sIgE determination could detect true hazelnut-allergic patients in 94% and 87% of overall patient population respectively (31, 32).
Based on the above studies, Cor a 14-sIgE determination could predict the oral food challenge (OFC) outcomes better than other allergens (Cor a 1, Cor a 8 and Cor a 9) and could thus reduce the number of OFC to be conducted (14, 22). This was evidenced from several studies which confirmed the ability of Cor a 14-sIgE in differentiating positive and negative outcomes after OFC. Cor a 14-sIgE was detected in 96.2% reactive vs. 36% non-reactive patients in the study conducted among 64 hazelnut-sensitized patients in Turkey (14). Similarly, in a US study, it was detectable in 85% and 35% of reactive and non-reactive patients, respectively (33). In addition, two studies conducted on 161 and 147 Dutch hazelnut-sensitized individuals, also confirmed higher levels of Cor a 14-sIgE in patients with objective symptoms (allergic symptoms seen on food challenge or convincing history of hazelnut allergy) (17, 34). 
Cross-Reactivity
Cor a 14 is a marker of primary sensitization to hazelnut; its cross-reactivity is limited to Jug r 1 from walnut and Car i 1 from pecan (25, 26).
Exposure
The main route of exposure is through ingestion (21).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Christian Fischer
Last reviewed: March 2021
References
  1. Barni S, Mori F, Piccorossi A, Sarti L, Pucci N, Maresca M, et al. Low-Dose Oral Food Challenge with Hazelnut: Efficacy and Tolerability in Children. International archives of allergy and immunology. 2019;178(1):97-100.
  2. Calamelli E, Trozzo A, Di Blasi E, Serra L, Bottau P. Hazelnut Allergy. Medicina (Kaunas, Lithuania). 2021;57(1).
  3. McWilliam V, Koplin J, Lodge C, Tang M, Dharmage S, Allen K. The Prevalence of Tree Nut Allergy: A Systematic Review. Current Allergy and Asthma Reports. 2015;15(9):54.
  4. Botha M, Basera W, Facey-Thomas HE, Gaunt B, Gray CL, Ramjith J, et al. Rural and urban food allergy prevalence from the South African Food Allergy (SAFFA) study. The Journal of allergy and clinical immunology. 2019;143(2):662-8.e2.
  5. Elizur A, Appel MY, Nachshon L, Levy MB, Epstein-Rigbi N, Golobov K, et al. NUT Co Reactivity - ACquiring Knowledge for Elimination Recommendations (NUT CRACKER) study. Allergy. 2018;73(3):593-601.
  6. Fedorova OS, Ogorodova LM, Fedotova MM, Evdokimova TA. [The prevalence of food allergy to peanut and hazelnut in children in Tomsk Region]. Voprosy pitaniia. 2014;83(1):48-54.
  7. Mahesh PA, Wong GW, Ogorodova L, Potts J, Leung TF, Fedorova O, et al. Prevalence of food sensitization and probable food allergy among adults in India: the EuroPrevall INCO study. Allergy. 2016;71(7):1010-9.
  8. McWilliam V, Peters R, Tang MLK, Dharmage S, Ponsonby AL, Gurrin L, et al. Patterns of tree nut sensitization and allergy in the first 6 years of life in a population-based cohort. The Journal of allergy and clinical immunology. 2019;143(2):644-50.e5.
  9. Ruiz Segura LT, Figueroa Pérez E, Nowak-Wegrzyn A, Siepmann T, Larenas-Linnemann D. Food allergen sensitization patterns in a large allergic population in Mexico. Allergologia et immunopathologia. 2020:In press.
  10. WHO/IUIS. Cor a 14 - Corylus avellana: WHO/IUIS Allergen Nomenclature Sub-Committee; 2021 [updated January 19, 2019February 16, 2021]. Available from: http://www.allergen.org/viewallergen.php?aid=233.
  11. Datema MR, Zuidmeer-Jongejan L, Asero R, Barreales L, Belohlavkova S, de Blay F, et al. Hazelnut allergy across Europe dissected molecularly: A EuroPrevall outpatient clinic survey. The Journal of allergy and clinical immunology. 2015;136(2):382-91.
  12. Masthoff LJ, van Hoffen E, Mattsson L, Lidholm J, Andersson K, Zuidmeer-Jongejan L, et al. Peanut allergy is common among hazelnut-sensitized subjects but is not primarily the result of IgE cross-reactivity. Allergy. 2015;70(3):265-74.
  13. Johnson J, Malinovschi A, Lidholm J, Petersson CJ, Nordvall L, Janson C, et al. Sensitization to storage proteins in peanut and hazelnut is associated with higher levels of inflammatory markers in asthma. Clin Mol Allergy. 2020;18:11.
  14. Buyuktiryaki B, Cavkaytar O, Sahiner UM, Yilmaz EA, Yavuz ST, Soyer O, et al. Cor a 14, Hazelnut-Specific IgE, and SPT as a Reliable Tool in Hazelnut Allergy Diagnosis in Eastern Mediterranean Children. J Allergy Clin Immunol Pract. 2016;4(2):265-72.e3.
  15. Valcour A, Lidholm J, Borres MP, Hamilton RG. Sensitization profiles to hazelnut allergens across the United States. Ann Allergy Asthma Immunol. 2019;122(1):111-6.e1.
  16. Faber MA, De Graag M, Van Der Heijden C, Sabato V, Hagendorens MM, Bridts CH, et al. Cor a 14: missing link in the molecular diagnosis of hazelnut allergy? International archives of allergy and immunology. 2014;164(3):200-6.
  17. Masthoff LJ, Mattsson L, Zuidmeer-Jongejan L, Lidholm J, Andersson K, Akkerdaas JH, et al. Sensitization to Cor a 9 and Cor a 14 is highly specific for a hazelnut allergy with objective symptoms in Dutch children and adults. The Journal of allergy and clinical immunology. 2013;132(2):393-9.
  18. Costa J, Mafra I, Carrapatoso I, Oliveira MB. Hazelnut Allergens: Molecular Characterization, Detection, and Clinical Relevance. Critical reviews in food science and nutrition. 2016;56(15):2579-605.
  19. Korte R, Happe J, Brümmer I, Brockmeyer J. Structural Characterization of the Allergenic 2S Albumin Cor a 14: Comparing Proteoform Patterns across Hazelnut Cultivars. J Proteome Res. 2017;16(2):988-98.
  20. Pfeifer S, Bublin M, Dubiela P, Hummel K, Wortmann J, Hofer G, et al. Cor a 14, the allergenic 2S albumin from hazelnut, is highly thermostable and resistant to gastrointestinal digestion. Mol Nutr Food Res. 2015;59(10):2077-86.
  21. Datema MR, van Ree R, Asero R, Barreales L, Belohlavkova S, de Blay F, et al. Component-resolved diagnosis and beyond: Multivariable regression models to predict severity of hazelnut allergy. Allergy. 2018;73(3):549-59.
  22. van Ree R, Aalberse RC. Allergens and the Allergenic Composition of Source Materials. In: Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Ollert M, editors. EAACI Molecular Allergology User's Guide: Using Molecular Allergology in the Clinical Practice. Zutich, Switzerland: The European Academy of Allergy and Clinical Immunology (EAACI); 2016. p. 11-20.
  23. Villalta D, Scala E, Mistrello G, Amato S, Asero R. Evidence of Cross-Reactivity between Different Seed Storage Proteins from Hazelnut (Corylus avellana) and Walnut (Juglans regia) Using Recombinant Allergen Proteins. International archives of allergy and immunology. 2019;178(1):89-92.
  24. Lamberti C, Nebbia S, Antoniazzi S, Cirrincione S, Marengo E, Manfredi M, et al. Effect of hot air and infrared roasting on hazelnut allergenicity. Food Chem. 2020:128174.
  25. Knol EF, Wickman M. Tree Nut and Seed Allergy. In: Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Ollert M, editors. EAACI Molecular Allergology User's Guide: Using Molecular Allergology in the Clinical Practice. Zutich, Switzerland: The European Academy of Allergy and Clinical Immunology (EAACI); 2016. p. 245-54.
  26. Scala E, Villalta D, Meneguzzi G, Giani M, Asero R. Storage molecules from tree nuts, seeds and legumes: relationships and amino acid identity among homologue molecules. European annals of allergy and clinical immunology. 2018;50(4):148-55.
  27. Bastiaan-Net S, Batstra MR, Aazamy N, Savelkoul HFJ, van der Valk JPM, Gerth van Wijk R, et al. IgE cross-reactivity measurement of cashew nut, hazelnut and peanut using a novel IMMULITE inhibition method. Clin Chem Lab Med. 2020;58(11):1875-83.
  28. Uotila R, Röntynen P, Pelkonen AS, Voutilainen H, Kaarina Kukkonen A, Mäkelä MJ. For hazelnut allergy, component testing of Cor a 9 and Cor a 14 is relevant also in birch-endemic areas. Allergy. 2020;75(11):2977-80.
  29. Nilsson C, Berthold M, Mascialino B, Orme M, Sjölander S, Hamilton R. Allergen components in diagnosing childhood hazelnut allergy: Systematic literature review and meta-analysis. Pediatr Allergy Immunol. 2020;31(2):186-96.
  30. Brandström J, Nopp A, Johansson SG, Lilja G, Sundqvist AC, Borres MP, et al. Basophil allergen threshold sensitivity and component-resolved diagnostics improve hazelnut allergy diagnosis. Clin Exp Allergy. 2015;45(9):1412-8.
  31. Carraro S, Berardi M, Bozzetto S, Baraldi E, Zanconato S. COR a 14-specific IgE predicts symptomatic hazelnut allergy in children. Pediatr Allergy Immunol. 2016;27(3):322-4.
  32. Eller E, Mortz CG, Bindslev-Jensen C. Cor a 14 is the superior serological marker for hazelnut allergy in children, independent of concomitant peanut allergy. Allergy. 2016;71(4):556-62.
  33. Kattan JD, Sicherer SH, Sampson HA. Clinical reactivity to hazelnut may be better identified by component testing than traditional testing methods. J Allergy Clin Immunol Pract. 2014;2(5):633-4 e1.
  34. Masthoff LJN, Blom WM, Rubingh CM, Klemans RJB, Remington BC, Bruijnzeel-Koomen C, et al. Sensitization to Cor a 9 or Cor a 14 has a strong impact on the distribution of thresholds to hazelnut. J Allergy Clin Immunol Pract. 2018;6(6):2112-4.e1.