Table of Contents
Component
f354 Ber e 1
f354 Ber e 1 Scientific Information
Type:
Component
Name; WHO/IUIS:
Ber e 1
Biological function:
2S albumin
Allergen code:
f354
Molecular Weight:
9 kDa
Other Names :
2S Albumin
Summary
Ber e 1 is a 9-kDa, 2S albumin recognized as the major allergen of Brazil nut, that is resistant to both, heat as well as pepsin digestion. This allergen belongs to the prolamin superfamily of proteins. The prevalence of allergy to Brazil nut has frequently been found in the United States and United Kingdom (UK), with specific immunoglobulin E reactivity to Ber e 1 commonly reported in individuals living in the UK. Interestingly, Ber e 1 has been considered as the first food allergen that may pass on from one plant to the other via genetic engineering technology. Furthermore, patients sensitive to Ber e 1 allergen may develop severe reactions, such as urticaria, laryngeal edema, or even anaphylactic shock. Ber e 1 has displayed sequence identity with 2S albumins from cocoa, upland cotton and English walnut; whereas sequence similarity with 2S albumins from pumpkin, arabdopsis, mabinlin, and rapeseed. Additionally, sequence identity and structural homology has also been found with 2S albumins of sesame and sunflower.
Epidemiology
Worldwide distribution
Allergy to Brazil nuts (Bertholletia excelsa) is frequently been reported in the United Kingdom (UK) as well as in the United States (US) (1). A systematic review involving 36 studies reported the prevalence of Brazil nut allergy to be between 24% and 33% in the UK and between 12% and 22% in the US, among tree-nut-allergic individuals (2).
A study was conducted in Italy on 11 symptomatic Brazil nut-allergic patients (severe anaphylaxis due to consumption of Brazil nut) and 10 asymptomatic patients (no clinical symptoms, but having positive immunoglobulin E (IgE) reactivity to Brazil nut). It was found that 100% of symptomatic patients showed IgE reactivity to Ber e 1, while none of the asymptomatic patients were positive for specific IgE (sIgE) to Ber e 1 (3).
In a study conducted on 56 patients who were evaluated for Brazil nut allergy, sIgE to recombinant (r) Ber e 1 was measured in 36 patients, among which 16 were found to be positive (44.4%) (4). Besides, another study conducted in UK among 18 patients with Brazil nut allergy, sIgE reactivity to Ber e 1 was reported in 94.4% of patients (17 out of 18) (5).
Interestingly, Ber e 1 is a protein rich in sulfur, containing amino acids like methionine (18% of total protein) and cysteine (8%). This property of 2S albumin of Brazil nut has made it as a   potential candidate in increasing the nutritional value of certain foods (tobacco, rapeseed, beans) and legumes (soybean and Vicia narbonensis) via genetic engineering technology. However, since Brazil nut is also reported to be allergic, the genetically-modified foods with Ber e 1 could present with allergic reactions, and hence should be considered for its safety. In line with this, a study evaluated the IgE reactivity to Ber e 1 (both Brazil nut 2S albumin and transgenic 2S albumin in soybean) in 9 Brazil nut-allergic patients. The Ber e 1-sIgE was found to be positive in 88.9% (8 out of 9) of these individuals. Also, 77.8% (7 out of 9) of Brazil nut-allergic patients showed IgE reactivity towards transgenic soybean protein, similar to 2S albumin of Brazil nut. In addition, it was found that this protein was not present in the non-transgenic extract of soybean (6). Thus, Ber e 1 could be considered as the first food allergen that may pass on from one plant to the other (7). 
Environmental Characteristics
Source and tissue
Ber e 1 is a major allergen found in the seed of Brazil nut tree. It is mainly a water-soluble, seed-storage protein, that constitutes of 30% of the total protein in the seeds (7).
The sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) and size-exclusion chromatography were used for the purification of natural Ber e 1 from water-soluble fraction of Brazil nut extract (3, 8). The molecule can also be generated by recombinant systems in Pichia pastoris, and further characterized by circular dichroism. (7). 
Clinical Relevance
Disease Severity
Ber e 1 is reported to be the major allergen of Brazil nut and found to be associated with the clinical expression of Brazil nut allergy in sensitized individuals (3). Furthermore, the IgE reactivity to 2S albumin of Brazil nut i.e. Ber e 1 has been reported to be correlated with the food-induced-anaphylaxis (9). In a study conducted on 11 symptomatic patients manifesting anaphylaxis (urticaria, laryngeal edema, or anaphylactic shock) on ingestion of Brazil nuts, Ber e 1 was recognized by the sera of all the patients (100%). In the same study, 10 non-symptomatic Brazil nut-sIgE sensitized individuals were studied, who were found negative for Ber e 1-sIgE (3).
Further, another study was conducted on a cohort of 36 patients with suspected allergy to Brazil nut. The patients were considered as true positive, if they reacted positive on oral challenge or skin prick test with a history of Brazil nut allergy, whereas the patients were considered as true negative if the oral challenge was negative or presented with a history of clinical non-reactivity. The sIgE reactivity to Ber e 1 was reported in 15 true positive patients, while it was found only in one true negative patient (4).
A recent study has postulated that naturally-existing plant lipids may play an essential role in the inherent allergenicity of Ber e 1 (10).
Molecular Aspects
Biochemistry
Ber e 1 is part of 2S albumin of the prolamin superfamily of proteins (7). The 2S albumins are small, water-soluble, globular proteins that consist of 2 subunits: 8 to10 kDa (large) and 3 to 4 kDa (small), connected via four disulfide linkages (11). The four α-helix structure, characteristic of 2S albumin, is put together via disulfide bridges (5). Further, these proteins possess large content of asparagine, arginine, cysteine, and glutamine residues. They are found in abundance in the seed cells and are helpful during germination as a result of their ability to donate nitrogen and sulfur (11). 
Ber e 1 is rich in methionine in addition to cysteine as compared to other 2S albumins (12). It has a molecular weight of approximately 12 kDa, which comprises a large subunit of ~9 kDa and a small subunit of ~3 kDa. The Ber e 1 precursor protein is a 18 kDa protein, which during the process of seed maturation, forms an intermediary protein of 15 kDa. Furthermore, this 15 kDa protein cleaves into a 12 kDa protein and finally matures into a 9 kDa and 3 kDa protein (7).
A characteristic property of Ber e 1 is that the hydrophobic cavity of the protein (located between the 3rd and 4th helix) is similar to non-specific lipid transfer proteins, and it can enclose a lipid component, that could be immunologically active. Further, Ber e 1 is unique in its interaction with copper ions, which can affect its allergenic potential (12, 13).
Ber e 1 has been found to be thermostable and hence, roasting of the allergen at 100°C for 20 min was not able to alleviate the allergenicity of the molecule. It has also been found to resist pepsin digestion (14). Moreover, based on the pH, the denaturation temperature of Ber e 1 was found to be between 80°C to 110°C (12).  Furthermore, it has been reported that even alkaline treatment of the allergen was shown to be ineffective in reducing the immunoreactivity (14). The high stability of the molecule is attributed to the four disulfide linkages, which maintain the tight fold of the protein (13). However, the disulfide linkage does not influence the allergenic potential of Ber e 1 (14).
Isoforms, epitopes, antibodies
Ber e 1.0101 is the major isoform of Ber e 1 (15).
Ber e 1 is encoded by multigene and is reported to possess more than 20 isoforms (7).
Furthermore, accumulating data from various studies have reported that Ber e 1-sIgE mainly binds to long and non-linear epitopes rather than linear epitopes as seen in other 2S albumins, such as Ara h 2 (peanut) (5, 7).
Cross-reactivity
Several 2S albumins from different sources have shown sequence similarity and/or identity among them. Ber e 1 was reported to be 36%, 39% and 44% identical to 2S albumins from cocoa (Theobroma cacao), upland cotton (Gossypium hirsutum) and English walnut (Juglans regia), respectively. Furthermore, the sequence similarity of 53%, 50%, 42%, 40% and >52% was observed with 2S albumins of pumpkin (Cucurbita maxima), arabdopsis (Arabidopsis thaliana), mabinlin (Capparis masaikai), rapeseed (Brassica napus), and cocoa, respectively (16, 17).
The structural homology between Ses i 2 (2S albumin from sesame seeds) and Ber e 1 was reported to be 40%. Moreover, the other 2S albumin from sesame i.e. Ses i 1 was reported to have some degree of amino-acid sequence homology with Ber e 1 (18). Furthermore, sequence homology between 2S albumin of sunflower and Ber e 1 was found to be 34% (19).
Common epitopes for IgE binding have been reported from Ber e 1 and other 2S albumins, like yellow mustard (Sin a 1), oriental mustard (Bra j 1) as well as English walnut (Jug r 1) (20).
Diagnostic Relevance
Disease Severity
Ber e 1 has been found to be the only allergen in Brazil nut, associated with clinical manifestations of its allergy. A study was conducted on 11 patients with positive IgE and clinical symptoms to Brazil nut (symptomatic) and 10 patients with only positive IgE but no clinical symptoms (non-symptomatic). The results revealed that only the 11 symptomatic patients were positive for sIgE to Ber e 1, while none of the 10 non-symptomatic patients showed IgE reactivity to Ber e 1. This suggests that sensitization to Ber e 1 is associated with the development of clinical allergy to Brazil nut (3).
Furthermore, it was found that sIgE-Ber e 1 could help differentiate patients with a true allergy to Brazil nuts, from the patients who showed only sensitization and no clinical symptoms towards Brazil nut ingestion (4).
Exposure
The main route of exposure is through ingestion (4).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: February 2021
References
  1. Borres MP, Maruyama N, Sato S, Ebisawa M. Recent advances in component resolved diagnosis in food allergy. Allergol Int. 2016;65(4):378-87.
  2. McWilliam V, Koplin J, Lodge C, Tang M, Dharmage S, Allen K. The Prevalence of Tree Nut Allergy: A Systematic Review. Curr Allergy Asthma Rep. 2015;15(9):54.
  3. Pastorello EA, Farioli L, Pravettoni V, Ispano M, Conti A, Ansaloni R, et al. Sensitization to the major allergen of Brazil nut is correlated with the clinical expression of allergy. J Allergy Clin Immunol. 1998;102(6 Pt 1):1021-7.
  4. Rayes H, Raza AA, Williams A, Matthews S, Arshad SH. Specific IgE to recombinant protein (Ber e 1) for the diagnosis of Brazil nut allergy. Clin Exp Allergy. 2016;46(4):654-6.
  5. Alcocer MJ, Murtagh GJ, Wilson PB, Progias P, Lin J, Archer DB. The major human structural IgE epitope of the Brazil nut allergen Ber e 1: a chimaeric and protein microarray approach. J Mol Biol. 2004;343(3):759-69.
  6. Nordlee JA, Taylor SL, Townsend JA, Thomas LA, Bush RK. Identification of a Brazil-nut allergen in transgenic soybeans. N Engl J Med. 1996;334(11):688-92.
  7. Alcocer M, Rundqvist L, Larsson G. Ber e 1 protein: the versatile major allergen from Brazil nut seeds. Biotechnol Lett. 2012;34(4):597-610.
  8. Murtagh GJ, Archer DB, Dumoulin M, Ridout S, Matthews S, Arshad SH, et al. In vitro stability and immunoreactivity of the native and recombinant plant food 2S albumins Ber e 1 and SFA-8. Clin Exp Allergy. 2003;33(8):1147-52.
  9. Pastorello EA, Pompei C, Pravettoni V, Brenna O, Farioli L, Trambaioli C, et al. Lipid transfer proteins and 2S albumins as allergens. Allergy. 2001;56 Suppl 67:45-7.
  10. Wang R, Ghumra A, Cochrane S, Fairclough L, Broughton R, Michaelson LV, et al. Defining lipids and T cell receptors involved in the intrinsic allergenicity of nut proteins. Clin Transl Allergy. 2020;10(1):54.
  11. Mendes C, Costa J, Vicente AA, Oliveira M, Mafra I. Cashew Nut Allergy: Clinical Relevance and Allergen Characterisation. Clinical reviews in allergy & immunology. 2019;57(1):1-22.
  12. Kluczkovski A, Scussel V. Brazil nut allergy: A review. African Journal of Pharmacy and Pharmacology. 2015;9:633-44.
  13. Rundqvist L, Tengel T, Zdunek J, Bjorn E, Schleucher J, Alcocer MJ, et al. Solution structure, copper binding and backbone dynamics of recombinant Ber e 1-the major allergen from Brazil nut. PLoS One. 2012;7(10):e46435.
  14. Vanga SK, Raghavan V. Processing effects on tree nut allergens: A review. Crit Rev Food Sci Nutr. 2017;57(17):3794-806.
  15. WHO/IUIS. Allergen Nomenclature "Ber e 1": WHO/IUIS Allergen Nomenclature Sub-Committee; 2020 [updated 16 July 201928 January 2021]. Available from: http://www.allergen.org/viewallergen.php?aid=127.
  16. Kochhar S, Gartenmann K, Guilloteau M, McCarthy J. Isolation and characterization of 2S cocoa seed albumin storage polypeptide and the corresponding cDNA. J Agric Food Chem. 2001;49(9):4470-7.
  17. Lopes JP, Kattan J, Doppelt A, Nowak-Wegrzyn A, Bunyavanich S. Not so sweet: True chocolate and cocoa allergy. J Allergy Clin Immunol Pract. 2019;7(8):2868-71.
  18. Beyer K, Bardina L, Grishina G, Sampson HA. Identification of sesame seed allergens by 2-dimensional proteomics and Edman sequencing: seed storage proteins as common food allergens. J Allergy Clin Immunol. 2002;110(1):154-9.
  19. Kelly JD, Hlywka JJ, Hefle SL. Identification of sunflower seed IgE-binding proteins. Int Arch Allergy Immunol. 2000;121(1):19-24.
  20. Moreno FJ, Mellon FA, Wickham MS, Bottrill AR, Mills EN. Stability of the major allergen Brazil nut 2S albumin (Ber e 1) to physiologically relevant in vitro gastrointestinal digestion. FEBS J. 2005;272(2):341-52.
  21. Tengel T, Alcocer MJ, Schleucher J, Larsson G. Complete assignment and secondary structure of the Brazil nut allergen Ber e 1. J Biomol NMR. 2005;32(4):336.