Whole Allergen
f278 Bay leaf
f278 Bay leaf Scientific Information
Type:
Whole Allergen
Display Name:
Bay leaf
Allergen code:
f278
Family:
Lauraceae
Latin Name:
Laurus nobilis
Other Names:
Bay leaf, Sweet Bay, Sweet Laurel, Laurel leaf, True Laurel, Mediterranean Bay leaf
Clinical Relevance
IgE-mediated reactions
Anecdotal evidence suggests that Bay leaf may cause symptoms of food allergy in sensitised individuals; however, no studies have been reported to date. A number of reports have described allergic dermatitis resulting from contact with Bay leaf. (1-4) Occupational asthma from inhalation of Bay leaf allergen may occur. (5)
The prevalence of allergy to Bay leaf could be expected to be higher in occupational settings, in particular in the food industry.
An individual with occupational asthma caused by several aromatic herbs –Thyme, Rosemary, Bay Leaf, and Garlic – has been described. The diagnosis was confirmed by several inhalation challenges. (5)
Allergic contact dermatitis from the leaf and the oil has been described. (1, 3) A 55-year-old woman presented with erythema and oedema over her knees, 3 days after the application of laurel oil to relieve joint pain. A patch test to the oil was positive. (4) Allergic contact dermatitis was reported following a massage with a mixture of olive oil and L. nobilis oil. Patch testing was 3+ positive to laurel oil and 2+ to the mixture of olive oil and laurel oil. (6)
A study examined the frequency of dermatologic allergic reactions to the Asteraceae species, using a Compositae mix; 118 of 3 851 (3.1%) individuals tested were positive. Further tests for Laurel oil were positive in 50.5% of cases.(7) Allergic contact dermatitis may be sesquiterpene-lactone-induced. (8)
Other reactions
A number of studies have indicated that Bay leaf may result in physical obstruction, including duodenal obstruction secondary to Bay leaf impaction. (9) Dried Bay leaf was described as an unusual cause of upper gastrointestinal tract haemorrhage,(10) and as an uncommon foreign body in the hypopharynx. (11)
Molecular Aspects
No allergens from this plant have yet been characterized.
Cross-reactivity
An extensive cross-reactivity among the different individual species of the genus could be expected, but in fact does not occur frequently.(12)
Cross-sensitization has been demonstrated between 2 allergenic plants, Frullania and Laurus Nobilis. (3, 13)
Compiled By
Last reviewed: April 2022
References
  1. Cheminat A, Stampf JL, Benezra C. Allergic contact dermatitis to laurel (Laurus nobilis L.): isolation and identification of haptens. Archives of dermatological research. 1984;276(3):178-81.
  2. Farkas J. Perioral dermatitis from marjoram, bay leaf and cinnamon. Contact dermatitis. 1981;7(2):121.
  3. Foussereau J, Muller JC, Benezra C. Contact allergy to Frullania and Laurus Nobilis: cross-sensitization and chemical structure of the allergens. Contact dermatitis. 1975;1(4):223-30.
  4. Ozden MG, Oztaş P, Oztaş MO, Onder M. Allergic contact dermatitis from Laurus nobilis (laurel) oil. Contact dermatitis. 2001;45(3):178.
  5. Lemière C, Cartier A, Lehrer SB, Malo JL. Occupational asthma caused by aromatic herbs. Allergy. 1996;51(9):647-9.
  6. Adişen E, Onder M. Allergic contact dermatitis from Laurus nobilis oil induced by massage. Contact dermatitis. 2007;56(6):360-1.
  7. Hausen BM. A 6-year experience with compositae mix. American journal of contact dermatitis : official journal of the American Contact Dermatitis Society. 1996;7(2):94-9.
  8. Gonçalo M, Gonçalo S. Allergic contact dermatitis from Dittrichia viscosa (L.) Greuter. Contact dermatitis. 1991;24(1):40-4.
  9. Tsang TK, Flais MJ, Hsin G. Duodenal obstruction secondary to bay leaf impaction. Annals of internal medicine. 1999;130(8):701-2.
  10. Skok P. Dried bay leaf: an unusual cause of upper gastrointestinal tract hemorrhage. Endoscopy. 1998;30(3):S40-1.
  11. Awerbuck DC, Briant TD, Wax MK. Bay leaf: an uncommon foreign body of the hypopharynx. Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery. 1994;110(3):338-40.
  12. L Y. Botanical relations and immunological cross-reactions in pollen allergy. 2nd ed Pharmacia Diagnostics AB Uppsala Sweden. 1982.
  13. Asakawa Y, Benezra C, Ducombs G, Foussereau J, Muller JC, Ourisson G. Editorial: Cross-sensitization between Frullania and Laurus nobilis: The allergen laurel. Archives of dermatology. 1974;110(6):957.