Whole Allergen
f279 Chili Pepper
f279 Chili Pepper Scientific Information
Type:
Whole Allergen
Display Name:
Chili Pepper
Allergen code:
f279
Family:
Solanaceae
Latin Name:
Capsicum annuum
Other Names:
Chili Pepper
Summary
In the Solanaceae family, the genus Capsicum includes C. annuum with common names chili pepper, paprika/sweet pepper, bell pepper, cayenne pepper and jalapeños.
Extensive IgE cross-reactivity among members of C. annuum is anticipated due to their close botanical relationship.
Allergy to Capsicum species is rare. Symptoms can occur after ingestion or inhalation of spice dust.
Route of Exposure
C. annuum is the primary species of genus Capsicum with more than 200 common names in use. The most common include chili pepper, paprika (sweet varieties), bell pepper, cayenne pepper and Jalapeños (Zhigila 2014).
Probably native to the tropics, these peppers produced flavourings that became known in the West through the spice trade. They are now grown mainly in China, Japan, Malaya, the Caribbean, Mexico, Hawaii, the Middle East, and Spain. Chili peppers tend to be eaten in very small amounts as spices or condiments, due to their strong flavours; as opposed to Sweet pepper, which is usually used as a vegetable.
Allergen exposure can occur by ingestion, skin contact or inhalation.
Clinical Relevance
Allergy to Capsicum species is rare. Symptoms can occur after ingestion or inhalation of spice dust. An extensive IgE cross-reactivity between members of C. annuum is expected, e.g. between chili pepper and sweet pepper (Takei 2022).
In a French series of 202 labial food challenges performed in 142 children with suspected food allergy, 156 were positive. The most common foods provoking reactions were egg white (75 cases), peanut (60 cases), mustard (23 cases), cow's milk (13 cases), and cod (8 cases). One child was positive to chili pepper (Rance 1997).
An Indian study of 24 children (aged 3 to 15 years) with documented deterioration in control of their perennial asthma – the purpose of which was to evaluate the possible effect of a specific elimination diet on symptoms – reported that specific IgE was raised to chili in 3 children (12.5%) (Agarkhedkar 2005).
Chili pepper has been implicated as a cause of eosinophilic esophagitis. In a study of 16 men and 7 women aged 18-57, nineteen had serum IgE specific for one or more food-associated allergens (median, 5 foods), with wheat, tomato, carrot, and onion identified most commonly. Sensitisation to chili pepper was documented in 2 cases (9%) (Roy-Ghanta 2008).
A severe asthma attack in a patient with premenstrual asthma was attributed to being triggered by chili pepper (Dikensoy 2001) and an anaphylactic reaction to pickled chili pepper was reported by (Galleani 2023).
‘Hunan hand’ is a contact dermatitis resulting from the direct handling of chili peppers containing capsaicin (Williams 1995) and erythema multiforme-like contact dermatitis has been reported (Raccagni 1995).
Molecular Aspects
Allergen Description
No allergens from chili pepper are described in the International Union of Immunological Societies Allergen Nomenclature ( http://www.allergen.org ).
However, the following allergens have been characterised from C. annuum sweet pepper (http://www.allergen.org), and chili pepper may contain similar allergens resulting in a high degree of IgE cross-reactivity between chili and sweet pepper.
Cap a 1, a thaumatin-like protein (Hoffmann-Sommergruber 2001)
Cap a 2, a profilin (Jensen-Jarolim 1998. Ebner 1998)
Cap a 7, Gibberellin-regulated protein (Takei 2022)
Other allergens detected include a Bet v 1 homologue, (Jensen-Jarolim 1998), an ascorbic acid oxidase and a 1,3-beta-glucanase (Wagner 2004) and a chitinase (Lee 2006).
In a study that demonstrated IgE reactivity to multiple spice allergens in workers exposed to high levels of inhalable spice dust, allergens of approximately 40 and 52 kDa were detected in chili pepper (Van der Walt 2010). Chili pepper IgE-binding proteins between 60 – 100 kDa was detected in (Galleani 2023).
Potential Cross Reactivity
Due to their close botanical relationship (Zhigila 2014, Olatunji 2018), extensive IgE cross-reactivity is expected among members of C. annuum, such as between chili pepper and paprika/sweet pepper (Takei 2022), and cayenne pepper (Airaksinen 2015).
Cross-reactivity to other plants containing the panallergens profilin and Bet v 1 could be expected to occur frequently. Patients with allergy to pepper and associated pollinosis show a high frequency of IgE reactivity to profilin (Wagner 2001).
Reviewed by Dr. Magnus Borres, March 2025
References
Agarkhedkar SR, Bapat HB, Bapat BN. Avoidance of food allergens in childhood asthma. Indian Pediatr 2005;42(4):362-366.
Airaksinen L, Riekki R, Vuokko A, Puustinen A. Paprika Rhinoconjunctivitis Case Reveals New Occupational Capsicum Allergens. American J Industrial Medicine 2015; 58:791–794.
Dikensoy O, Bayram NG, Filiz A. Severe asthma attack in a patient with premenstrual asthma: hot pepper is the possible trigger. Respiration 2001;68(2):227.
Ebner C, Jensen-Jarolim E, Leitner A, Breiteneder H. Characterization of allergens in plant-derived spices: Apiaceae spices, pepper (Piperaceae), and paprika (bell peppers, Solanaceae). Allergy 1998;53(46 Suppl):52-54.
Galleani C, Valdelvira R, Carmen Diéguez M, Crespo JF, Cabanillas B. Anaphylaxis to pickled chili pepper (Capsicum frutescens): Role of pickling processing in the allergic reactivity. Ann Allergy Asthma Immunol. 2023;130(3):369-370.
Hoffmann-Sommergruber K, Witty M, Campillo N, Krebitz M, Fuchs H, et al. Pru P 2, Mal d 2 and Cap a 1 are members of the thaumatin family and allergens: common structural features [Poster] 8th International Symposium on Problems of Food Allergy, Venice. 2001;11-13.
Jensen-Jarolim E, Santner B, Leitner A, Grimm R, Scheiner O, Ebner C, Breiteneder H. Bell peppers (Capsicum annuum) express allergens (profilin, pathogenesis-related protein P23 and Bet v 1) depending on the horticultural strain. Int Arch Allergy Immunol 1998;116(2):103-109.
Lee MF, Hwang GY, Chen YH, Lin HC, Wu CH. Molecular cloning of Indian jujube (Zizyphus mauritiana) allergen Ziz m 1 with sequence similarity to plant class III chitinases. Mol Immunol 2006; 43:1144-1151.
Olatunji, T. L.& Afolayan, A. J. Contributions to the Classification of Capsicum annuum L. and Capsicum frutescens L. in West Africa Using Morphological Traits. Notulae Botanicae Horti Agrobotanici Cluj-Napoca. 2018; 47:135–142.
Raccagni AA, Bardazzi F, Baldari U, Righini MG. Erythema-multiforme-like contact dermatitis due to capsicum. Contact Dermatitis 1995;33(5):353-354.
Rance F, Dutau G. Labial food challenge in children with food allergy. Pediatr Allergy Immunol 1997;8(1):41-44.
Roy-Ghanta S, Larosa DF, Katzka DA. Atopic characteristics of adult patients with eosinophilic esophagitis. Clin Gastroenterol Hepatol 2008;6(5):531-535.
Takei M, Nin C, Iizuka T, Pawlikowski M, Selva MA, Chantran Y, Nakajima Y, Zheng J, Aizawa T, Ebisawa M, Sénéchal H, Poncet P. Capsicum Allergy: Involvement of Cap a 7, a New Clinically Relevant Gibberellin-Regulated Protein Cross-Reactive With Cry j 7, the Gibberellin-Regulated Protein From Japanese Cedar Pollen. Allergy Asthma Immunol Res. 2022;14(3):328-338.
Van der Walt A, Lopata AL, Nieuwenhuizen NE, Jeebhay MF. Work-related allergy and asthma in spice mill workers - The impact of processing dried spices on IgE reactivity patterns. Int Arch Allergy Immunol 2010;152(3):271-278.
Wagner S, Breiteneder H. The latex-fruit syndrome. Biochem Soc Trans 2001;30(6):935-940.
Wagner S, Radauer C, Hafner C, Fuchs H, Jensen-Jarolim E, Wuthrich B, Scheiner O, Breiteneder H. Characterization of cross-reactive bell pepper allergens involved in the latex-fruit syndrome. Clin Exp Allergy 2004;34(11):1739-1746.
Williams SR, Clark RF, Dunford JV. Contact dermatitis associated with capsaicin: Hunan hand syndrome. Ann Emerg Med 1995;25(5):713-715.
Zhigila DA, AbdulRahaman AA, Kolawole OS, Oladele FA. Fruit Morphology as Taxonomic Features in Five Varieties of Capsicum annuum L. Solanaceae. Journal of Botany 2014; 4:1-6.