Whole Allergen
f286 Mare's milk
f286 Mare's milk Scientific Information
Type:
Whole Allergen
Display Name:
Mare's milk
Allergen code:
f286
Latin Name:
Equus spp.
Other Names:
Mare's milk
WHO/ICD-11 code:
XM8RU4
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Clinical Relevance
Clinical symptoms in cow's milk allergic patients have been described.
Cow’s milk allergic patients using ointments containing mare’s milk have suffered severe clinical symptoms . Cross reactivity between cow’s milk and mare’s milk has been shown in RAST inhibition. The authors suggest the cross reactivity is linked to sensitization to casein(1). On the other hand, heat labile proteins, presumably a-lactalbumin and b-lactoglobulin, of mare’s milk showed no relation to the corresponding cow’s milk proteins as judged by IgE response, skin tests and clinical sensitivity in a middle-aged woman (2).
Compiled By
Last Reviewed-  April 2022
References
  1. Kalveram, CM; Kalveram, K. Cross reactivity of cow’s milk and mare’s milk antigens. Allergy Clin Immunol News; 1991; Suppl 1: 616.
  2. Gall, H; Kalveram, CM; Sick, H; Sterry, W. Allergy to the heat-labile proteins a-lactalbumin and b-lactoglobulin in mare’s milk. JAllergy Clin Immunol; 1996; 97: 1304-1307.