Type:
Whole Allergen
Whole Allergen
Whole Allergen
Swordfish
f312
Xiphidae
Xiphias gladius
Swordfish
XM95M8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be
Swordfish is an enormous mackerel-like fish representing the only member of the family Xiphidae. It feeds on schools of fishes. The prolongation of the forepart of the skull into a horizontally flattened "sword" is a formidable weapon which can pierce wooden boats. An Atlantic and Mediterranean fish, it is seen only occasionally in the Pacific. A patient with clinical sensitivity to swordfish had IgE antibodies to a 25 kD protein but to no other protein from 10 fish species tested. The allergen may be specific for the family Xiphidae (1).
Last reviewed: June 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.