Whole Allergen
f337 Sole
f337 Sole Scientific Information
Type:
Whole Allergen
Display Name:
Sole
Allergen code:
f337
Family:
Soleidae
Latin Name:
Solea solea
Other Names:
Sole
WHO/ICD-11 code:
XM95M8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Clinical Relevance
The sole, also called Dover sole, black sole, true sole, tounge, etc, is a highly valued fish. The name sometimes refers to all members of the family Soleidae, and also frozen fillets of other cheaper flatfishes, e.g. flounders or dabs from the family Pleuronectidae, may appear on the market falsely labeled as sole. Clinical sensitivity and IgE anti-bodies against sole were studied and discussed in relation to sensitivity to other types of fish (1, 2).      
Compiled By
Last Reviewed : June 2022
References
  1. de Martino, M; Novembre, E; Galli, L; de Marco, A; Botarelli, P; Marano, E; Vierucci, A. Allergy to different fish species in cod-allergic children: In vivo and in vitro studies. J Allergy Clin Immunol; 1990; 86: 909-914.
  2. Pascual, C; Martín Esteban, M; Fernández Crespo, J. Fish allergy: Evaluation of the importance of cross-reactivity. J Pediatr; 1992; 121: S29-34.