Whole Allergen
f5 Rye
f5 Rye Scientific Information
Type:
Whole Allergen
Display Name:
Rye
Allergen code:
f5
Family:
Poaceae (Gramineae)
Species:
Secale cereale
Route of Exposure:
Inhalation
Source Material:
Rye grain/flour
Latin Name:
Secale cereale
Other Names:
Rogge
WHO/ICD-11 code:
XM6JA2
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Rye is one of the unique and the hardiest cereals among all grains. Rye grain (Secale cereale) is different from ryegrass pollen (Lolium perenne), wild rye grass pollen, and cultivated ryegrass. Rye is extensively used in bread-making, alcohol fermentation, paper making, wood industry, used as fuel, and feed for livestock.  Believed to have originated in Afghanistan and Turkish areas, rye is now grown in a cool region and is adapted to various adverse environmental conditions. Nevertheless, rye is also one of the most common sources of allergy, especially in Europe, where bread is extensively consumed.
Inhalation of rye flour may cause respiratory symptoms and elicit an allergic response. Rye flour allergy is reported among people working or associated with bakeries and are exposed to rye flour. Depending on the route of exposure, rye allergy may have different clinical manifestations including classical food allergy symptoms that affect the skin, gastrointestinal system, or respiratory tract; occupational rhinitis or asthma and contact dermatitis; and even exercise-induced anaphylaxis. Allergens from rye grain have been characterized and Sec c 38 is found to be the major allergen, belonging to the family of α- amylase inhibitors.
Allergens from rye show profound cross-reactivity with other cereal grains like wheat, barley, and limited cross-reactivity with oat and even potato extracts. Besides cereals, allergens of rye also exhibit cross-reactivity with allergens from poppy seeds, hazelnut, soya, fungal α-amylases, and with grass and tree pollens.
Sensitized individuals are advised to avoid rye flour in diet and strict labeling laws to label gluten-containing cereals have been established in various countries. 
Allergen
Nature
Rye (Secale cereale) is considered a unique grain from graminoid cereals. It has historical significance. Rye is cross-pollinated by wind (1). It is one of the toughest cereal crops that seeds from late summer to mid-autumn. It is sensitive to seeding depth compared to other cereals and grows up to 2 inches deep (2). Rye is a small grain, and its growth is the best among all the winter grains (3).
Gluten-containing cereals like rye, wheat, barley are one of the common allergenic foods (4). Rye flour is the source of this food allergy, which depending on the route of exposure, can present different clinical manifestations (5).
Like other cereal allergens, the allergens in rye exhibit inherent resistance to gastrointestinal enzymes (such as pepsin, trypsin and pancreatin), thereby eliciting the allergic response on reaching the intestine (5).
Habitat
Rye grows best in cool and temperate zones, although it is well adapted in different climatic zones (2).
Taxonomy
Taxonomic tree of Rye (6)  
Domain Eukaryota
Kingdom Viridiplantae
Phylum Streptophyta
Class Magnoliopsida
Order Poales
Family Poaceae
Tribe Triticeae
Genus Secale
Species Secale cereale
Tissue
The main chemical constituents of rye grain are starch, dietary fiber, mineral matter, and protein (7). Prolamins are the major storage proteins in cereal grains (8). Secalins, the allergenic molecules in rye, are alcohol-soluble seed storage proteins (prolamins) found in the endosperm of rye grain (8-10).
Epidemiology
Worldwide distribution
Baur et al. (1998) studied the frequency of work-related asthma in 89 bakers from the Bochum area (Group A). They summarized the data from a previous study involving 104 bakers (Group B) filing compensation claims for the baker’s asthma. The study involved a control group with 43 healthy people of the same area who did not work in the bakeries. They found that 34% of the subjects from Group A, 50% from Group B, and only 10% of the Control group were positive for rye flour in IgE determination tests (11). A study on 24 Spanish employees (92% males) from the baking industry with a history of baker’s asthma showed that 71% of the workers were positive for rye flour in skin prick tests (SPTs), and 67% showed positive IgE to rye flour (12).
A cross-sectional study was performed across 31 bakeries involving 517 workers across supermarket stores in South Africa to study the asthma phenotypes in these workers exposed to flour dust in the bakery. It was found that about 33% of workers documented work-related asthma due to rye flour specifically. However, these workers handled only a small proportion of products made up of rye flour (13).
A study from a Korean allergy clinical center involving 5340 patients with different allergic diseases demonstrated the IgE sensitization rate and cross-reactivity to different home-made agricultural products and observed that rye grain sensitization rate was 9.5% (14).
Allergy to rye has also been reported in Japan and other western countries (15).
Risk factors
Bakers are at a very high risk of developing occupational asthma from exposure to rye flour used in making bread and other bakery products (16). A study also showed that woodworkers with occupational allergy represent a risk group to rye flour allergy since rye flour is used as a glue in the woodwork (17).
Pediatric issues
Out of different cereal allergens known to cause allergy in children, a study on 40 newborns aged between 3 to 6 months detected rye as one of the potential sources of allergens. Symptoms such as diarrhea, vomiting, eczema, and weight loss were elicited on feeding the babies with cereal formula (18).
Environmental Characteristics
Living environment
Rye grain shows excellent growth in poor soils (7). Rye can be cultivated in a variety of adverse environmental conditions (1). Rye can be grown on infertile, acidic, sandy, or poorly prepared lands as well (2).
Worldwide distribution
Rye is considered to have originated in Afghanistan and Turkish regions. The primary domestication of rye is seen in Turkey, Iran, Central Asia and Transcaucasia. It was introduced into Europe and can be found along the Mediterranean route. It is a stable and popular crop in Central Europe, Northern Europe, and Russia. Related wild rye is grown in Southwest Asia, the Mediterranean basin, Central Asia, and Caucasus (1).
Route of Exposure
Main
Inhalation of rye flour can cause respiratory symptoms and elicit an allergic response (19).
Secondary
Ingestion or even skin contact with cereal rye can cause allergic symptoms, contact dermatitis, exercise-induced anaphylaxis, or gut-related symptoms (5, 20).
Clinical Relevance
Allergic rhinitis and rhino-conjunctivitis (ARC)
Depending on the route of exposure, rye and wheat allergy may have different clinical manifestations including classical food allergy symptoms that affect the skin, gastrointestinal system, or respiratory tract; occupational rhinitis or asthma and contact dermatitis; and even exercise-induced anaphylaxis (5). Moreover, allergens in rye flour, similar to other cereals are resistant to gastrointestinal enzymes and can provoke a reaction upon ingestion (5).
A 38-year-old male baker presented with a two-month history of rhinitis, asthma, and urticaria on arms by exposure to rye flour. The patient showed heightened sensitivity to rye than to other cereals. Rye has been studied to elicit a more robust physiological bronchial response than other grains like wheat (21).
A 35-year-old man working in a bakery for seven years reported aggravation of allergic rhino-conjunctivitis (ARC) upon exposure to rye flour. A 32-year baker from Cuba complained of ARC on rye flour exposure in the bakery where he worked for three years (22).
A 32- year old male baker with a history of smoking and ARC in Japan presented aggravation of ARC, chest tightness, cough, and wheezing while handling rye flour (23).
In a study on rye allergy to woodworkers (since rye four is used in the wood industry), all nine subjects exposed to wood dust and having a history of occupational allergy showed a positive response to commercial rye flour extracts in skin prick and conjunctival tests (17).
Asthma
Inhalation of flour from rye, wheat, barley, soya bean, and buckwheat are reported to trigger respiratory allergic symptoms and asthma and among them, rye, wheat, or barley are mostly known to be responsible for these allergies (19)Allergens in rye flour are known to cause occupational asthma, particularly Baker’s asthma since rye flour is popularly being used to produce “special bread” (5, 16).
A 38-year old baker exhibited asthma and other respiratory symptoms on exposure to rye flour or mixes of rye flour only, and not any other cereal (21).
An old, asymptomatic baker who shifted to working from a bakery in Cuba to Spain complained of developing respiratory symptoms, including asthma, which aggravated with rye flour exposure. He reported severe and more frequent asthma attacks requiring immediate emergency hospital admission due to rye four allergy (22).
Food allergy syndrome and Anaphylaxis
Ingestion of rye can trigger food allergy symptoms in the gut and skin and may cause anaphylaxis (5). Anaphylaxis following the ingestion of rye has been documented. A case of a 61- year old woman without any history of drug allergy, suffering from rye-dependent exercise-induced anaphylaxis upon ingestion of toasted rye bread, has been reported. The woman developed generalized erythema, dizziness, hypotension,and facial angioedema after a 30-minute walk, one-hour post-ingestion of rye (24). A 32-year old Japanese baker with a history of ARC presented with anaphylactic symptoms including urticaria in the upper body, facial swelling, dyspnea, and angioedema on eyelids after consumption of bread containing 50% rye flour (23). Similarly, another study demonstrated a case of a 38-year old male baker with a history of urticaria on arms after exposure to rye flour (21).
Contact dermatitis
Flour-associated contact dermatitis is frequently reported in sensitized individuals. The most commonly responsible agents are wheat and rye. A woman with a history of atopic eczema on fingers since childhood reportedly developed itchy rashes on her hand upon handling a dough made of both wheat and rye and wet chapatis. Dry flour did not trigger any symptoms. Also, the patient did not develop symptoms of allergy or AR after eating cooked chapatis. She tested positive in SPTs for both wheat and rye individually and together (25).
Prevention and Therapy
Prevention strategies
Avoidance
Dietary elimination of allergic food has been shown to improve the status of symptoms due to food-borne allergies. In most countries, it is mandatory to have proper labeling for all gluten-containing grains, including rye (9).
Other topics
The potential of a selected cohort of sourdough lactic acid bacteria in hydrolyzing the allergens in wheat and rye cereal proteins during fermentation, followed by treatment with digestive enzymes has been evaluated. Serum studies from sensitized individuals showed that during the treatment of sourdough wheat and rye bread, the potential IgE- binding proteins were degraded. The decrease in the allergenicity potential of treated rye and wheat bread was ascribed to the selected sourdough lactic acid bacteria's proteolytic activity in producing pre-digested flours with IgE-binding proteins, no longer resistant to digestive enzymes (5).
Molecular Aspects
Allergenic molecules
According to the WHO/IUIS nomenclature, the allergens in rye include:
  1.  Sec c 38 (renamed from Sec c 1): a 13.5 kDa, dimeric α-amylase/trypsin inhibitors / α-amylase inhibitor  (20, 26).
  2. Sec c 5: a 30 kDa, group 5 grass pollen allergen (20).
  3. Sec c 20: a 70 kDa, gamma-secalin (20).
Sec c 38 is the major allergen from rye flour with a positive SPT in 71% of patients with baker’s asthma (26).
91% of patients reporting wheat-dependent exercise-induced anaphylaxis exhibited IgE antibodies to Sec c 20. Similarly, 67% of patients showed Sec c 20 in their sera as per the STPs (20).
Other allergens with limited characterization include RDAI-1 and RAI-3 which elicited an allergic response in 86% and 57% of patients in a study. These allergens are members of α- amylase/ trypsin inhibitor family (27).
Cross-reactivity
An extensive cross-reactivity is observed between rye flour, wheat, and barley flour. This cross-reactivity is attributed to the family of proteins gliadins and glutenins (28). Sec c 38, a key allergen from rye, and Barley dimeric protein (BDP) show a 70% identical N-terminal sequence (Garcia-Casado1995).
A study demonstrated that  γ-70 and γ-35 secalins of rye cross-react with ῳ-5 gliadin, a major allergen in wheat-dependent exercise-induced anaphylaxis, indicating that rye can induce symptoms in patients with wheat-dependent, exercise-induced anaphylaxis (29).
Fungal α-amylase is used as a dough additive in numerous bakeries and is another potential cause of baker’s asthma. A study assessing the case reports of two different bakers with an allergy to wheat, barley, and rye showed that both the cases were also sensitive to fungal α- amylase, thereby implying that some, if not broad, cross-reactivity exists between cereal amylases and fungal amylases (22).
Rye grain also shows weak cross-reactivity with oat allergens (30).
Inhibition tests in potato sensitized subjects have demonstrated that rye grain, wheat flour, soya bean, Hop. J and corn extracts inhibited IgE binding to extracts from potato (14).
Allergens in sesame show cross-reactivity with rye, although this cross-reactivity's clinical significance has still not been established (31). Cross-reactivity of rye is seen with allergens in poppy seed, hazelnut, and soya (19, 32, 33).
Merget et al. (2011) demonstrated a significant cross-reactivity between rye flour and grass and tree pollens. Study subjects positive to challenge tests for rye flour also exhibited the highest concentration to ryegrass IgE and IgE to tree pollen (34).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: November  2020
References
  1. Hagenblad J. OHR, Forsberg N.E.G. et al. . Geographical distribution of genetic diversity in Secale landrace and wild accessions. BMC Plant Biol 2016;16(23).
  2. SARE. Managing cover crops profitably. 3rd edition ed. USA: SARE; 2012.
  3. Knott C. HE, Henning J., Lee c., Smith R. Grain, Forage, and Cover crop guide for Kentuck 2020.
  4. Boye JI. Food allergies in developing and emerging economies: need for comprehensive data on prevalence rates. Clinical and Translational Allergy 2012;25(2):1-9.
  5. Rizzello C.G. DAM, Coda R. et al. Use of selected sourdough lactic acid bacteria to hydrolyze wheat and rye proteins responsible for cereal allergy. . Eur Food Res Technol. 2006;223:405–11.
  6. NCBI. Secale cereale: National Centre for Biotechnology Information (NCBI). Available from: https://www.ncbi.nlm.nih.gov/Taxonomy/Browser/wwwtax.cgi?mode=Info&id=4550&lvl=3&p=has_linkout&p=blast_url&p=genome_blast&lin=f&keep=1&srchmode=1&unlock. Accessed on 17.10.2020.
  7. Hansen H.B. MB, Andersen S.B., Jørgensen J.R., Hansen A. . Grain characteristics, chemical composition, and functional properties of rye (Secale cereale L.) as influenced by genotype and harvest year. J Agric Food Chem  2004;52(8):2282-91.
  8. Breiteneder H. RC. A classification of plant food allergens. J Allergy Clin Immunol 2004;113(5):821-30.
  9. Kliewer K. VC, Cassin A.M., Abonia J. . Should wheat, barley, rye, and/or gluten be avoided in a 6-food elimination diet? The Journal of Allergy and Clinical Immunology 2015;137(4).
  10. Shimizu Y. NS, Endo T.R. Detection of the Sec-1 locus of rye by a PCR-based method. Genes Genet Syst. 1997;72(4):197-203.
  11. Baur X DP, Sander I Baker’s asthma: Still among the most frequent occupational respiratory disorders. J Allegy Clin Immunol. 1998;102:984–97.
  12. Quirce S. F-NM, Escudero C., Cuesta J., de Las Heras M., Sastre J. . Bronchial responsiveness to bakery-derived allergens is strongly dependent on specific skin sensitivity. Allergy        2006;61(10):1202-8.
  13. Baatjies R LA, Sander I et al. . Determinants of asthma phenotypes in supermarket bakery workers. . EurRespir J. 2009;34:825–33.
  14. Yoon S.H. KYM, Kim S.H., Suh C.H., Nahm D.H., Park H.S. . The sensitization rate and cross-reactivity to homemade agrivultural products in adult allergy patients. KoreanJ Asthma Allergy Clin Immunol. 2005;25(4):269-75.
  15. Gangal S.V. MBK. Food allergy-how much of a problem really is this in India? Journal of Scientific and Industrial Research 2003;62:755-65.
  16. Bensefa L. VC, Tabka F., Causse-Sounillac E., Fabries J.F., Choudat D. . Rye flour induces a stronger early bronchial response than wheat flour in occupational asthma. . Allergy 2004;59: 833-8.
  17. Armentia A. GCG, Vega J., Sanchez-Monge R., Mendez J., Salcedo G. . Occupational allergy to rye flour un carpenters. Allergy. 1997;52:1151-2.
  18. Armentia A. RR, Callejo A., Martin-Esteban M., Martin-Santos J.M., Salcedo G. et. al. . Allergy after ingestion or inhalation of cereals involves similar allergens in different ages. . Clin Exp Allergy. 2002;32:1216-22.
  19. Sandiford C.P. TRD, Newman-Taylor A.J. Identification of crossreacting wheat, rye, barley and soya flour allergens using sera from individuals with wheat-induced asthma. Clin Exp Allergy 1995;25(4):340-9.
  20. WHO/IUIS. Allergen Nomenclature Sub-Committee 2019. Available from: http://www.allergen.org/search.php?allergenname=&allergensource=Secale+cereale&TaxSource=&TaxOrder=&foodallerg=all&bioname= Accessed on 17.10.2020.
  21. Ehrlich R. PR. Baker’s asthma with a predominant clinical response to rye flour. American Journal of Industrial Medicine 2005;48:153-5.
  22. Letrán A. PA, Barranco P., Salcedo G., Pascual C., Quirce S. . Rye flour allergens: an emerging role in baker's asthma. Am J Ind Med 2008;51(5):324-8.
  23. Oshikata C, Tsurikisawa, N., Saito, A., Yasueda, H., & Akiyama, K. Occupational asthma from exposure to rye flour in a Japanese baker. Respirology Case Reports. 2014;2(3):102-4.
  24. García-Menaya JM CG, Cordobés-Durán C, Mahecha AC, Bobadilla-González P. . Rye-dependent exercise-induced anaphylaxis. Ann Allergy Asthma Immunol 2016;117(5):566-8.
  25. Malinauskiene L. IM. Protein contact dermatitis caused by allergy to chapatti flour. Acta Derm Venereol 2013;93(1):91-3.
  26. Garcia-Casado G. AA, Sanchez- Monge R., Sanchez L.M., Lopez-Otin G., Salcedo G. . A major baker’s asthma allergen from rye flour is considerably more active than its barley counterpart. FEBS 1995:36-40.
  27. García-Casado G AA, Sánchez-Monge R, Malpica JM, Salcedo G. Rye flour allergens associated with baker's asthma. Correlation between in vivo and in vitro activities and comparison with their wheat and barley homologues. Clin Exp Allergy 1996 26(4): 428-35.
  28. Snégaroff J. B-MI, Pecquet C., Branlard G., Laurière M. . Study of IgE antigenic relationships in hypersensitivity to hydrolyzed wheat proteins and wheat-dependent exercise-induced anaphylaxis. Int Arch Allergy Immunol 2006;139(3):201-8.
  29. Palosuo K. AH, Varjonen E., Kalkkinen N., Reunala T. Rye gamma-70 and gamma-35 secalins and barley gamma-3 hordein cross-react with omega-5 gliadin, a major allergen in wheat-dependent, exercise-induced anaphylaxis. Clin Exp Allergy 2001;31(3):466-73.
  30. Gilissen L.J.W.J. Meer vdMIM, Marinus J.M.M. Reducing the incidence of allergy and intolerance to cereals. Journal of Cereal Science. 2014;59:337-53.
  31. Dalal I. GM, Katz Y. . Sesame seed food allergy. Curr Allergy Asthma Rep  2012;12(4):339-45.
  32. Oppel T. TP, Pryzbilla B., Wollenberg A. . Cross-reactivity from poppy seed to buckwheat in a food allergic patient with poppy seed anaphylaxis.113(2): 244. Journal of Allergy and Clinical Immunology. 2004;113(2).
  33. Müller U. LD, Hoffmann A. et al. . Allergens in raw and roasted hazelnuts (Corylus avellana) and their cross-reactivity to pollen.” 2000; 212: . Eur Food Res Technol 2000;212:2–12.
  34. Merget R. SI, van Kampen V., Beckmann U., Heinze E., Raulf-Heimsoth M., Bruening T. Allergic asthma after flour inhalation in subjects without occupational exposure to flours: an experimental pilot study. Int Arch Occup Environ Health. 2011;84(7):753-60.