Whole Allergen
f59 Octopus
f59 Octopus Scientific Information
Type:
Whole Allergen
Display Name:
Octopus
Allergen code:
f59
Family:
Octopodidae
Latin Name:
Octopus vulgaris
Other Names:
Octopus
WHO/ICD-11 code:
XM66P3
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Clinical Relevance
IgE antibodies have been measured in sera from Japanese patients with atopic dermatitis (1).
Octopus vulgaris is a cephalopod mollusk of the family Octopodidae, an octopus with eight suckered arms and a round mouth. IgE antibodies have been measured in sera from Japanese patients with atopic dermatitis (1). Also measured in children with allergy (2). Sensitivity to octopus is also common in southern Europe (3). In a study using Pharmacia CAP System, octopus and squid showed a low degree of correlation with each other (4).
Molecular Aspects
Cross-reactivity
Octopus and squid showed a low degree of correlation with each other (4). IgE antibodies to octopus were also measured in children with allergy (2).
References
  1. Lindqvist A IZ, Tanaka A, Yman L. Seafood specific IgE in atopic dermatitis. Ann Allergy Asthma Immunol. 1993:70:58.
  2. Yamada M TS. Clinical evaluation of Pharmacia CAP System new food and inhalant allergens. Japanese Soc Allergol 1992.
  3. Castillo R, Carrilo T, Blanco C, Quiralte J, Cuevas M. Shellfish hypersensitivity: clinical and immunological characteristics. Allergologia et immunopathologia. 1994;22(2):83-7.
  4. al CTe. Clinical evaluation of Pharmacia CAP System new food allergens. Jap Soc Pediatric Allergol. 1992