Type:
Whole Allergen
Whole Allergen
Whole Allergen
Jack mackerel/Scad
f60
Carangidae
Trachurus japonicus
Jack mackerel/Scad
XM95M8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
IgE antibodies to Jack mackerel have been measured in asthmatic adults (1), atopic dermatitis patients (2), and food-allergic children (3).
This fish is not a mackerel but a jack. It is also called Japanese scad. There are several species of Trachurus, some also called horse mackerel. Other names used are saurel, cigar fish and round robin. The name horse mackerel is also sometimes applied to certain species of tuna and bonito
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fish (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small.
Last Reviewed- April 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.