Type:
Whole Allergen
Whole Allergen
Whole Allergen
Wild rye grass
g70
Poaceae (Gramineae)
Elymus triticoides
Leymus triticoides, Elymus condensatus
This grass is native to and widespread in western North America, from Washington State to California, and east to Montana. It often forms large clumps in the wild, but is also grown for forage and ground cover, especially for erosion control in low- and medium-elevation saline and alkali areas. In Europe the closely-related species Lymegrass (E. arenarius) is common in the dunes along coasts.
Wild Rye grass looks like a cross between Bermuda grass and Salt grass. It is a cool-season perennial, forming extensive creeping rhizomes. The stems are upright, hollow, usually unbranched, and up to 1.2m tall. Blades are up to 10cm long, about 1cm wide, flat to involute, at least somewhat stiff, and rough to the touch. The inflorescences are spikes up 25cm long, mostly 2 per node, with spikelets containing 3 to 8 flowers each.
It flowers between June and August, and the seeds ripen from September to October. The flowers are hermaphrodite (have both male and female organs) and are pollinated by wind. Seed germination (requiring cold) is poor. Seedlings are weak and compete poorly with weeds and other grasses in the early developmental stages. However, once established, the grass is very rhizomatous and maintains stands for many years.
This grass is unlike most others in the genus Elymus because of its very slender and short spikes, its long-creeping rhizomes, and its paired spikelets.
Meadows, damp ravines, river flats, sand dunes, often in salty areas; erosion-prone slopes; cultivated beds.
The seed may be ground into flour and used to make to make bread, cakes, or porridge. (The hairs on the seed must be removed, traditionally by singeing, before consumption.)
No allergens from this plant have yet been characterised.
The pantemperate tribe Triticeae is notable for its cereal genera: Wheat, Barley and Rye. The close relation speaks in favour of cross-reactivity between Wild Rye grass g70, Cultivated Wheat g15, Cultivated Rye grass g12, Barley g201, and Couch grass (Agropyron repens), as well as Lymegrass (Elymus arenarius). Extensive cross-reactivity among the different individual species of the genus may be expected. There should be cross-reactivity with other members of the family Poaceae, particularly in the subfamily Pooideae (Rye grass (g5), Canary grass (g71), Meadow grass (g8), Timothy (g6), Cocksfoot (g3), Meadow Fescue (g4), Velvet (g13), Redtop (g9), Meadow Foxtail (g16), Wild Rye grass (g70)) (1, 2)
Anecdotal evidence suggests that Wild Rye grass pollen may induce asthma, allergic rhinitis and allergic conjunctivitis in sensitised individuals; however, no studies have been reported to date.
In Missouri, USA, the close relative E. canadensis (Canada wildrye or Canada wild rye) has been implicated as a minor contributor in hay fever, and other genus members have been suspected elsewhere, particularly in California (3).