Whole Allergen
g71 Canary grass
g71 Canary grass Scientific Information
Type:
Whole Allergen
Display Name:
Canary grass
Allergen code:
g71
Family:
Poaceae
Species:
Phalaris arundinacea
Route of Exposure:
Inhalation
Latin Name:
Phalaris arundinacea
Other Names:
Reed Canary grass, Reed Canarygrass, Ribbon grass,
Summary
Phalaris arundinacea (canary grass) is a widely distributed perennial wetland reed with highly-mobile pollen that can induce hypersensitive respiratory and cutaneous reactions in sensitized individuals. 
Allergen
Nature
Pollen produced by P. arundinacea can induce hypersensitive respiratory (asthma and rhinoconjunctivitis) and cutaneous reactions in sensitized individuals (2-4).
Taxonomy 
Taxonomic tree of P. arundinacea (canary grass) (1)
Domain
Eukaryota
Kingdom
Plantae
Phylum
Spermatophyta
Subphylum
Angiospermae
Class
Monocotyledonae
Family
Poaceae
Genus
Phalaris
Environmental Characteristics
Worldwide distribution 
P. arundinacea is a perennial wetland reed which is widely distributed throughout North and Central America, most of Africa, Europe, the Middle East and Asia, including Pacific islands (1).
Considered an invasive species, canary grass grows 0.6–2 m high and can spread rapidly along watercourses and ditch systems (1). The invasive success of canary grass may be related to its highly plastic phenotypic responses to environmental stresses (5, 6), and a number of studies have demonstrated a beneficial ecological role for P. arundinacea in phytoremediation of contaminated soils (7, 8), denitrification (9) and greywater reuse (10).
Route of Exposure
Main 
Large quantities of highly mobile seed are produced in the first year of life, and canary grass also spreads vegetatively via vigorous and dense rhizome growth (1, 3, 6). Direct exposure of skin to grass has been reported to elicit hypersensitivity reactions including itchy urticaria and angioedema (3).
Clinical Relevance
A retrospective cross-sectional study of 894 patients with seasonal allergic rhinitis in Argentina assessed skin prick tests (SPTs) to selected pollen species belonging to three subfamilies of the Poaceae family, which between them account for 95% of allergy-relevant grass species (2). In this study, patients were most frequently sensitized to extracts from the Pooideae subfamily (86.8%; CI 84.4%–88.9%), and P. arundinacea (subfamily Pooideae) presented the highest frequency of sensitization of all species tested overall (62.2%; CI 59.0%–65.3%) (2).
Ramon et al. (2017) described a 4-year old male child who developed acute urticaria and mild neurologic/respiratory symptoms after exposure to grasses including P. arundinacea (3). On two occasions, the child laid down on grass and within several minutes experienced generalized itching, angioedema, erythema and respiratory symptoms which resolved following treatment with glucocorticoids and/or antihistamines (3). Notably, SPTs for this child were negative for a broad range of foods and airborne allergens but significantly positive for three grasses including P. arundinacea (3).
Luis et al. (2017) described two case reports of patients with a personal history of seasonal allergic rhinoconjunctivitis and asthma to pollen, who developed perennial symptoms after keeping parakeet birds at home (4). SPTs in both patients were positive to a range of seed and grass substances including canary grass, and the authors concluded that asthma and rhinoconjunctivitis after inhalation of bird food were rare but should be considered as a potential cause of respiratory allergy in bird-keepers (4).
Molecular Aspects
Allergenic molecules
To date, no allergens have been characterized from P. arundinacea specifically.  However, several protein allergens have been isolated from the pollen of P. aquatica, a closely-related bulbous canary grass, which suggests these allergens may be present in P. arundinacea (11, 12):
·       Pha a 1, a 34 kDa protein and major Group 1 allergen
·       Pha a 5, a major Group 5 allergen with four isoforms
Cross-reactivity
Variable degrees of cross-reactivity may occur between grass pollen species sharing Group 1 and/or Group 5 grass allergens (13). Radioallergosorbent testing (RAST) of sera from subjects sensitized to inhaled wheat or flour indicated that there was significant reaction with seed extracts of 12 different cereals, including canary grass (14).
Carbohydrate-reactive IgE antibodies attributed to grass pollen sensitization have been found to cross-react with glycan structures from other allergen sources, particularly vegetable foods (15). Extensive cross-reactivity is also possible between profilins, a family of Group 12 allergen proteins which are highly conserved throughout the plant kingdom and present in all tissues (15).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer
Last reviewed: December  2021
References
  1. CABI. Phalaris arundinacea (reed canary grass) Wallingford, UK: CAB International 2021 [cited 2021 5.11.21]. Available from: https://www.cabi.org/isc/datasheet/55423.
  2. Ramon GD, Barrionuevo LB, Viego V, Vanegas E, Felix M, Cherrez-Ojeda I. Sensitization to subtropical grass pollens in patients with seasonal allergic rhinitis from Bahia Blanca, Argentina. World Allergy Organ J. 2019;12(9):100062.
  3. Ramon GD, Croce V, Chérrez-Ojeda I. Anaphylaxis in a 4-year-old male caused by contact with grasses: A case report. World Allergy Organization Journal. 2017;10.
  4. Luis EM, Matas AE, Moguel IG, Herrera LH, De las Cuevas Moreno N, Fernández CR. RESPIRATORY ALLERGY DUE TO MILLET IN BIRD-KEEPERS: TWO CASES REPORT. Journal of Allergy and Clinical Immunology. 2017;139(2):AB6.
  5. Weston LM, Mattingly KZ, Day CTC, Hovick SM. Potential local adaptation in populations of invasive reed canary grass (Phalaris arundinacea) across an urbanization gradient. Ecology and Evolution. 2021;11(16):11457-76.
  6. Lavergne S, Molofsky J. Increased genetic variation and evolutionary potential drive the success of an invasive grass. Proceedings of the National Academy of Sciences. 2007;104(10):3883-8.
  7. Novakovskiy AB, Kanev VA, Markarova MY. Long-term dynamics of plant communities after biological remediation of oil-contaminated soils in far north. Scientific Reports. 2021;11(1):4888.
  8. Polechońska L, Klink A. Accumulation and distribution of macroelements in the organs of Phalaris arundinacea L.: Implication for phytoremediation. J Environ Sci Health A Tox Hazard Subst Environ Eng. 2014;49(12):1385-91.
  9. Gordon BA, Lenhart C, LaPara TM. A comparison of nitrate removal and denitrifying bacteria populations among three wetland plant communities. J Environ Qual. 2020;49(1):210-9.
  10. Chowdhury RK. Greywater reuse through a bioretention system prototype in the arid region. Water Sci Technol. 2015;72(12):2201-11.
  11. Suphioglu C, Singh MB. Cloning, sequencing and expression in Escherichia coli of Pha a 1 and four isoforms of Pha a 5, the major allergens of canary grass pollen. Clin Exp Allergy. 1995;25(9):853-65.
  12. Suphioglu C, Singh MB, Simpson RJ, Ward LD, Knox RB. Identification of canary grass (Phalaris aquatica) pollen allergens by immunoblotting: IgE and IgG antibody-binding studies. Allergy. 1993;48(4):273-81.
  13. Matthiesen F, Løwenstein H. Group V allergens in grass pollens. II. Investigation of group V allergens in pollens from 10 grasses. Clin Exp Allergy. 1991;21(3):309-20.
  14. Baldo BA, Krilis S, Wrigley CW. Hypersensitivity to inhaled flour allergens. Comparison between cereals. Allergy. 1980;35(1):45-56.
  15. Andersson K, Lidholm J. Characteristics and immunobiology of grass pollen allergens. Int Arch Allergy Immunol. 2003;130(2):87-107.