Whole Allergen
i202 Grain weevil
i202 Grain weevil Scientific Information
Type:
Whole Allergen
Display Name:
Grain weevil
Allergen code:
i202
Family:
Curculionidae
Latin Name:
Sitophilus granarius
Summary
Insect
An insect, which may result in allergy symptoms in sensitised individuals.
Exposure
Allergen Exposure
Sitophilus granarius is a common pest in stored grain, and a common kitchen pest.
The grain weevil is a member of the family of beetles known as Curculionidae, or true weevils. It is tiny, and chestnut brown or black. It has an elongated snout on the front of the head, at the tip of which are the mouth parts. Grain weevils are associated with all kinds of whole grain. Besides the grain weevil, there are two other almost indistinguishable species of weevil that are important pests of grain: the rice weevil (S. oryzae) and the maize weevil (S. zeamais). (1)
Females produce numerous eggs six times a year, and deposit them inside grain kernels.
The allergy-causing components persist after processing (cleaning and milling) of the grain. The principal exposure occurs while bakers are exposed to the grain weevil allergens, together with other dust and flour allergen, but also during milling procedures.
Potential Cross Reactivity
A German study has shown that another member of the genus, S. oryzae, the rice weevil, caused IgE-mediated allergy among people living in a former grain store. This study found several binding components in rice weevil, ranging from 30 to 70 kDa, and also observed cross-reactivity to the grain weevil. (2, 5, 6)
Clinical Experience
a. IgE-mediated reactions
Although grain weevil can be found in homes, it has been reported primarily as an occupational allergen, (2, 4, 7 )and one of the causes of ‘Millworker’s asthma’, (1, 8) ‘grain-worker's asthma’, (9) and ‘Farmer’s lung’. (7) Hypersensitivity pneumonitis has also been described. (10)
Two laboratory workers, who spent a considerable time each day handling grain infested by grain weevil, developed allergic rhinitis and asthma; one after exposure to weevil dust for only six months, and the other after three years. The former worker also noticed sneezing when she was cooking using flour. One of the workers was so sensitive to weevil dust that she had to leave the laboratory; the other worker, by using an extractor fan and wearing a mask, could remain free of symptoms. Skin-prick testing with extracts of grain weevil faecal dust was positive. Consequently, 100 patients attending an allergy clinic for seasonal hay fever symptoms were also skin-prick tested with extracts of S. granaries, of which 51 were positive. In 18, the wheal size was at least 6 mm in diameter. The authors concluded that the significance of the positive responses in these 18 patients was unexplained. (1, 8)
In view of these two patients, a pilot study was undertaken on 75 volunteer millworkers and controls to determine the extent of sensitivity to grain weevil. A millworker was defined as anyone who worked in a flour mill or mill producing animal feed from mixed cereals, or who worked in grain-storing silos. Skin-testing with weevil and mixed flour extracts found that 57% had a positive response to the weevil extract, and 68% a positive response to the mixed flour extract. In the control group, 34% were positive to the weevil extract and 17% to mixed flour. Of the initial 75 millworkers, 15 were selected for further study, based on a positive skin response to the weevil, and a history of a productive cough and chest tightness and wheezing when exposed to mill dust. The forced expiratory volume in one second (FEV1) showed significant reductions of 20% and 15% in two subjects following inhalation of weevil extract. One worker developed wheezing and cough. Twenty-five of the control subjects with positive skin responses to the grain weevil demonstrated no significant FEV1 changes to a similar provocation inhalation test. (8)
In a more recent South African study that examined mill employees in the maize wet-milling industry to determine sensitisation to common allergens and to maize products, of 75 patients tested for specific IgE, 18 were found to have specific IgE to storage mite and 13 (20%) to storage mite and grain weevil. (11)
A study of 66 Danish bakers whose skin-prick tests were positive to extracts of S. granarius reported that 54% had elevated levels of IgE against whole-body extracts of S. granarius. A range of allergens were detected, and a heterogenous response found with a range of sensitisation to the various allergens. (2) Whether the bakers were symptomatic following exposure to this organism was not reported and the authors cautioned that “as for baker's allergy, it is evident that many other factors in the baker's environment besides grain weevil may cause allergy, including flour components (e.g. wheat or other grain proteins), baking-improving additives (e.g. fungal enzymes), and bakery-infesting pests (mites and other insects).
An earlier study of 440 Swedish farmers evaluating the Phadiatop® reported that 29 were RAST-positive to G. intestinalis, and six to S. granarius. The calculated overall prevalence of a positive RAST to G. inteslinalis and S. granarius in the whole farming population (n= 2578) of Gotland was therefore 5.2% and 1.1% respectively. (12)
b. Other reactions
Nil reported.
References
  1. Frankland AW, Kunn JA. Asthma caused by the grain weevil. Br J Ind Med 1965;22:157-159.
  2. Herling C, Svendsen UG, Schou C. Identification of important allergenic proteins in extracts of the granary weevil (Sitophilus granarius). Allergy 1995;50(5):441-6.
  3. Berzhets VM, Radikova OV, Khlgatian SV, Berzhets AI, Kropotova IS. Evaluation of specific activity of preparations of allergens from synanthropic insects. Zh Mikrobiol Epidemiol Immunobiol 2006;(7):74-78.
  4. Sauvaget J, Aerts J, Gacouin JC. Two cases of respiratory disease with grain weevil precipitins; Sitophilus granarius. Rev Fr Allergol Immunol Clin 1978;18(1):1-4.
  5. Gupta S, Jain S, Chaudhry S, Agarwal MK. Role of insects as inhalant allergens in bronchial asthma with special reference to the clinical characteristics of patients. Clin Exp Allergy 1990;20(5):519-24.
  6. Kleine-Tebbe J, Jeep S, Josties C, Meysel U, O'Connor A, Kunkel G. IgE-mediated inhalant allergy in inhabitants of a building infested by the rice weevil (Sitophilus oryzae). Ann Allergy 1992;69(6):497-504.
  7. Ebner H, Feldner H, Kraft D. Some observations on farmer's lung (author's transl). [German] Wien Klin Wochenschr 1981 Dec 11;93(23):729-33.
  8. Lunn JA. Millworkers' asthma: allergic responses to the grain weevil (Sitophilus granarius). Br J Ind Med 1966 Apr;23(2):149-52.
  9. Southcott RV. Injuries from Coleoptera. Med J Aust 1989;151(11-12):654-9.
  10. Bielory L. Hypersensitivity pneumonitis: occupational exposure to Sitophilus granarius and Thermoactinomyces vulgaris. Md State Med J 1982 Dec;31(12):25-6.
  11. Bartie C, Calverley AE, Rees D. Sensitisation to maize in the wetmilling industry. Curr Allergy Clin Immunol 2006;19(3):138-41.
  12. Van Hage-Hamsten M, Johansson SG. In vitro diagnosis of farmers' IgE-mediated allergy by Phadiatop and three new multiallergen RAST analyses. Allergy 1993 Jul;48(5):345-8.