Whole Allergen
k74 Silk
k74 Silk Scientific Information
Type:
Whole Allergen
Display Name:
Silk
Allergen code:
k74
Latin Name:
Bombyx mori
Other Names:
Silk
Route of Exposure
Occupational
Cultured silk from Bombyx mori, is used for exclusive textiles. Workers in the silk industry are heavily exposed. In addition to pure silk they are exposed to silk waste containing potent contaminating allergens such as insect material(1).
Clinical Relevance
IgE-mediated reactions
Allergic respiratory symptoms are well known among silk industry workers and have been attributed to proteins from the silk worm(2). The substance glueing together the thread filaments, sericin, was previously suspected but this hypothesis has not been confirmed (1). Dust containing pure silk is not a potent allergen.
Compiled By
Last Reviewed-  April 2022
References
  1. Johansson SGO, Wüthrich B, Zortea-Caflisch C: Nightly asthma caused by allergens in silkfilled bed quilts: Clinical and immunological studies. J Allergy Clin Immunol 1985, 75, 452-459.
  2. Kobayashi S: Occupational asthma due to inhalation of pharmacological dusts and other chemical agents with some reference to other occupational asthmas in Japan. In Allergology Proceedings VIII Int Congr Allergol eds. Y Yamamura et al., Exerpta Medica, Amsterdam 1973, 127-132.