Table of Contents
Component
k87 Asp o 21, alpha-amylase
k87 Asp o 21, alpha-amylase Scientific Information
Type:
Component
Name; WHO/IUIS:
Asp o 21, alpha-amylase
Allergen code:
k87
Molecular Weight:
53 kDa
Environmental Characteristics
Baker's asthma caused by flour dust exposure is a classic example of occupational asthma. To improve industrial processes and product quality, several non-cereal additives are today regularly present in baking flours. The starch-degrading enzyme alpha-amylase from Aspergillus oryzae and soybean flour (Glycine max) are examples of such additives that, together with the cereal components, are responsible for sensitization and IgE-mediated symptoms in bakers and others exposed to baking flours (1).
Clinical Relevance
Sensitization to enzymes in bakeries is a common cause of occupational asthma (2, 3) and may also result in contact dermatitis (4). The prevalence of sensitization to alpha-amylase in symptomatic bakers is about 25% (5, 6). Epidemiologic studies have shown that alpha-amylase exposure is strongly associated with enhanced risk of sensitization of bakery workers (7, 8). Bread made from flour containing alpha-amylase from Aspergillus oryzae can trigger allergic reactions in sensitized subjects (9, 10). Clinical reactions after ingestion of baked bread do not occur frequently in those sensitized to Aspergillus fumigatus (11).
Baker's asthma caused by flour dust exposure is a classic example of occupational asthma. To improve industrial processes and product quality, several non-cereal additives are today regularly present in baking flours. The starch-degrading enzyme alpha-amylase from Aspergillus oryzae and soybean flour (Glycine max) are examples of such additives, that together with the cereal components are responsible for sensitization and IgE-mediated symptoms in bakers and others exposed to baking flours (1). Sensitization to enzymes in bakeries is a common cause of occupational asthma (2, 3) and may also result in contact dermatitis (4). The prevalence of sensitization to alpha-amylase in symptomatic bakers is about 25% (5, 6). Epidemiologic studies have shown that alpha-amylase exposure is strongly associated with enhanced risk of sensitization of bakery workers (7, 8).
Work-related respiratory symptoms in small bakeries were significantly associated with specific IgE to alpha-amylase (12). In a study of 83 pharmaceutical-industry workers, sensitization was demonstrated in 26 workers by skin testing. Specific IgE antibodies were found in 24/26 of the workers with positive skin tests. No amylase-specific IgE antibodies were detected in non-exposed controls (13). Alpha-amylase hypersensitivity with positive skin tests and specific IgE antibodies was also observed in millers (14). Fungi naturally growing on stored cereal grains were assumed to be the source of the sensitization. This assumption is supported by the observation of common epitopes in amylase and extracts of Aspergillus oryzae (15).
The enzyme alpha-amylase, also referred to as allergen Asp o 2, loses its enzyme activity when heated. However, it has been shown to retain some allergenic activity even at 200° C (5, 16) and bread made from flour containing alpha -amylase from Aspergillus oryzae can trigger allergic reactions in sensitized subjects (9, 10). Clinical reactions after ingestion of baked bread do not occur frequently in those sensitized to Aspergillus fumigatus (11).
Molecular Aspects
Non-cereal additives such as the starch-degrading enzyme alpha-amylase from Aspergillus oryzae are today regularly present in baking flours.
Cross-reactivity
Alpha-amylase hypersensitivity with positive skin tests and specific IgE antibodies was also observed in millers (14). Fungi naturally growing on stored cereal grains were assumed to be the source of the sensitization. This assumption is supported by the observation of common epitopes in amylase and extracts of Aspergillus oryzae (15).
Compiled By
Last reviewed: June 2022.
References
  1. Baur X, Weiss W, Sauer W, Fruhmann G, Kimm KW, Ulmer WT, et al. [Baking ingredients as a contributory cause of baker's asthma]. Deutsche medizinische Wochenschrift (1946). 1988;113(33):1275-8.
  2. Blanco Carmona JG, Juste Picón S, Garcés Sotillos M. Occupational asthma in bakeries caused by sensitivity to alpha-amylase. Allergy. 1991;46(4):274-6.
  3. Moneret-Vautrin DA, Kanny G, Lagrange A. [Occupational asthma caused by organic substances]. La Revue de medecine interne. 1994;15 Suppl 2:216s-25s.
  4. Morren MA, Janssens V, Dooms-Gossens A, Van Hoeyveld E, Cornelis A, De Wolf-Peeters C, et al. alpha-Amylase, a flour additive: an important cause of protein contact dermatitis in bakers. Journal of the American Academy of Dermatology. 1993;29(5 Pt 1):723-8.
  5. Baur X, Chen Z, Sander I. Isolation and denomination of an important allergen in baking additives: alpha-amylase from Aspergillus oryzae (Asp o II). Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 1994;24(5):465-70.
  6. Baur X, Degens PO, Sander I. Baker's asthma: still among the most frequent occupational respiratory disorders. The Journal of allergy and clinical immunology. 1998;102(6 Pt 1):984-97.
  7. Houba R, Heederik DJ, Doekes G, van Run PE. Exposure-sensitization relationship for alpha-amylase allergens in the baking industry. American journal of respiratory and critical care medicine. 1996;154(1):130-6.
  8. Houba R, van Run P, Doekes G, Heederik D, Spithoven J. Airborne levels of alpha-amylase allergens in bakeries. The Journal of allergy and clinical immunology. 1997;99(3):286-92.
  9. Baur X, Czuppon AB. Allergic reaction after eating alpha-amylase (Asp o 2)-containing bread. A case report. Allergy. 1995;50(1):85-7.
  10. Kanny G, Moneret-Vautrin DA. alpha-Amylase contained in bread can induce food allergy. The Journal of allergy and clinical immunology. 1995;95(1 Pt 1):132-3.
  11. Cullinan P, Cook A, Jones M, Cannon J, Fitzgerald B, Taylor AJ. Clinical responses to ingested fungal alpha-amylase and hemicellulase in persons sensitized to Aspergillus fumigatus? Allergy. 1997;52(3):346-9.
  12. Jeffrey P, Griffin P, Gibson M, Curran AD. Small bakeries--a cross-sectional study of respiratory symptoms, sensitization and dust exposure. Occupational medicine (Oxford, England). 1999;49(4):237-41.
  13. Losada E, Hinojosa M, Quirce S, Sánchez-Cano M, Moneo I. Occupational asthma caused by alpha-amylase inhalation: clinical and immunologic findings and bronchial response patterns. The Journal of allergy and clinical immunology. 1992;89(1 Pt 1):118-25.
  14. Moneo I, Alday E, Gonzalez-Muñoz M, Maqueda J, Curiel G, Lucena R. alpha-Amylase hypersensitivity in non-exposed millers. Occupational medicine (Oxford, England). 1994;44(2):91-4.
  15. Quirce S, Cuevas M, Díez-Gómez M, Fernández-Rivas M, Hinojosa M, González R, et al. Respiratory allergy to Aspergillus-derived enzymes in bakers' asthma. The Journal of allergy and clinical immunology. 1992;90(6 Pt 1):970-8.
  16. Baur X, Czuppon AB, Sander I. Heating inactivates the enzymatic activity and partially inactivates the allergenic activity of Asp o 2. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 1996;26(2):232-4