Whole Allergen
m3 Aspergillus fumigatus
m3 Aspergillus fumigatus Scientific Information
Type:
Whole Allergen
Display Name:
Aspergillus fumigatus
Allergen code:
m3
Family:
Trichocomaceae
Route of Exposure:
Inhalation
Latin Name:
Aspergillus fumigatus
Other Names:
Neosartorya fumigata, Sartorya fumigata (obsolete)
WHO/ICD-11 code:
XN5Z7
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Allergen
Summary
Aspergillus fumigatus (A. fumigatus) is a saprotroph and opportunistic filamentous fungus widely distributed in the environment. A. fumigatus is a spore-forming, thermotolerant fungus that reproduces asexually through spores in the form of conidia. It is a cosmopolitan pathogen distributed all over the five continents (Europe, Asia, America, Africa and Australia). Inhalation is the main route of exposure to Aspergillus spores. A. fumigatus is the causal agent of allergic and infectious diseases affecting dozens of millions of people globally. Allergic bronchopulmonary aspergillosis (ABPA) is the most severe Aspergillus-related allergic disease, affecting 1-2.5% of asthmatic patients and up to 10% of cystic fibrosis patients during their lifetime. Other significant allergic Aspergillus-related diseases are allergic fungal rhinosinusitis (AFRS) and severe asthma with fungal sensitization (SAFS). Up to now, 23 allergens of A. fumigatus have been characterized. Among them, Asp f 1 and Asp f 2 are major allergens, Asp f 3 has extensive cross-reactivity, and Asp f 4 and Asp f 6 are associated with ABPA diagnosis. Moreover, Asp f 1 and Asp f 2 are markers of genuine sensitization to A. fumigatus. Cross-reactivity has been mainly observed between A. fumigatus and other fungal species.
Nature
Aspergillus fumigatus (A. fumigatus) is a saprotroph and opportunistic filamentous fungus widely distributed in the environment. It sporulates abundantly with airborne conidia (asexual spores). It can grow and survive at a wide range of temperatures (25°C to 55°C) (1, 2). However, seasonal variation is seen in its growth influenced by geographical location (1). Elevated spore counts are reported to occur mostly during the summer, monsoon and autumn seasons due to effect of temperature and humidity in certain regions like India (1, 3). In contrast, there are studies, which reported weak or no correlation between A. fumigatus spore counts and seasonal variation with the persistent occurrence of low level of A. fumigatus throughout the year (1).
The conidia of A. fumigatus are small (2-3 µm), much more hydrophobic than those produced by other molds and can disperse easily. A. fumigatus shares remarkable stress tolerance features, such as thermotolerance (A. fumigatus conidia survive at 70°C), xerotolerance, resistance to acid pH, tolerance to low O2 levels, and nutritional versatility with other Aspergillus species (2, 4, 5).
Habitat
The main ecological niche of A. fumigatus is environmental decaying organic matter, where it contributes to carbon and nitrogen recycling. Consequently, A. fumigatus is abundant in soil, decaying plants, stored grain and fruit, silage, compost, stagnant water, sewage. The optimal conditions for A. fumigatus growth and conidiogenesis are a temperature of 37°C and an oxygen-rich gaseous environment. Under these conditions, A. fumigatus growth occurs almost independently of the degree of moisture and water activity (i.e., proportion of water which is unbound and thus available for microorganismal growth) (2, 5-7).
Taxonomy
There are more than 300 species belonging to the genus Aspergillus. Among them A. fumigatus is the most frequently occurring species in the environment (8) and also the most relevant for human disease. Below is the taxonomic classification of A. fumigatus.
Taxonomic tree of Aspergillus fumigatus (8)
Domain
Eukaryota
Kingdom
Fungi
Phylum
Ascomycota
Class
Eurotiomycetes
Order
Eurotiales
Family
Trichocomaceae
Genus
Aspergillus
Species
A. fumigatus
Tissue
A. fumigatus is a spore-forming filamentous fungus that reproduces asexually by producing spores in the form of conidia. Conidial heads are typically columnar (up to 400 x 50 µm) and uniseriate. Conidiophore stipes are short, smooth-walled, and have conical-shaped terminal vesicles, which support a single row of phialides on the upper two thirds of the vesicle. Conidia grow into hyphae whose cell wall is basically composed of fibrillar β-1, 3-glucan and chitin (8).
Epidemiology
Worldwide Distribution
In the vast majority of humans, daily inhalation of A. fumigatus spores does not result in disease, thanks to effective mucociliary clearance and innate immune defenses involving neutrophils and macrophages (9). However, A. fumigatus is the causal agent of allergic and infectious diseases affecting dozens of millions of people worldwide (10).
Aspergillus-related allergic (type I hypersensitivity, IgE) diseases mainly affect asthmatic and cystic fibrosis (CF) patients, although rhino-sinusal forms may develop in otherwise nonaffected subjects (11). Aspergillus sensitization, evidenced by skin or blood specific IgE tests, is associated with poorer lung function in adults and children with asthma, CF and chronic obstructive bronchopulmonary disease (COPD) (12-14).  
Hypersensitivity pneumonitis (type III hypersensitivity, IgG) due to A. fumigatus may develop in the context of occupational exposure, for example in farmers and in rural, malt, or stucco workers (15, 16).
Allergic bronchopulmonary aspergillosis (ABPA), the most severe Aspergillus-related allergic disease, has an estimated global prevalence of almost 5,000,000, affecting mostly asthmatic and CF patients (10, 17).  
ABPA is estimated to affect 1-2.5% of asthmatic patients and up to 10% of CF patients (18-20). These figures may vary with the severity of asthma, comorbidities, fungal ecology, diagnostic criteria, and country of residence (21, 22). Considering the subgroup of A. fumigatus-sensitized asthma, a systematic review and meta-analysis found that ABPA diagnostic criteria were met by 40% of patients (95% CI 27–53, range 6% - 68%). A. fumigatus sensitization (AFS) itself is a frequent finding in bronchial asthma, demonstrated in 28% of patients (95% CI 24–34, range 15% - 48%). The prevalence of ABPA was found to be high (70%) in severe asthmatic patients as shown in a study among 50 severe asthmatic patients (23). Finally, the prevalence of ABPA appeared as significantly higher in industrialized than in developing countries (25.6% vs. 12.7%, p = 0.0001) (21). Also, it was reported significantly higher in adults than in children (10.1% vs. 8.9%, p<0.0001) (20).
Other allergic Aspergillus-related diseases are allergic fungal rhinosinusitis (AFRS) and severe asthma with fungal sensitization (SAFS) (24, 25).The prevalence of AFRS is reported to range from 2.7 to 19% (24). AFRS is a unique form of immune-mediated non-invasive fungal rhinosinusitis (26). It is definitely higher in arid and tropical climates, such as in the Asia-Pacific region, Australia, Thailand, Malaysia, India, the Middle East, Saudi Arabia, North Africa, and southeastern and southwestern parts of the US, especially the Mississippi basin (26, 27). Reported prevalence in general population may be as high as 100 to 500 per 100,000 in India and Israel. Climate influence is prominent, as demonstrated by a prevalence of 0.4% in Northern US states compared to over 10% in Southern ones. AFRS can be caused by A. fumigatus, but also by other Aspergillus species, e.g. A. flavus, and by dematiaceous fungi (26 - 28).
SAFS is another phenotype of severe asthma associated with fungal sensitization in adults having overlapping characteristics with ABPA. However, it is distinct from ABPA as it presents without serious bronchopulmonary impairment (bronchiectasis and mucus plugging) and is diagnosed only on exclusion of ABPA (14, 29). As per a multi-national prevalence estimate analysis of 43 reports, the average global incidence of SAFS was observed to be 154 cases/100,000 cases. Further, the incidence in South and Central America was found to be significantly higher than other continents like Asia, Europe, Africa or North America (10). Prevalence rate of SAFS due to Aspergillus species (spp.) sensitization was found to be 18.3% among the 109 uncontrolled severe asthmatic patients in a cross-sectional study in Singapore (30). Similarly a Japanese study on 124 severe asthma patients reported sensitization prevalence of Aspergillus spp.to be 11% (31). Aspergillus-related infections can be divided into invasive and non-invasive forms, with the former preferentially affecting immunocompromised patients. Invasive aspergillosis (IA) has an estimated global annual incidence of 4.10 cases/100,000 cases, and chronic pulmonary aspergillosis (CPA) is estimated at a global annual incidence of 22 cases/1,000,000 cases as per a global and multi-national prevalence study. A high mortality rate is associated with such infections (10).
Pediatric issues
Allergic A. fumigatus-related diseases such as ABPA, SAFS, and AFRS are slightly less frequent in children than in adults. ABPA occurrence in children younger than 4 years is infrequent (10).
Environmental Characteristics
Living environment
Soil is the main habitat of Aspergillus spp. which has adapted to grow in adverse environmental conditions (29). The most prominent outdoor places where it grows are plants and dead and decayed organic matter. Freshwater and marine habitats are also favorable niches. In indoor environments it is mostly found on the surfaces of buildings, household appliances, in drinking water, and on dust (2). A. fumigatus can grow at 37°C, explaining its ability to colonize the airways and induce potentially severe infections in immunocompromised hosts (32).
Worldwide distribution
A. fumigatus is a cosmopolitan, coping with virtually all environments: deserts, forests, wetland, and cultivated soils around the world (33). Further, it is a ubiquitous pathogen distributed all over the globe covering all the five continents (Europe, Asia, America, Africa and Australia) (32, 34).
Route of Exposure
Main
Large amounts of conidia are released from the mycelial mass in the atmosphere. Humans are estimated to inhale 100 to 1,000 conidia each day from the atmosphere. Due to their small size, in the absence of efficient muco-ciliary clearance, Aspergillus conidia easily reach lung alveoli and may cause colonization even in normal subjects, and respiratory tract infections in susceptible hosts (8, 35).
Secondary
Other secondary route of A. fumigatus exposure may include transmission through contaminated water, also referred to as ‘wet route of transmission’. This may include drinking of contaminated water, accidental inhalation of fluid into the lungs or even through contact with exposed wounds (36). The widespread presence of A. fumigatus in stagnant water and water supply systems in hospitals poses considerable infectious risks for immunocompromised patients (37, 38). This source of contamination from potable water supply of hospitals still poses a hazard risk for immunocompromised patients as per a recent study (39).
Clinical Relevance
Aspergillus spp. is known to cause a spectrum of lung disorders including allergic (AFRS, ABPA, IgE-mediated asthma, SAFS) and infectious diseases (invasive aspergillosis, chronic pulmonary aspergillosis) (17, 25, 40).
Allergic rhinitis and rhinosinusitis
AFRS is classified as a form of non-invasive, eosinophilic, chronic fungal rhinosinusitis (27, 28). From a pathophysiologic viewpoint, AFRS is closely related to ABPA, exhibiting inflammation, Th2-skewed anti-A. fumigatus lymphocyte responses with eosinophil involvement, while IgE production is inconsistently demonstrated. AFRS is rarely found alone, as it has been demonstrated that 66% of AFRS patients may have co-existing allergic rhinitis (AR), 90% of them are fungi-sensitized (increased IgE levels)  and approximately 50% may have co-existing asthma (26, 27). Sensitization is more frequent in patients with atopic diseases; conversely, apparent monosensitization to molds is infrequent. In atopic patients, fungal antigenic material is released and induces a chain of immunologic reactions favoring the development of AFRS.  In a study on 131 AR patients, 32 (24.4%) displayed Aspergillus sensitization, of whom 28 had associated sinusitis; however, only 2 patients met all the criteria for allergic Aspergillus rhinosinusitis (27, 41). AFRS can be complicated by concomitant fungal presence and persistence in non-invasive forms, e.g. mycetoma (aspergilloma), a fungal ball in the sinonasal cavities (29, 42), or even invasive forms with local or systemic infection. The latter group is beyond the scope of the allergy referral, as their treatment relies on antifungal medication and surgery (24, 42).
Asthma
The two most important conditions associated with sensitization to Aspergillus spp. in asthmatic patients are ABPA and SAFS (25, 43).
Allergic bronchopulmonary aspergillosis (ABPA)
ABPA is an immune-mediated lung disease due to hypersensitivity to Aspergillus that mostly occurs in individuals suffering from asthma and CF (17, 43), however it can also occur in non-asthmatics (40). Sensitized individuals may develop allergic reactions on recurrent inhalation of Aspergillus spores, that is mainly a type I (IgE-mediated) allergic reaction. Additionally, type-III (IgG-mediated immune complex) and type IV (cell-mediated, mainly eosinophils) reactions also play a role (25, 40, 43).
Typically, ABPA follows a flare and relapse pattern. Asthma exacerbations, poor asthma control, recurrent pulmonary infiltrate resulting in irreversible bronchial remodeling with bronchiectasis form the natural history of ABPA. Another distinctive feature of ABPA is expectoration of brownish mucous plugs (mucus along with fungal hyphae and eosinophils), observed in 5-69% of the patients and representing the clinical counterpart of the computed tomography scan “mucoid impactions”  (17, 40). Asthma severity in ABPA patients can range from mild intermittent asthma to severe persistent asthma that may be corticosteroid dependent (19). In CF patients, ABPA-related exacerbation with reduced lung function can be accompanied by hemoptysis, increased microbial colonization, pneumothorax, and worsened nutritional status (23). In both asthmatic and CF patients, untreated ABPA results in severe lung damage with central bronchiectasis, fibrosis and respiratory failure (17, 40).
The diagnosis of ABPA in asthma or CF patients is complicated due to overlapping clinical features and involves clinical history, immunological findings (such as increased levels of both total serum IgE and A. fumigatus–sIgE as well as Aspergillus-specific IgG), radiological observations (17, 23). The ISHAM (International Society for Human and Animal Mycology) working group has proposed a classification of ABPA in seven stages, ranging from asymptomatic (serological) expression to respiratory failure (17, 40).
A prospective study on 35 severe asthmatic ABPA patients reported the prevalence of serological ABPA at 68.5%, compared 31.4% for that of the full-blown form of ABPA with bronchiectasis (23).
Severe asthma with fungal sensitization (SAFS)
Aspergillus species sensitization has been reported to enhance asthma severity, resulting in worsening of asthma control. A subset of patients diagnosed with severe asthma and fungal sensitization was defined as SAFS (30, 31, 44). In contrast to ABPA, SAFS does not result in mucoid impactions or bronchiectasis. Additionally, it is associated only with severe asthma, unlike ABPA that can also occur in mild and moderate asthmatic patients (25).
A retrospective study conducted in United Kingdom (UK) among 135 patients with severe asthma reported a prevalence of 60% for SAFS due to A. fumigatus (44). In Singapore and Japan, the reported prevalence of Aspergillus spp. sensitization among severe asthmatic patients was 11.7% (n=206) and 11% (n=124) respectively (30, 31). The occurrence of SAFS was reported at a younger age as compared to patients without fungal sensitization (31). In accordance to this study, another study conducted in UK on 82 children (4-17 years) reported presence of SAFS in 46% children with onset of asthma at early age (45).
Atopic Dermatitis (AD)
A cross-sectional study in China conducted among 59 AD patients without respiratory disease found Aspergillus spp. sensitization in 81.4% of patients along with other fungal sensitizations like Penicillium spp. or Candida spp. (46). The contribution of cross-reactions is probably important as suggested by a retrospective study conducted in the Czech Republic, which found only 8% of 60 AD patients were sensitized to Asp f 1, a marker of genuine sensitization to A. fumigatus (47).
Other Diseases
Due to its ability to interact with multiple players of the host immune response, A. fumigatus induces a continuous spectrum of allergic and infectious diseases (10). (see above, “Epidemiology” section).  
Prevention and Therapy
Prevention Strategies
Avoidance
Aspergillus spp. is an environmental pathogen (health-care-acquired linked with humid and dusty environment), infection may occur due to contamination of the health-care facility. The implicated environmental vehicles are mainly air filters, air conditioners, damaged ventilation system, air filter frames etc.(48). To prevent the spread of A. fumigatus spores, HEPA filtration limits the conidial air counts (36). A protected environment with high efficiency HEPA filtration along with laminar flow, least air leakage in the room, increased number of hourly air changes, as well as regular maintenance of HEPA filtration systems are recommended in hospital rooms (particularly for immunocompromised patients) (48-50).
Molecular Aspects
Allergenic molecules
Fungal species are characterized by a large array of allergenic molecules, and A. fumigatus is an outstanding example, with 23 characterized allergens featured in the WHO/IUIS database accessed on February 8th, 2021 (51). The allergens of A. fumigatus are difficult to classify as major and minor allergens owing to high variation in the sensitization pattern among different allergic manifestations of A. fumigatus (52). (pg 131). The table below provides the information of the allergenic molecules of A. fumigatus identified by WHO/IUIS (51).
Allergens
Molecular weight
Biochemical name
Allergenicity
Asp f 1
18 kDa
Mitogilin family
·  Major secreted allergen (53, 54)
·  Species specific and marker of genuine sensitization (54, 55).
·  Not present in spores, produced after gemination process (55)
·  Significant in asthmatic and ABPA patients (54).
·  Recognised by 83% of ABPA patients and 45% of A. fumigatus allergic patients (52) (pg 133)
Asp f 2
37 kDa
·  Major allergen (54).
·  Species specific and marker of genuine sensitization (54, 55).
·  Intracellular allergen solely identified by ABPA patients (asthma and CF) (52).          (pg 366)
·  Recognised by 100% of ABPA patients while none of the A. fumigatus allergic patients (52). (pg 133)
Asp f 3
19 kDa
Peroxysomal protein
·  Species non-specific (54). 
·  ABPA-related intracellular protein (54).
·  Recognised by 88% of ABPA patients and 52% of A. fumigatus allergic patients (52) (pg 133).
·  May be helpful in differentiating between ABPA and non-ABPA patients (54).
Asp f 4
30 kDa
unknown
·  Species specific (55, 56)
·  Specific for ABPA diagnosis (53)
·  ABPA-related intracellular protein solely recognized by ABPA patients (asthma and CF) (52) (pg 132 and 134).
·  Recognised by 80% of ABPA patients while none of the A. fumigatus allergic patients (52) (pg 133).
Asp f 5
40 kDa
Metalloprotease
·  Recognised by 93% of ABPA patients and 74% of A. fumigatus allergic patients (52) (pg 133).
Asp f 6
26.5 kDa
Mn superoxide dismutase
·  Species non-specific (54).
·  Specific for ABPA diagnosis (53)
·  ABPA-related intracellular protein solely recognized by ABPA patients (asthma and CF) (52) (pg 132 and 134).
·  High IgE levels observed in patients with comorbid atopic dermatitis in A. fumigatus sensitized asthmatic patietns (57)
·  Recognised by 55% of ABPA patients while none of the A. fumigatus allergic patients (52) (pg 133).
Asp f 7
12 kDa
·  Recognised by 46% of ABPA patients and 29% of A. fumigatus allergic patients (52) (pg 133).
Asp f 8
11 kDa
Ribosomal protein P2
·  Recognised by 15% of ABPA patients and 8% of A. fumigatus allergic patients (52) (pg 133).
Asp f 9
34 kDa
·  Recognised by 89% of ABPA patients and 31% of A. fumigatus allergic patients (52) (pg 133).
Asp f 10
34 kDa
Aspartate protease
·  Recognised by 28% of ABPA patients and 3% of A. fumigatus allergic patients (52) (pg 133).
Asp f 11
24 kDa
Peptidyl-prolyl isomerase
·  Recognised by 90% of A. fumigatus allergic patients (52) (pg 133).
 Asp f 12
90 kDa
Heat shock protein P90
·  Recognized by serum IgE and IgG from  ABPA patients (58, 59)
Asp f 13
34 kDa
Alkaline serine protease
·  Detected in the airway of AIA patients and may co-relate with asthma severity (60).
Asp f 15
16 kDa
Unknown
·  NA
Asp f 16
43 kDa
Unknown
·  Recognised by 70% of ABPA patients (52) (pg 133).
Asp f 17
27 kDa
·  NA
Asp f 18
34 kDa
Vacuolar serine protease
·  Recognised by 79% of A. fumigatus allergic patients (52) (pg 133).
Asp f 22
46 kDa
Enolase
·  Recognised by 86% of A. fumigatus allergic patients (52) (pg 133).
Asp f 23
44 kDa
L3 ribosomal protein
·   Identified following IgG and IgE screening of cDNA library with sera from ABPA patients (61)
Asp f 27
18 kDa
Cyclophilin
·  Recognised by 75% of ABPA patients (52) (pg 133).
Asp f 28
13 kDa
Thioredoxin
·  Recognised by 30% of ABPA patients (52) (pg 133).
Asp f 29
13 kDa
Thioredoxin
·  Recognised by 50% of ABPA patients (52) (pg 133).
Asp f 34
20 kDa
PhiA cell wall protein
·  Recognised by 93% of ABPA patients and 46% of A. fumigatus allergic patients (52) (pg 133).
kDa: kilodaltons, IgE: Immunoglobulin E, AIA: A. fumigatus sensitized asthma ABPA: Allergic bronchopulmonary aspergillosis, CF: Cystic fibrosis, NA: Not available.
Biomarkers of severity
 Major allergens Asp f 1 and Asp f 2 are considered as markers of genuine sensitization to A. fumigatus and could help differentiate primary sensitization from cross-reactive sensitization (54 - 56). However, sensitization to Asp f 1 and Asp f 2 is reported in both ABPA and asthmatic patients (54).  IgE reactivity to Asp f 3, Asp f 4 and Asp f 6 is considered specific for ABPA in the context of genuine sensitization (IgE to Asp f 1 or Asp f 2), but Asp f 3 and Asp f 6 are cross-reactive and not specific for A. fumigatus sensitization, while Asp f 4 is said to be specific for A. fumigatus (54, 55). Thus, combined detection of IgE reactivity to all 5 allergens of A. fumigatus can help to identify A. fumigatus sensitized ABPA patients (54).
Furthermore, IgE reactivity to Asp f 1, Asp f 4, and Asp f 6 was found to be higher in ABPA patients as compared to asthmatic patients. Thus, the levels of IgE antibodies towards all 5 allergens could help to differentiate between ABPA and A. fumigatus sensitized asthma patients (53). In addition, mono-sensitization to Asp f 6 was reported in association to AD, possibly as a result of cross-reaction with fungi other than A. fumigatus, thus possessing a very low risk of ABPA (55).
Cross-reactivity
Extensive cross-reactivity is linked to the homology between allergens found in A. fumigatus and other allergenic molds, mainly Cladosporium, Penicillium, Alternaria and Malassezia.
The table below provides the list of most common cross-reactive A. fumigatus allergens with other fungal allergens (55):
A. fumigatus allergen
Fungal Homologs IUIS allergens
Cross-reactive species
Asp f 3
Pen c 3
Cand b 2
Mala f 3, 4
Penicillium citrinum
Candida boidinii
Malassezia furfur
Asp f 5
-
Aspergillus oryzae (52) (pg 134).
Asp f 6
Alt a 14
Mala s 11
Alternaria alternata
Malassezia sympodialis
Asp f 8
Alt a 5
Cla h 5
Alternaria alternata
Cladosporium herbarum
Asp f 12
Alt a 3
Mala s 10
Alternaria alternata
Malassezia sympodialis
Asp f 13
Pen ch 13
Penicillium chrysogenum (53)
Asp f 18
Pen ch 18
Penicillium chrysogenum (53)
Asp f 22
Pen c 22
Alt a 6
Cla h 6
Penicillium citrinum
Alternaria alternata
Cladosporium herbarum
Asp f 23
Pen b 26
Alt a 12
Cla h 12
Penicillium brevicompactum
Alternaria alternata
Cladosporium herbarum
Asp f 27
Mala s 6
Malassezia sympodialis
Asp 28, 29
Mala s 13
Malassezia sympodialis
Explained Results
Allergen information
A. fumigatus is a common aeroallergen of the indoor and outdoor environments. It is a saprotroph and opportunistic filamentous fungus (1). Major allergens Asp f 1 and Asp f 2 are markers of genuine sensitization to A. fumigatus (54).
Clinical information
A. fumigatus is the causal agent of potentially severe allergic (allergic rhino-sinusitis, ABPA, IgE-mediated asthma, SAFS) and infectious diseases with high rates of morbidity and mortality (17, 25, 40).
Cross-reactivity
 Aspergillus fumigatus shows important cross-reactivity, mainly with other fungi and yeast species  (52, 53, 55).
References
1. Alshareef F, Robson GD. Prevalence, persistence, and phenotypic variation of Aspergillus fumigatus in the outdoor environment in Manchester, UK, over a 2-year period. Sabouraudia. 2014;52(4):367-75.
2. Paulussen C, Hallsworth JE, Álvarez‐Pérez S, Nierman WC, Hamill PG, Blain D, et al. Ecology of aspergillosis: insights into the pathogenic potency of Aspergillus fumigatus and some other Aspergillus species. Microbial biotechnology. 2017;10(2):296-322.
3. Priyamvada H, Singh RK, Akila M, Ravikrishna R, Verma RS, Gunthe SS. Seasonal variation of the dominant allergenic fungal aerosols - One year study from southern Indian region. Sci Rep. 2017;7(1):11171.
4. Van De Veerdonk FL, Gresnigt MS, Romani L, Netea MG, Latge J-P. Aspergillus fumigatus morphology and dynamic host interactions. Nature Reviews Microbiology. 2017;15(11):661.
5. Rhodes JC. Aspergillus fumigatus: growth and virulence. Medical mycology. 2006;44 Suppl 1:S77-81.
6. Hall LA, Denning DW. Oxygen requirements of Aspergillus species. J Med Microbiol. 1994;41(5):311-5.
7. Alonso V, Cavaglieri L, Ramos AJ, Torres A, Marin S. Modelling the effect of pH and water activity in the growth of Aspergillus fumigatus isolated from corn silage. J Appl Microbiol. 2017;122(4):1048-56.
8. Fang W, Latgé J-P. Microbe profile: Aspergillus fumigatus: a saprotrophic and opportunistic fungal pathogen. Microbiology. 2018;164(8):1009.
9. Bartemes KR, Kita H. Innate and adaptive immune responses to fungi in the airway. J Allergy Clin Immunol. 2018;142(2):353-63.
10. Bongomin F, Gago S, Oladele RO, Denning DW. Global and multi-national prevalence of fungal diseases—estimate precision. Journal of fungi. 2017;3(4):57.
11. Li E, Knight JM, Wu Y, Luong A, Rodriguez A, Kheradmand F, et al. Airway mycosis in allergic airway disease. Advances in immunology. 2019;142:85-140.
12. Fairs A, Agbetile J, Hargadon B, Bourne M, Monteiro WR, Brightling CE, et al. IgE sensitization to Aspergillus fumigatus is associated with reduced lung function in asthma. Am J Respir Crit Care Med. 2010;182(11):1362-8.
13. Fillaux J, Bremont F, Murris M, Cassaing S, Rittie JL, Tetu L, et al. Assessment of Aspergillus sensitization or persistent carriage as a factor in lung function impairment in cystic fibrosis patients. Scand J Infect Dis. 2012;44(11):842-7.
14. Welsh KG, Holden KA, Wardlaw AJ, Satchwell J, Monteiro W, Pashley CH, et al. Fungal sensitization and positive fungal culture from sputum in children with asthma are associated with reduced lung function and acute asthma attacks respectively. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 2020.
15. Quirce S, Vandenplas O, Campo P, Cruz MJ, de Blay F, Koschel D, et al. Occupational hypersensitivity pneumonitis: an EAACI position paper. Allergy. 2016;71(6):765-79.
16. Greenberger PA. Hypersensitivity pneumonitis: A fibrosing alveolitis produced by inhalation of diverse antigens. J Allergy Clin Immunol. 2019;143(4):1295-301.
17. Agarwal R, Sehgal IS, Dhooria S, Aggarwal AN. Developments in the diagnosis and treatment of allergic bronchopulmonary aspergillosis. Expert review of respiratory medicine. 2016;10(12):1317-34.
18. Denning DW, Pleuvry A, Cole DC. Global burden of allergic bronchopulmonary aspergillosis with asthma and its complication chronic pulmonary aspergillosis in adults. Medical mycology. 2013;51(4):361-70.
19. Patel G, Greenberger PA. Allergic bronchopulmonary aspergillosis. Allergy and Asthma Proceedings. 2019;40(6):421-4.
20. Maturu VN, Agarwal R. Prevalence of Aspergillus sensitization and allergic bronchopulmonary aspergillosis in cystic fibrosis: systematic review and meta-analysis. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 2015;45(12):1765-78.
21. Agarwal R, Aggarwal A, Gupta D, Jindal S. Aspergillus hypersensitivity and allergic bronchopulmonary aspergillosis in patients with bronchial asthma: systematic review and meta-analysis. The international journal of Tuberculosis and Lung Disease. 2009;13(8):936-44.
22. Mortezaee V, Mahdaviani SA, Pourabdollah M, Hassanzad M, Mirenayat MS, Mehrian P, et al. Diagnosis of Allergic Bronchopulmonary Aspergillosis in patients with persistent allergic asthma using three different diagnostic algorithms. Mycoses. 2020.
23. Bhankhur D, Singla N, Aggarwal D, Chander J. Prevalence of allergic bronchopulmonary aspergillosis among patients with severe bronchial asthma in a tertiary care hospital in Northern India. Indian Journal of Pathology and Microbiology. 2019;62(1):111.
24. Wiesmuller GA, Heinzow B, Aurbach U, Bergmann KC, Bufe A, Buzina W, et al. Abridged version of the AWMF guideline for the medical clinical diagnostics of indoor mould exposure: S2K Guideline of the German Society of Hygiene, Environmental Medicine and Preventive Medicine (GHUP) in collaboration with the German Association of Allergists (AeDA), the German Society of Dermatology (DDG), the German Society for Allergology and Clinical Immunology (DGAKI), the German Society for Occupational and Environmental Medicine (DGAUM), the German Society for Hospital Hygiene (DGKH), the German Society for Pneumology and Respiratory Medicine (DGP), the German Mycological Society (DMykG), the Society for Pediatric Allergology and Environmental Medicine (GPA), the German Federal Association of Pediatric Pneumology (BAPP), and the Austrian Society for Medical Mycology (OGMM). Allergo J Int. 2017;26(5):168-93.
25. Shah A, Panjabi C. Allergic Bronchopulmonary Aspergillosis: A Perplexing Clinical Entity. Allergy Asthma Immunol Res. 2016;8(4):282-97.
26. Chakrabarti A, Kaur H. Allergic aspergillus rhinosinusitis. Journal of Fungi. 2016;2(4):32.
27. Panjabi C, Shah A. Allergic Aspergillus sinusitis and its association with allergic bronchopulmonary aspergillosis. Asia Pacific Allergy. 2011;1(3):130-7.
28. Denning DW, Chakrabarti A. Pulmonary and sinus fungal diseases in non-immunocompromised patients. Lancet Infect Dis. 2017;17(11):e357-e66.
29. Latge J-P, Chamilos G. Aspergillus fumigatus and aspergillosis in 2019. Clinical microbiology reviews. 2019;33(1).
30. Goh KJ, Yii ACA, Lapperre TS, Chan AK, Chew FT, Chotirmall SH, et al. Sensitization to Aspergillus species is associated with frequent exacerbations in severe asthma. J Asthma Allergy. 2017;10:131-40.
31. Masaki K, Fukunaga K, Matsusaka M, Kabata H, Tanosaki T, Mochimaru T, et al. Characteristics of severe asthma with fungal sensitization. Ann Allergy Asthma Immunol. 2017;119(3):253-7.
32. Pringle A, Baker D, Platt J, Wares J, Latge J, Taylor J. Cryptic speciation in the cosmopolitan and clonal human pathogenic fungus Aspergillus fumigatus. Evolution. 2005;59(9):1886-99.
33. Pennerman K, Al-Maliki H, Lee S, Bennett J. Fungal volatile organic compounds (VOCs) and the genus Aspergillus.  New and Future Developments in Microbial Biotechnology and Bioengineering: Elsevier; 2016. p. 95-115.
34. Chowdhary A, Kathuria S, Xu J, Meis JF. Emergence of azole-resistant Aspergillus fumigatus strains due to agricultural azole use creates an increasing threat to human health. PLoS pathog. 2013;9(10):e1003633.
35. Latgé JP. Aspergillus fumigatus and aspergillosis. Clinical microbiology reviews. 1999;12(2):310-50.
36. Warris A, Verweij P. Clinical implications of environmental sources for Aspergillus. Medical mycology. 2005;43(Supplement_1):S59-S65.
37. Anaissie EJ, Stratton SL, Dignani MC, Summerbell RC, Rex JH, Monson TP, et al. Pathogenic Aspergillus species recovered from a hospital water system: a 3-year prospective study. Clinical Infectious Diseases. 2002a;34(6):780-9.
38. Arvanitidou M, Spaia S, Velegraki A, Pazarloglou M, Kanetidis D, Pangidis P, et al. High level of recovery of fungi from water and dialysate in haemodialysis units. Journal of Hospital Infection. 2000;45(3):225-30.
39. Caggiano G, Diella G, Triggiano F, Bartolomeo N, Apollonio F, Campanale C, et al. Occurrence of Fungi in the Potable Water of Hospitals: A Public Health Threat. Pathogens. 2020;9(10).
40. Prasad R, Kacker R, Gupta N. Allergic bronchopulmonary aspergillosis: Indian scenario. 2019;33(2):63-9.
41. Sahay S, Shah A. Allergic rhinitis: Aspergillus sensitisation increases the severity of sinusitis in “blockers” as compared to “sneezers and runners” 162. Allergy: European Journal of Allergy and Clinical Immunology. 2008;63(88).
42. Callejas CA, Douglas RG. Fungal rhinosinusitis: what every allergist should know. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 2013;43(8):835-49.
43. Shah A, Panjabi C. Allergic aspergillosis of the respiratory tract. European Respiratory Review. 2014;23(131):8-29.
44. Farrant J, Brice H, Fowler S, Niven R. Fungal sensitisation in severe asthma is associated with the identification of Aspergillus fumigatus in sputum. J Asthma. 2016;53(7):732-5.
45. Castanhinha S, Sherburn R, Walker S, Gupta A, Bossley CJ, Buckley J, et al. Pediatric severe asthma with fungal sensitization is mediated by steroid-resistant IL-33. J Allergy Clin Immunol. 2015;136(2):312-22 e7.
46. Chang FY, Lee JH, Yang YH, Yu HH, Wang LC, Lin YT, et al. Analysis of the serum levels of fungi-specific immunoglobulin E in patients with allergic diseases. Int Arch Allergy Immunol. 2011;154(1):49-56.
47. Čelakovská J, Bukač J, Vaňková R, Krcmova I, Krejsek J, Andrýs C. Sensitisation to molecular allergens of Alternaria alternata, Cladosporium herbarum, Aspergillus fumigatus in atopic dermatitis patients. Food and Agricultural Immunology. 2019;30(1):1097-111.
48. Chinn RY, Sehulster L. Guidelines for environmental infection control in health-care facilities; recommendations of CDC and Healthcare Infection Control Practices Advisory Committee (HICPAC). 2003.
49. Anaissie EJ, Stratton SL, Dignani MC, Lee C-K, Mahfouz TH, Rex JH, et al. Cleaning patient shower facilities: a novel approach to reducing patient exposure to aerosolized Aspergillus species and other opportunistic molds. Clinical infectious diseases. 2002b;35(8):e86-e8.
50. Patterson TF, Thompson III GR, Denning DW, Fishman JA, Hadley S, Herbrecht R, et al. Practice guidelines for the diagnosis and management of aspergillosis: 2016 update by the Infectious Diseases Society of America. Clinical Infectious Diseases. 2016;63(4):e1-e60.
51. WHO/IUIS. "Aspergillus fumigatus" Allergen Nomenclature 2019 [February, 8th 2021]. Available from: http://www.allergen.org/search.php?allergenname=&allergensource=Aspergillus+fumigatus&TaxSource=&TaxOrder=&foodallerg=all&bioname=.
52. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27:1-250.
53. Luo W, Hu H, Wu Z, Wei N, Huang H, Zheng P, et al. Molecular allergen sensitization of Aspergillus fumigatus between allergic bronchopulmonary aspergillosis and A fumigatus-sensitized asthma in Guangzhou, Southern China. J Clin Lab Anal. 2020;34(10):e23448.
54. Carsin A, Romain T, Ranque S, Reynaud-Gaubert M, Dubus JC, Mege JL, et al. Aspergillus fumigatus in cystic fibrosis: An update on immune interactions and molecular diagnostics in allergic bronchopulmonary aspergillosis. Allergy. 2017;72(11):1632-42.
55. Fukutomi Y, Tanimoto H, Yasueda H, Taniguchi M. Serological diagnosis of allergic bronchopulmonary mycosis: Progress and challenges. Allergology international : official journal of the Japanese Society of Allergology. 2016;65(1):30-6.
56. Volgger V, Louza J, Gellrich D, Eder K, Gröger M. Value of Component Resolved Diagnostics to Aspergillus fumigatus in Patients with Upper Airway Complaints. Int Arch Allergy Immunol. 2021;182(2):120-30.
57. Tanimoto H, Fukutomi Y, Yasueda H, Takeuchi Y, Saito A, Watai K, et al. Molecular-based allergy diagnosis of allergic bronchopulmonary aspergillosis in Aspergillus fumigatus-sensitized Japanese patients. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 2015;45(12):1790-800.
58. Kumar A, Reddy LV, Sochanik A, Kurup VP. Isolation and characterization of a recombinant heat shock protein of Aspergillus fumigatus. J Allergy Clin Immunol. 1993;91(5):1024-30.
59. Singh B, Sharma GL, Oellerich M, Kumar R, Singh S, Bhadoria DP, et al. Novel cytosolic allergens of Aspergillus fumigatus identified from germinating conidia. J Proteome Res. 2010;9(11):5530-41.
60. Basu T, Seyedmousavi S, Sugui JA, Balenga N, Zhao M, Kwon Chung KJ, et al. <em>Aspergillus fumigatus</em> alkaline protease 1 (Alp1/<em>Asp f13</em>) in the airways correlates with asthma severity. Journal of Allergy and Clinical Immunology. 2018;141(1):423-5.e7.
61. Saxena S, Madan T, Muralidhar K, Sarma PU. cDNA cloning, expression and characterization of an allergenic L3 ribosomal protein of Aspergillus fumigatus. Clin Exp Immunol. 2003;134(1):86-91.