Whole Allergen
re212 Swine urine proteins
re212 Swine urine proteins Scientific Information
Type:
Whole Allergen
Display Name:
Swine urine proteins
Allergen code:
re212
Family:
Suidae
Species:
Sus scrofa
Route of Exposure:
Inhalation
Latin Name:
Sus scrofa
Other Names:
Pig, hog
Summary
Swine (Sus scrofa) urine proteins are inhalant allergens that cause respiratory symptoms in sensitized individuals. In addition to widespread use as production animals, pigs can be found as household pets. As such swine urine protein allergens are an important cause of allergic respiratory disease, particularly in occupational disease among farmworkers, swine production workers, and swine veterinarians.
Allergen
Nature
In addition to being common production animals, pigs (small pigs, mini pigs, or teapot pigs) are uncommon but popular pets in the home (1); in 2017, every 100 people shared the world with 13 pigs (2).
The sensitizing, allergenic agents in pigs are proteins found in their urine, hair, and dander (1, 3).
Taxonomy      
Taxonomic tree of Sus scrofa (4)
Domain
Eukaryota
Kingdom
Metazoa
Phylum
Chordata
Subphylum
Vertebrata
Class
Mammalia
Order
Artiodactyla
Family
Suidae
Genus
Sus.
Epidemiology
Worldwide distribution 
Respiratory diseases are commonly seen in workers in swine production facilities. Studies have shown up to 50% of workers in swine production facilities have bronchitis, organic dust toxic syndrome, hyperreactive airway disease, chronic mucous membrane irritation, or other respiratory problems (5). High frequencies of respiratory symptoms and airway obstruction are also seen in swine veterinarians (5) with work-related symptoms including rhinitis symptoms (60.3%), cough and chest tightness (55.2%), and wheezing (35.1%) (6).
Risk factors 
Exposure to swine urine occurs in work settings, such as farms (7), veterinary clinics (6) and production facilities (5, 6), or in the home (1). A survey of European animal farmers found that pig farmers were at highest risk of developing work-related respiratory symptoms, versus farmers working with cattle, poultry, and sheep (7).
A history of atopic disease predisposes individuals to sensitization (3); studies have shown that atopic people are more at risk of developing asthma from exposure to environmental antigens than non-atopic persons (3).
Proximity to swine confinement facilities is a risk factor for respiratory symptoms; prevalence of wheezing among adolescents was 5% higher at schools located within 3 miles of confined swine feeding, compared with those located beyond 3 miles, and 24% higher at schools in which swine odor was discernible twice per month, compared with schools with no discernible odor (8).
Environmental Characteristics
Living environment
High and chronic levels of airborne allergens in confined swine enclosures can lead to local airway and lung inflammation, as well as systemic effects in sensitized individuals (9).
Worldwide distribution 
Pigs are bred throughout the world for meat, with an estimated 112 million tons produced in 2014 [OWID]. China dominates global pigmeat output, producing just under half of total production, while other major producers include the United States, Germany, Spain, and Brazil (10).
Route of Exposure
Main
Inhalation (9, 11).
Clinical Relevance
Exposure to swine urine allergens is a significant cause of IgE-mediated occupational allergy and asthma in both exposed and naïve workers in swine production (11).
In a review of 14 epidemiologic studies from four countries (n=2,786), cough and phlegm were the most common symptoms (range, 12‒55%), with frequent reports of tightness of chest and wheezing (12‒33%), and episodes of toxic dust syndrome. Acute symptoms directly associated with work were between 1.5‒2x more prevalent than chronic symptoms, however, while IgE antibodies to swine house allergens were common, no relation to symptoms could be found (12).
A recent systematic evidence update reported the prevalence and incidence of respiratory symptoms in subjects working with swine. In a range of cross-sectional studies, exposure to swine was associated with increased risk of asthma and wheeze (odds ratio [OR]=3.4, 95% CI 1.6‒7.0), rhinoconjunctivitis (OR=1.5, 95% CI 1.2‒1.9), and chronic bronchitis (range OR: 3.2 to 4.3) (9).
A population study of animal farmers in Denmark, Germany, Switzerland, and Spain found a significant dose-response relationship for pig farmers (n=885) between daily hours worked inside animal houses and respiratory symptoms. In this study, farmers keeping only pigs had significantly higher ORs for work-related shortness of breath (OR=1.50, 95% CI 1.18‒1.92), cough without phlegm (OR=1.78, 95% CI 1.43‒2.21), wheezing (OR=1.54, 95% CI 1.16‒2.05), nasal irritation (OR=1.48, 95% CI 1.17-1.88) and flu-like illness (OR=1.37, 95% CI 1.05-1.78) than cattle farmers, and the highest risk of all work-related symptoms of all farmers evaluated except cough with phlegm. However, pig farmers had a lower OR of nasal allergies compared to other farmers (7).
A case study of four female workers at intensive swine production facilities in Canada reported that each developed acute onset of wheezing and cough suggestive of asthma within weeks of commencing full-time employment. None of the workers were aware of preexisting asthma, allergy or hay fever, and all reported improvements when exposure to swine ceased. However, two individuals remained mildly symptomatic up to 4–5 months after exposure (11).
One case study reported a highly atopic female (aged 21) with eczema who developed severe itching and shortness of breath within 5 minutes of entering an agricultural pig house. The patient’s eczema settled 2 hours after leaving the building. A skin prick test with 1.0 g/L pig urine extract elicited a 3 mm skin weal, and inhalation of a nebulized dose of pig urine (0.0015 g) for one minute caused a decline in forced expiratory volume of 70% (from 2.3 to 0.7 L). Additionally, a radioallergosorbent test (RAST) demonstrated positive specific IgE antibody to pig urine (3).
Conversely, a population-based cross-sectional study in Italy suggested that swine breeding did not increase, and might decrease, the risk of pollen sensitization and allergic disease. In this study of native-born swine breeders (n=78) and the general population (‘non-breeders’, n=82), breeders had lower prevalences of respiratory allergy (12.8% vs 31.1%, respectively, p=0.002), asthma (6.4% vs 15.8%, p=0.059), rhinitis (16.7% vs 51.2%, p<0.001), persistent cough (5.1% vs 15.9%, p=0.028), and sensitization to grass (7.7% vs 25.6%, p=0.002) (5).
Molecular Aspects
Allergenic molecules
The following allergenic molecules have been characterized from swine urine (table adapted from Allergome.org (13).
Allergen
Type
Mass (kDa)
Sus s 1
Serum albumin
70
Cross-reactivity
There are limited data available regarding the cross-reactivity of swine urine allergens with those of other species.
An early study identified a jointly-recognized cat and pig serum albumin. Inhibition experiments using sera of 10 patients with specific IgE to cat dander and pork showed that the spectrum of IgE reactivity to cat serum albumin completely contained IgE reactivity to porcine serum albumin, which the authors suggested meant that sensitization to cat was the primary event (14).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:April 2022
References
  1. Díaz-Perales A, González-de-Olano D, Pérez-Gordo M, Pastor-Vargas C. Allergy to uncommon pets: new allergies but the same allergens. Front Immunol. 2013;4:492-.
  2. Rauw WM, Rydhmer L, Kyriazakis I, Øverland M, Gilbert H, Dekkers JC, et al. Prospects for sustainability of pig production in relation to climate change and novel feed resources. Journal of the Science of Food and Agriculture. 2020;100(9):3575-86.
  3. Harries MG, Cromwell O. Occupational asthma caused by allergy to pigs' urine. Br Med J (Clin Res Ed). 1982;284(6319):867-.
  4. CABI. Sus scrofa Wallingford, UK2019 [cited 2022 31.01.22]. Available from: https://www.cabi.org/isc/datasheet/119688.
  5. Galli L, Facchetti S, Raffetti E, Donato F, D'Anna M. Respiratory diseases and allergic sensitization in swine breeders: a population-based cross-sectional study. Ann Allergy Asthma Immunol. 2015;115(5):402-7.
  6. Poole JA, LeVan TD, Slager RE, Qiu F, Severa L, Yelinek J, et al. Bronchodilator responsiveness in swine veterinarians. J Agromedicine. 2007;12(2):49-54.
  7. Radon K, Danuser B, Iversen M, Jörres R, Monso E, Opravil U, et al. Respiratory symptoms in European animal farmers. European Respiratory Journal. 2001;17(4):747-54.
  8. Mirabelli MC, Wing S, Marshall SW, Wilcosky TC. Asthma symptoms among adolescents who attend public schools that are located near confined swine feeding operations. Pediatrics. 2006;118(1):e66-75.
  9. Sigsgaard T, Basinas I, Doekes G, de Blay F, Folletti I, Heederik D, et al. Respiratory diseases and allergy in farmers working with livestock: a EAACI position paper. Clinical and Translational Allergy. 2020;10(1):29.
  10. Ritchie H, Roser M. Meat and Dairy Production Our World in Data 2017.
  11. Dosman JA, Lawson JA, Kirychuk SP, Cormier Y, Biem J, Koehncke N. Occupational asthma in newly employed workers in intensive swine confinement facilities. Eur Respir J. 2004;24(4):698-702.
  12. Donham KJ. Health effects from work in swine confinement buildings. American Journal of Industrial Medicine. 1990;17(1):17-25.
  13. allergome.org. Suidae 2021 [cited 2022 31.01.22]. Available from: https://www.allergome.org/script/search_step2.php.
  14. Hilger C, Kohnen M, Grigioni F, Lehners C, Hentges F. Allergic cross-reactions between cat and pig serum albumin. Study at the protein and DNA levels. Allergy. 1997;52(2):179-87.