Whole Allergen
t17 Japanese cedar
t17 Japanese cedar Scientific Information
Type:
Whole Allergen
Display Name:
Japanese cedar
Allergen code:
t17
Family:
Taxodiaceae
Species:
Cryptomeria japonica
Route of Exposure:
Inhalation
Latin Name:
Cupressus japonica
Other Names:
Sugi tree
Synonyms:
Cupressus japonica
WHO/ICD-11 code:
XM3940
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Japanese cedar trees (Cryptomeria japonica) are distributed widely across Japan, Taiwan and southern China and cultivated in Europe and North America as an ornamental tree. Cryptomeria pollen is a major allergen source associated with allergic rhinitis, asthma, atopic dermatitis and food-pollen allergy syndrome. Japanese cedar pollen shows high levels of cross-reactivity with other members of the Cupressaceae family including Japanese, Mediterranean and Arizona cypresses and mountain juniper. 
Allergen
Nature
The Japanese cedar tree (Cryptomeria japonica) is an evergreen tree native to Japan, Taiwan and coastal regions of China. It is often cultivated in Europe and North America as an ornamental tree (1, 2). The Cryptomeria pollen season starts in early February and peak pollen counts are seen between late February and early March, with pollen dispersing >100 km from its source (3). However, the high level of cross-reactivity between Cryptomeria and other members of the Cupressaceae family could extend the effective pollen season from December to March (4). Cedar pollen grains are relatively large (>30 µm) (5).
Taxonomy
Taxonomic tree of Cryptomeria japonica (6)
Domain
Eukaryota
Kingdom
Plantae
Phylum
Spermatophyta
Subphylum
Gymnospermae
Class
Pinopsida
Family
Taxodiaceae
Genus
Cryptomeria
Tissue
Tree pollen
Epidemiology
Worldwide distribution 
Cryptomeria japonica pollen is the most common allergen causing seasonal allergic rhinitis in Japan (2, 7). It can disperse >100km from its source, causing widespread pollinosis in major cities such as Tokyo and Osaka (3). There are regional differences in sensitization to Cryptomeria pollen across Japan, although these are relatively small compared to other tree pollens, due to the widespread distribution of cedar trees (7). In South Korea, Japanese cedar was the major outdoor allergen in spring only in Jeju district; this was attributed to localized cultivation of cedar trees (8, 9).
There appears to be a significant worldwide increase in the prevalence of cedar pollinosis (3, 10). In a 2008 national survey of Japanese otorhinolaryngologists, the prevalence of cedar pollinosis among their families was 26.5%; 10% higher than that recorded in a similar survey 10 years earlier (3).
Risk factors 
Co-morbid allergic rhinitis and a family history of cedar pollinosis increased the risk of both subjective and physician diagnosed Cryptomeria pollen allergy in Japanese patients (11). Repeated exposure to cedar pollen for over two seasons increased the risk of sensitization in Korean adults (12).
Pediatric issues 
IgE sensitization to Japanese cedar pollen is common in Japanese children at age 5 years and increases over time; this sensitization is clearly associated with allergic disease (10). In 2008, the prevalence of cedar pollinosis in Japan ranged from 1.1% in children aged 0-4 years to 31.4% in children aged 10-19 years (3). In another Japanese study, high levels of sensitization to the major Cryptomeria pollen allergen (Cry j 1) were found in 32.8% of children aged 5 years and 57.8% of children aged 9 years (10). A recent study of 258 pollen-allergic children aged 13 years found 95.7% were sensitized to Cry j 1 (13). Children sensitized to Cryptomeria showed clinical symptoms of wheeze, asthma, eczema and/ or rhinitis (10).
In a regional study of allergic Japanese children, cedar pollen-specific IgE was detected in approximately 60% of Grade 4 and 5 school children from inland sample areas, compared with 23% of children from a coastal sample area (3).
Environmental Characteristics
Worldwide distribution 
Cryptomeria japonica is a naturally growing endemic species in Japan and is widely used to create artificial forests (7). It is used in agroforestry for timber and fuel and cultivated as a boundary, shade, windbreak, or erosion control species and as an ornamental tree (6).
Detection
In Japan, pollen is often sampled using the gravimetric method via a Durham sampler, rather than the volumetric Burkard spore trap commonly used in Western countries (3).
Clinical Relevance
Japanese cedar pollen is the most common cause of seasonal pollen allergy in Japan (1, 11). During the spring it contributes significantly to allergic rhinoconjunctivitis and sinusitis (2). Sensitization to cedar (defined as specific IgE levels of class 1 or above) was found in 78.8% of 382 young Japanese adults; the prevalence of cedar pollinosis was 41.2% based on self-reported symptoms and 22.2% based on physician diagnosis (11). Although cedar pollinosis peaks in patients aged 30–50 years, the age of onset is decreasing and it is now commonly diagnosed in children aged 5 years and over (3, 10).
Rhinitis and asthma symptoms can be exacerbated during Cryptomeria pollen season, even in patients who are not sensitized to this pollen. Both Cryptomeria pollen and its major allergen (Cry j 1) induced eosinophil adhesiveness, generated super-oxide anion free radicals (O2-) and released eosinophil-derived neurotoxin in non-sensitized healthy donors as well as in Cryptomeria-sensitized patients (14).
Asthma
Cryptomeria japonica pollen is a risk factor for bronchial asthma in Japanese adults (2). Seasonal increases in the level of pro-inflammatory eicosanoids in the bronchial mucosa of cedar-sensitized patients can reduce their pulmonary function, even when receiving maintenance treatment for asthma (15). Among 152 asthmatic and cedar-sensitized patients, asthma control worsened during the pollen season despite 84% of patients receiving concomitant treatment for rhinitis (16).
Atopic Dermatitis
Cryptomeria japonica pollen affects the severity of atopic dermatitis (2).
Other diseases
Of 76 Japanese patients with chronic obstructive pulmonary disease (COPD), 35.5% demonstrated specific IgE to Japanese cedar pollen (5).
Pollen from Cryptomeria japonica is an important factor in oral allergy syndrome (OAS) (2). Pollen food allergy syndrome (PFAS) refers to pollen-allergic patients who develop OAS on consuming fruits and vegetables. Age-independent studies suggest that 10-13% of Japanese patients with cedar pollinosis also have PFAS (13). Among 59 children aged 13 years with PFAS, the top causative foods were pineapple and kiwi, and 93.2% were sensitized to Cry j 1, the major pollen allergen of Japanese cedar (13).
Molecular Aspects
Allergenic molecules
Allergens of the Cupressaceae family (4, 17).
Allergen name
Protein group (if known)
Jun v 3
Thaumatin-like protein
Jun a 3
Thaumatin-like protein
Cup s 3
Thaumatin-like protein
Jun v 1
Pectate lyase
Jun a 1
Pectate lyase
Cup s 1
Pectate lyase
Cup a 1
Pectate lyase
Cha o 1
Pectate lyase
Cry j 1
Pectate lyase
Jun a 2
Polygalacturonase
Cha o 2
Polygalacturonase
Cry j 2
Polygalacturonase
Jun o 4
Calmodulin/ Polcalcin-like protein
Cry j 4
Polcalcin
Cup s 7
Gibberellin-regulated protein
Jun a 7
Gibberellin-regulated protein
Cry j 7
Gibberellin-regulated protein
The major allergen of Cryptomeria japonica is Cry j 1. It shares 78.6% sequence identity with Cha o 1, the major allergen of the Japanese cypress (Chamaecyparis obtusa) and 79% with Cup a 1, the major allergen of the Arizona cypress (Cupressus arizonica) (1). The major Mediterranean cypress allergen (Cup s 1) and mountain juniper allergen (Jun a 1) are also part of this dominant allergen group belonging to the pectate lyase protein family, to which more than 90% of Cupressaceae pollen-allergic patients are sensitized (17). 
Biomarkers of severity
The ratio of Japanese cedar pollen-specific IgA to total IgA (rIgA) in nasal secretions of symptomatic, cedar-sensitized patients increases significantly during pollen season and correlates with rhinitis symptoms. Asymptomatic cedar-sensitized patients do not show increased rIgA, suggesting that rIgA is a useful antigen-specific biomarker for allergic rhinitis or pollinosis (18).
Cross-reactivity
The major allergens of Japanese cedar, Japanese cypress (Chamaecyparis obtusa), Arizona cypress (Cupressus arizonica), Mediterranean cypress (Cupressus sempervirens) and mountain juniper (Juniperus ashei) are highly cross-reactive with each other and together share 70%–95% sequence identity (17).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer
Last reviewed:January 2022
References
  1. Asam C, Hofer H, Wolf M, Aglas L, Wallner M. Tree pollen allergens-an update from a molecular perspective. Allergy. 2015;70(10):1201-11.
  2. LOINC. Japanese Cedar IgE Ab [Units/volume] in Serum 2021 [cited 2021 11.24.21]. Available from: https://loinc.org/14035-0/.
  3. Okamoto Y, Horiguchi S, Yamamoto H, Yonekura S, Hanazawa T. Present Situation of Cedar Pollinosis in Japan and its Immune Responses. Allergology International. 2009;58(2):155-62.
  4. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  5. Toyota H, Sugimoto N, Kobayashi K, Suzuki Y, Takeshita Y, Ito A, et al. Comprehensive analysis of allergen-specific IgE in COPD: mite-specific IgE specifically related to the diagnosis of asthma-COPD overlap. Allergy, Asthma & Clinical Immunology. 2021;17(1):13.
  6. CABI. Cryptomeria japonica (Japanese cedar) 2021 [cited 2021 24.11.21]. Available from: https://www.cabi.org/isc/datasheet/14482.
  7. Minami T, Fukutomi Y, Inada R, Tsuda M, Sekiya K, Miyazaki M, et al. Regional differences in the prevalence of sensitization to environmental allergens: Analysis on IgE antibody testing conducted at major clinical testing laboratories throughout Japan from 2002 to 2011. Allergol Int. 2019;68(4):440-9.
  8. Park SC, Hwang CS, Chung HJ, Purev M, Al Sharhan SS, Cho HJ, et al. Geographic and demographic variations of inhalant allergen sensitization in Koreans and non-Koreans. Allergol Int. 2019;68(1):68-76.
  9. Sung M, Kim SW, Kim JH, Lim DH. Regional Difference of Causative Pollen in Children with Allergic Rhinitis. J Korean Med Sci. 2017;32(6):926-32.
  10. Yamamoto-Hanada K, Borres MP, Åberg MK, Yang L, Fukuie T, Narita M, et al. IgE responses to multiple allergen components among school-aged children in a general population birth cohort in Tokyo. World Allergy Organization Journal. 2020;13(2):100105.
  11. Nakamura S, Tsunoda S, Sakaida H, Masuda S, Said AS, Takeuchi K. Analysis of factors associated with cedar pollen sensitization and development of pollinosis in a young Japanese adult population. Allergol Int. 2019;68(1):39-45.
  12. Suh MJ, Yi HJ, Kim JH, Lee K-H, Hong S-C, Kang Jw. Number of seasonal exposures to Japanese cedar pollen increases the risk of sensitization: Observational study in Korean adults. Scientific Reports. 2019;9(1):10496.
  13. Kiguchi T, Yamamoto-Hanada K, Saito-Abe M, Sato M, Irahara M, Ogita H, et al. Pollen-food allergy syndrome and component sensitization in adolescents: A Japanese population-based study. PLoS One. 2021;16(4):e0249649.
  14. Miyauchi S, Nakagome K, Noguchi T, Kobayashi T, Ueda Y, Soma T, et al. Japanese cedar pollen upregulates the effector functions of eosinophils. Asia Pac Allergy. 2021;11(3):e26.
  15. Soma T, Uchida Y, Nakagome K, Hoshi R, Nagata M. Eicosanoids seasonally impact pulmonary function in asthmatic patients with Japanese cedar pollinosis. Allergol Int. 2020;69(4):594-600.
  16. Hojo M, Ohta K, Iikura M, Hirashima J, Sugiyama H, Takahashi K. The impact of co-existing seasonal allergic rhinitis caused by Japanese Cedar Pollinosis (SAR-JCP) upon asthma control status. Allergol Int. 2015;64(2):150-5.
  17. Ehrenberg AE, Klingebiel C, Östling J, Larsson H, Mattsson L, Vitte J, et al. Characterization of a 7 kDa pollen allergen belonging to the gibberellin-regulated protein family from three Cupressaceae species. Clinical & Experimental Allergy. 2020;50(8):964-72.
  18. Suzuki M, Yokota M, Ozaki S, Matsumoto T, Nakamura Y. Japanese Cedar Pollen-Specific IgA in Nasal Secretions and Nasal Allergy Symptoms. Ann Otol Rhinol Laryngol. 2019;128(4):330-7.