Whole Allergen
t2 Grey alder Pollen
t2 Grey alder Pollen Scientific Information
Type:
Whole Allergen
Display Name:
Grey alder Pollen
Allergen code:
t2
Family:
Betulaceae
Species:
Alnus incana
Route of Exposure:
Inhalation
Latin Name:
Alnus incana
Other Names:
Speckled alder
WHO/ICD-11 code:
XM7QD6
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Grey alder (Alnus incana) belongs to family Betulaceae which also consists of birch, hazel, oak, and hornbeam tree species. It is a deciduous tree that can grow up to 24 meters. Its flowering season is from February till May and sheds pollen of size 15-40 µm. It usually favors high humidity thus can be seen near riverbanks and marshy lands. This species originated from central Europe and spread to Russia, Siberia, Asia, the US and other parts of the world. Alder is one of the significant contributors for pollinosis and inducer of respiratory allergy such as rhinitis, rhino-conjunctivitis as well as bronchial asthma. The main route of exposure for this pollen is through inhalation. To date, no allergens from grey alder pollen have been characterized. But a major allergen from common alder (Alnus glutinosa) Alt g 1 has been recognized. It shows extensive cross-reactivity with major allergens from tree pollen of the Betulaceae family.
Allergen
Nature
Grey alder (Alnus incana) is a deciduous, wind-pollinated tree and can age to nearly 60 years. It is small to moderate in size, with a height up to 24 meters. It is small in comparison to its peer common alder (Alnus glutinosa). It has smooth bark, which is grey and has fissures. The leaves of this tree are oval with sharp tips. Leaves are partly green and grey. It is a monoecious plant with a flowering season from February till May (1). Grey alder generally has flowering first, followed by black alder (2).
Habitat
Grey alder is commonly located on rivers and lakes sides, near wet meadows and marshland areas. It usually favors high humidity and can withstand high acidity up to 3.5 to 4 pH levels without any issues. It grows at elevated areas up to 1800 meters in the Caucasus region and central parts of Europe (1).
Taxonomy
Grey alder belongs to the genus Alnus belonging to the Betulaceae family and order Fagales, which also includes Betula (birch), Corylus (hazel), Quercus (oak) and Carpinus (hornbeam) species.
Taxonomic tree of Grey Alder
Domain Eukaryota
Kingdom Plantae
Phylum Spermatophyta
Subphylum Angiospermae
Class Dicotyledonae 
Order  Fagales
Family Betulaceae
Genus Alnus
Species Alnus Incana
Tissue
Alder pollen has a size ranging from 15 to 40 µm. It has a thick ektexine layer enveloping the pores. The pore count in grey alder is around 3 to 6 (4).
Epidemiology
Worldwide distribution
A steady rise has been seen in the number of allergies due to tree pollen in many parts of Europe. Studies have shown that about 10 to 15% of Poland's people and around 15 % in Norway suffer from pollinosis. The major contributors to pollinosis are alder, hazel, and birch trees (5).
In a study conducted from 2002 to 2005 in Croatia, pollens from the Betulaceae family accounted for 34% of the total pollen count in 864 patients with respiratory allergy such as rhinitis, rhino-conjunctivitis as well as bronchial asthma. Alder was responsible for 6% of the above count. The number of patients with polysensitization to the Betulaceae family pollen was 12.88%. The highest rate of allergy (46.22%) was found in the 31 to 50 years age-group, whereas the lowest rate of allergy (23.12%) was found in more than 51 years age groups(6).
In a study conducted in Korea in 2017, alder was the second most found tree pollen allergy in children (n=57) with allergic rhinitis or asthma. The tree pollen count of alder in Busan was 2nd highest with 4078 grains per cubic mm. The sensitization rates for alder was 35.1 percent in these children. Among these, oral allergy syndrome was found in 12.3% of the children (7).
Environmental Characteristics
Worldwide distribution
Grey alder has its origin from the central parts of Europe. It has extended to France, Russia, Caucasus, and Siberia eventually. Later it became common in Scandinavia and was found to be present in Britain also. One of its subspecies, hirsuta, has its origin from the central and northeast parts of the Asia region (1). The studies have found that alder pollen has been found to have traveled long distances from the Alps to Fennoscandia and from Belarus, Ukraine, to Rzeszow. It has also been reported in Greece (8). In Canada, it is found mainly in Alberta, British Columbia, Yukon, and the US, particularly in West Virginia, Maryland (9). Its subspecies like rugosa and tenuifolia are formerly from the northern regions of the United States and Canada(1).
Route of Exposure
Main
The main route of exposure for grey alder pollen is through the airway (2). Alder pollen triggers allergic reaction at the concentrations of 50 grains per cubic millimetres in 24 hours (6).
Clinical Relevance
Alder pollen (a member of the Fagales order) is a significant cause of asthma, allergic rhinitis, and allergic conjunctivitis.
Allergic rhinitis (AR) and Allergic rhinoconjunctivitis (ARC)
Allergic rhinitis is a condition affecting the quality of life of the individual. It is accompanied by other conditions like asthma, otitis media, nasal polyps, sinusitis, and lower respiratory tract infections (10). A study was conducted on 88 patients with respiratory allergy (allergic rhinitis with or without asthma). Sensitization to alder allergen was tested through specific-IgE presence and skin prick testing. The results showed sensitization to alder allergen ranged from 13.6 to 25 percent of the patients (11).
Asthma
In one of the Korean studies conducted in 57 patients, allergic rhinitis was seen in 98.2% of patients. The study also reported other symptoms such as allergic conjunctivitis, atopic dermatitis, bronchial asthma, food allergy, and oral allergy syndrome in 56.1%, 42.1%, 28.1%, 24.6%, and 12.3% patients, respectively. Alder was the second most found tree pollen in patients with allergic rhinitis or asthma. The sensitization rates to alder were 35.1 percent among these patients (7).
Atopic dermatitis
Patients with atopic dermatitis might develop the disease flare-up after exposure to an allergen such as grey alder. The most common sites include areas not covered by clothes, including the head and neck region. In a study done in 125 patients with atopic dermatitis, grey alder sensitization was found in 4.8% (12).
Prevention and Therapy
Prevention strategies
Avoidance
Alder allergy can be reduced or avoided by staying indoors even when the pollen count is low (13).
Allergen immunotherapy
A double-blind, placebo-controlled study was conducted in 22 patients with allergic rhinitis to evaluate intranasal immunotherapy's safety and efficacy. They received immunotherapy or placebo for 4 months. Immunotherapy consisted of birch and grey alder allergens. During the pollen season, the patients who took immunotherapy showed fewer sneezing episodes and runny nose compared to placebo and these differences were statistically significant. The requirement for drug treatment was also significantly decreased (14).
Molecular Aspects
Allergenic molecules
No allergens have been characterized by grey alder (Alnus incana). But some allergens have been characterized by common alder (Alnus glutinosa). The major allergen found in alder is Aln g I (2). The molecular weight of this allergen Aln g I was estimated to be about 16 to 20 kd, which corresponds to 160 to 180 amino acid sequences (15). Aln g 2, a profilin (actin-binding) allergen, and Aln g 4 a calcium-binding allergen has been identified (16-18). The WHO/IUIS has listed the following alder allergens.
Allergen
Biochemical Name
Molecular Weight (kDa)
Allergenicity
Aln g I
Pathogenesis-related protein, PR-10, Bet v 1 family member
18
In the test of immunoblot done in 9 tree-pollen allergic individuals, all of them showed positive IgE binding to Aln g I (19)
Aln g 4
Polcalcin
6-7
In a test for IgE reactivity, Aln g 4 showed positive results in 18% of the individuals with pollen allergy (20)
Biomarkers of severity
Aln g I the major allergen in alder is responsible for allergenicity (2).
Cross-reactivity
Alder pollen allergen (Aln g I) shows extensive cross-reactivity with major allergens from tree pollen such as birch (Bet v I) and hazel (Cor a I) in polyacrylamide gel electrophoresis studies (6, 21). Pollens from hazel, alder, oak, beech, hornbeam and chestnut have structurally related allergens with common IgE-binding sites responsible for their cross-reactivity(22).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: November 2020
References
  1. Durrant T, de Rigo D, Caudullo G. Alnus incana in Europe: distribution, habitat, usage and threats. 2016.
  2. Smith M, Emberlin J, Stach A, Czarnecka-Operacz M, Jenerowicz D, Silny W. Regional importance of Alnus pollen as an aeroallergen: a comparative study of Alnus pollen counts from Worcester (UK) and Poznań (Poland). Ann Agric Environ Med. 2007;14(1):123-8.
  3. CABI. Invasive Species Compendium 2019 [11-11-2020]. Available from: https://www.cabi.org/isc/datasheet/4578.
  4. Leopold E, Birkebak J, Reinink-Smith L, Jayachandar A, Narváez P, Zaborac-Reed S. Pollen morphology of the three subgenera of Alnus. Palynology. 2012;36:1-21.
  5. Piotrowska K. Comparison of Alnus, Corylus and Betula pollen counts in Lublin (Poland) and Skien (Norway). Ann Agric Environ Med. 2004;11(2):205-8.
  6. Peternel R, Milanović S, Hrga I, Mileta T, Čulig J. Incidence of Betulaceae pollen and pollinosis in Zagreb, Croatia, 2002-2005. Annals of agricultural and environmental medicine : AAEM. 2007;14:87-91.
  7. Park Y, Hwang Y. Tree Pollen Sensitization and Cross-Reaction of Children with Allergic Rhinitis or Asthma. Kosin Medical Journal. 2019;34:126.
  8. Ghasemifard H, Ghada W, Estrella N, Lüpke M, Oteros J, Damialis A, et al. High post-season Alnus pollen loads successfully identified as long-range transport of an alpine species. Atmospheric Environment. 2020:117453.
  9. Plant Guide [Internet]. USDA NRCS. 2006. Available from: https://plants.usda.gov/plantguide/pdf/cs_alinr.pdf.
  10. Kouzegaran S, Zamani MA, Faridhosseini R, Rafatpanah H, Rezaee A, Yousefzadeh H, et al. Immunotherapy in Allergic Rhinitis: It's Effect on the Immune System and Clinical Symptoms. Open Access Maced J Med Sci. 2018;6(7):1248-52.
  11. Bogomolov A. Diagnostic value of methods for alder (Alnus incana) allergen sensitization determination in people with respiratory allergy. Tuberculosis, Lung Diseases, HIV Infection. 2019:72-7.
  12. Nečas M. Atopy patch testing with airborne allergens. Acta Dermatovenerol Alp Pannonica Adriat. 2013;22(2):39-42.
  13. Jantunen J, Saarinen K, Rantio-Lehtimäki A. Allergy symptoms in relation to alder and birch pollen concentrations in Finland. Aerobiologia. 2011;28.
  14. Cirla AM, Sforza N, Roffi GP, Alessandrini A, Stanizzi R, Dorigo N, et al. Preseasonal intranasal immunotherapy in birch-alder allergic rhinitis: A double-blind study. Allergy. 1996;51(5):299-305.
  15. Valenta R, Breiteneder H, Pettenburger K, Breitenbach M, Rumpold H, Kraft D, et al. Homology of the major birch-pollen allergen, Bet v I, with the major pollen allergens of alder, hazel, and hornbeam at the nucleic acid level as determined by cross-hybridization. Journal of Allergy and Clinical Immunology. 1991;87(3):677-82.
  16. Wopfner N, Dissertori O, Ferreira F, Lackner P. Calcium-binding proteins and their role in allergic diseases. Immunol Allergy Clin North Am. 2007;27(1):29-44.
  17. Allergome. ALn g 2 2016 [11-11-2020]. Available from: http://www.allergome.org/script/dettaglio.php?id_molecule=8.
  18. Becker S, Gröger M, Jakob T, Klimek L. The benefit of molecular diagnostics in allergic rhinitis. Allergo Journal International. 2017;26(8):301-10.
  19. Breiteneder H, Ferreira F, Reikerstorfer A, Duchene M, Valenta R, Hoffmann-Sommergruber K, et al. Complementary DNA cloning and expression in Escherichia coli of Aln g I, the major allergen in pollen of alder (Alnus glutinosa). J Allergy Clin Immunol. 1992;90(6 Pt 1):909-17.
  20. Hayek B, Vangelista L, Pastore A, Sperr WR, Valent P, Vrtala S, et al. Molecular and immunologic characterization of a highly cross-reactive two EF-hand calcium-binding alder pollen allergen, Aln g 4: structural basis for calcium-modulated IgE recognition. J Immunol. 1998;161(12):7031-9.
  21. Ebner C, Ferreira F, Hoffmann K, Hirschwehr R, Schenk S, Szépfalusi Z, et al. T cell clones specific for Bet v I, the major birch pollen allergen, crossreact with the major allergens of hazel, Cor a I, and alder, Aln g I. Mol Immunol. 1993;30(15):1323-9.
  22. Kleine-Tebbe J. Bet v 1 and its Homologs: Triggers of Tree-Pollen Allergy and Birch Pollen-Associated Cross-Reactions. 2017. p. 21-42.