Whole Allergen
t206 Chestnut
t206 Chestnut Scientific Information
Type:
Whole Allergen
Display Name:
Chestnut
Allergen code:
t206
Family:
Fagaceae
Species:
C. sativa
Route of Exposure:
Inhalation
Latin Name:
Castanea sativa
Other Names:
Chestnut, Sweet chestnut, European chestnut, Spanish chestnut
Summary
Chestnut trees (Castanea spp.) are distributed across the Northern hemisphere, particularly in Spain and the wider Mediterranean region. Chestnut pollen is produced in large quantities and disperses over long distances. Castanea sativa pollen is a recognized allergen causing rhinoconjunctivitis and asthma. Chestnut pollen is part of the birch-homologous group of allergens, although recent evidence for cross-reactivity with other tree pollens is inconclusive.
Allergen
Nature
Chestnut trees are members of the Fagaceae family, which includes oaks (Quercus spp.) and beeches (Fagus spp.). The sweet chestnut, Castanea sativa var. Miller, is common in deciduous forests throughout Europe, especially in Spain and the wider Mediterranean region (1, 2). Chestnut trees are widely distributed in the Northern hemisphere, from northwest Africa to southwest Asia (2, 3).
Castanea pollen is wind-borne and disperses over long distances due to its small size (2). Pollen production is very high, with 10–12 billion pollen grains per tree released during the pollen season (1). The European chestnut pollen season occurs from June to August, with a peak in late June/early July (4). In South Korea, the chestnut pollen season varies between late April and July according to region (3).
Castanea pollen constitutes 5-7% of annual pollen concentrations in Spain, where high pollen counts correlate with high air temperature (1).
Taxonomy
Taxonomic tree of Castanea species (5)
Domain
Eukaryota
Kingdom
Plantae
Phylum
Spermatophyta
Subphylum
Angioospermae
Class
Dicotyledonae
Family
Fagaceae
Genus
Castanea
Tissue
Tree pollen
Epidemiology
Worldwide distribution 
Allergy to chestnut pollen represents a major cause of pollinosis in sub-Mediterranean areas (6).
Environmental Characteristics
Worldwide distribution 
Chestnut trees are used in agroforestry, for charcoal production and for ornamental purposes (CABI 2021). In Spain, chestnut trees are economically important as a source of pollen for region-specific honey (1).
Castanea sativa produces edible fruits (chestnuts) which may be consumed whole, or ground to produce chestnut flour used for baking and in animal feed (CABI 2021). China and Korea are among the world’s largest producers of chestnuts, with Italy, Spain, Portugal, France and Greece the principal European producers (LOINC 2021).
Route of Exposure
Main
The main route of exposure is inhalation.
Detection
Chestnut pollen may be sampled using a vacuum air sampler such as the Burkard spore trap (3).
Clinical Relevance
Chestnut pollen allergy is associated with allergic rhinoconjunctivitis and asthma (4).
Prevention and Therapy
Allergen immunotherapy
Immunotherapy with the reference allergen Bet v 1 (birch pollen) induced highly variable repertoires of cross-reactive IgG1 and IgG4 antibodies to other members of the birch-homologous group, including chestnut pollen (7). Current data suggest that chestnut pollen allergy does not show sufficient immunological cross-reactivity to respond successfully to immunotherapy (8).
Prevention strategies 
Avoidance
Allergenic pollen calendars are a simple method for forecasting chestnut pollen counts, as demonstrated using data from eight collection stations in South Korea over 10 years (3). Artificial neural networks (a type of computational learning) have proved to be a valid tool for predicting Castanea pollen concentration for one day ahead and, less reliably, for two and three days ahead of exposure (1).
Molecular Aspects
Allergenic molecules
Allergenic molecules of Castanea sativa (adapted from EAAC Molecular Allergology User’s Guide 2016) (9)
Allergen name
Biochemical name
Cas s 1
PR-10, Bet v 1 family member
Cas s 5
Chitinase
Cas s 8
Non-specific lipid transfer protein type 1
Cas s 9
Cytosolic class 1 small heat shock protein
Cas s 1 is the major pollen allergen of Castanea sativa. The major pollen allergens of the Fagales order belong to the pathogenesis-related protein class 10 (PR-10), which includes a large group of aeroallergens (10). All major allergens of the birch-homologous group of pollens share structural homology and biochemical properties with the major birch pollen allergen, Bet v 1 (4, 7).
Cross-reactivity
The European Medicines Agency includes Castanea sativa in the birch-homologous group of tree pollens. The major allergen of chestnut pollen (Cas s 1) demonstrates a 75% (N-terminal) amino acid sequence homology with Bet v 1, the major allergen of birch pollen (4). The earlier pollen seasons for hazel and alder may act to prime patients for birch pollen allergy, followed by allergic responses to beech and chestnut pollen (4).
In a study of 17 Bet v 1-sensitized, birch pollen-allergic patients, all subjects displayed IgE reactivity with the most common isoforms of Cas s 1 (7). However, a larger study examining 397 birch-allergic patients before and after treatment with standardized quality tree sub-lingual immunotherapy (SLIT) tablets did not find significant IgE or IgG4 cross-reactivity to chestnut, suggesting that chestnut pollen is not immunologically part of the birch-homologous group (8).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer
Last reviewed:January 2022
References
  1. Astray G, Fernández-González M, Rodríguez-Rajo FJ, López D, Mejuto JC. Airborne castanea pollen forecasting model for ecological and allergological implementation. Sci Total Environ. 2016;548-549:110-21.
  2. López-Orozco R, Hernández-Ceballos MÁ, Galán C, García-Mozo H. Atmospheric Pathways and Distance Range Analysis of Castanea Pollen Transport in Southern Spain. Forests. 2020;11(10):1092.
  3. Shin JY, Han MJ, Cho C, Kim KR, Ha JC, Oh JW. Allergenic Pollen Calendar in Korea Based on Probability Distribution Models and Up-to-Date Observations. Allergy Asthma Immunol Res. 2020;12(2):259-73.
  4. Biedermann T, Winther L, Till SJ, Panzner P, Knulst A, Valovirta E. Birch pollen allergy in Europe. Allergy. 2019;74(7):1237-48.
  5. CABI. Castanea (chestnut) Wallingford, UK2021 [cited 2021 15.11.21]. Available from: https://www.cabi.org/isc/datasheet/16586.
  6. Hirschwehr R, Jäger S, Horak F, Ferreira F, Valenta R, Ebner C, et al. Allergens from birch pollen and pollen of the European chestnut share common epitopes. Clin Exp Allergy. 1993;23(9):755-61.
  7. Grilo JR, Kitzmüller C, Aglas L, Sánchez Acosta G, Vollmann U, Ebner C, et al. IgE-cross-blocking antibodies to Fagales following sublingual immunotherapy with recombinant Bet v 1. Allergy. 2021;76(8):2555-64.
  8. Würtzen PA, Grønager PM, Lund G, Gupta S, Andersen PS, Biedermann T, et al. Simplified AIT for allergy to several tree pollens-Arguments from the immune outcome analyses following treatment with SQ tree SLIT-tablet. Clin Exp Allergy. 2021;51(2):284-95.
  9. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  10. Asam C, Hofer H, Wolf M, Aglas L, Wallner M. Tree pollen allergens-an update from a molecular perspective. Allergy. 2015;70(10):1201-11