Manuel, an 18-year-old with a history of rhino-conjunctivitis, visit his healthcare provider after developing oral symptoms, throat thickening, and urticaria after eating a piece of cake containing hazelnuts.
Manuel's healthcare provider conducts a full clinical history and physical examination and decides to test using components.
A boy with a history of rhino-conjunctivitis develops oral symptoms and urticaria after eating a piece of cake.
| Test | Type | Manuel's Results |
| Olive pollen | Whole Allergen | +5 |
| Peach | Whole Allergen | +3 |
Manuel had a negative skin prick test to grass, mugwort, dog, cat, and mite.
| Test | Type | Manuel's Results |
| Olive pollen | Whole Allergen | 28.5 |
| Peach | Whole Allergen |
8.1 |
Based on history, evaluation, and testing their previous healthcare provider recommended that Manuel use antihistamines during spring/summer and to avoid peaches.
These results together with this patient's case history and symptoms, help confirm the diagnosis.
| Test | Type | Manuel's Results |
| Olive | Whole Allergen | 35.4 |
| Peach | Whole Allergen | 12.9 |
| Hazelnut | Whole Allergen | 3.1 |
| Cor a 1 | Allergen Component | <0.1 |
| Cor a 8 | Allergen Component | 3.5 |
| Cor a 9 | Allergen Component | <0.1 |
| Cor a 14 | Allergen Component | <0.1 |
| Pru p 3 | Allergen Component | 11.5 |
| Ole e 1 | Allergen Component | <0.1 |
| Ole e 7 | Allergen Component | 21.2 |
| Ole e 9 | Allergen Component | 2.7 |
The presence of IgE antibodies to Cor a 8 confirms that Manuel’s hazelnut allergy is lipid transfer protein (LTP) dependent. He has no detectable IgE antibodies to storage proteins, but his LTP-dependent sensitization to hazelnut shows that he runs a risk of even more severe reactions than urticaria. He may experience reactions to other nuts and fruits that contain LTP, e.g., walnuts and apples.1,2
REFINED DIAGNOSIS
HEALTHCARE PROVIDER MANAGEMENT PLAN
FOLLOW UP
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.