Search
Thermo Scientific Arctic and Glacier Series refrigerated bath circulators offer outstanding cooling power, expansive temperature ranges, powerful force/suction pumps, and sophisticated digital control technology designed to ensure accuracy and reproducibility of your liquid temperature control procedures. The stainless steel reservoir is offered in multiple capacities with a variety of reservoir openings and depth dimensions for maximum application flexibility.
→ Temperature range -50 to 200°C
→ Pump capacity, 24 lpm
→ Extensive programming/ communication options
→ Available in 3 bath sizes
→ Temperature range -40 to 200°C
→ Pump capacity 20 lpm
→ UL Listed series on 115V models
→ Available in 6 bath sizes
→ Temperature range -25 to 150°C
→ Pump capacity, 17 lpm
→ Smallest, most compact series
→ Available in 6 bath sizes
*Make sure you consider cooling capacity too, especially if you're looking to circulate
Advanced | Standard | |||||
| Temperature Stability (°C) | -10 to 100 | -10 to 100 | ||||
| Cooling Capacity (click to learn more) | 240W | 240W | ||||
| Maximum Flow Rate (lpm at 0 mbar/psi) | 20 | 17 | ||||
| Maximum Pressure (mbar/psi at ) lpm) | 475/6.89 | 300/4.35 | ||||
| Communication Options | ||||||
| USB port | Optional | Optional | ||||
| Remote Sensor Port | ✓ | -- | ||||
| Ramp programs | Up to 1 | -- | ||||
| Net Weight (kg/lb) | 27.5/60.6 | 27.5/60.6 | ||||
| Compliance | CE/ROHS/WEEE?UL | CE/ROHS/WEEE | ||||
| Languages | English, German, French, Spanish, Italian | English, German, French, | ||||
| Bath Model | Volume | 115/60 | 230/50 | 100/50-60 | 115/60 | 230/50 | 100/50-60 |
| A10 | 6 Liters | 1555108 | 1555101 | 1555106 | 1525108 | 1525101 | 1525106 |
| A10 | 6 Liters with USB Port | 1565108 | 1565101 | 1565106 | 1535108 | 1535101 | 1535106 |
| A10B | 30 Liters | 1554108 | 1554101 | 1554106 | 1524108 | 1524101 | 1524106 |
| A10B | 30 Liters with USB Port | 1564108 | 1564101 | 1564106 | 1534108 | 1534101 | 1534106 |
Premium | Advanced | Standard | |||||||
| Temperature Range (°C) | -25 to 200 | -25 to 150 or 200 USB provides 200° range | -25 to 100 or 150 USB provides 150° range | ||||||
| Cooling Capacity (click to learn more) | 500W | 500W | 500W | ||||||
| Maximum Flow Rate (lpm at 0 mbar/psi) | 24 | 20 | 17 | ||||||
| Maximum Pressure (mbar/ psi at lpm) | 560/8.12 | 475/6.89 | 300/4.35 | ||||||
| Communication Options | |||||||||
| USB port | ✓ | Optional | Optional | ||||||
| Remote Sensor Port | ✓ | ✓ | -- | ||||||
| Ramp programs | Up to 10 | Up to 1 | -- | ||||||
| Net Weight (kg/lb) | 36.1/79.5 | 36.1/79.5 | 36.1/79.5 | ||||||
| Compliance | CE/ROHS/WEEE | CE/ROHS/WEEE/UL | CE/ROHS/WEEE | ||||||
| Languages | English, German, French,Spanish, Italian Chinese, Japanese | English, German, French, Spanish, Italian | English, German, French | ||||||
| Bath Model| Volume | 115/60 | 230/50 | 100/ 50-60 | 115/60 | 230/50 | 100/ 50-60 | 115/60 | 230/50 | 100/ 50-60 |
| A25|12 Liters | --- | --- | --- | 1555258 | 1555251 | 1555256 | 1525258 | 1525251 | 1525256 |
| A25|12 Liters with USB Port | 1575258 | 1575251 | 1575256 | 1565258 | 1565251 | 1565256 | 1535258 | 1535251 | 1535256 |
| A25 |21 Liters | --- | --- | --- | 1554258 | 1554251 | 1554256 | 1524258 | 1524251 | 1524256 |
| A25B|21 Liters with USB Port | --- | --- | --- | 1564258 | 1564251 | 1564256 | 1534258 | 1534251 | 1534256 |
| CE/ROHS/WEEE, UL |
| CE/ROHS/WEEE, UL |
| English, German, French, Spanish, Italian Chinese, Japanese |
PC200 | AC200 | SC150L | |||||||||
| Temperature Stability (°C) | -45 to 200 | -45 to 200 | -28 to 200 | ||||||||
| Cooling Capacity (click to learn more) | 900W | 900W | 900W | ||||||||
| Maximum Flow Rate (lpm at 0 mbar/psi) | 24 | 24 | 24 | ||||||||
| Maximum Pressure (mbar/psi at ) lpm) | 560/8.12 | 560/8.12 | 560/8.12 | ||||||||
| Communication Options | |||||||||||
| USB port | ✓ | ✓ | ✓ | ||||||||
| Remote Sensor Port | ✓ | ✓ | |||||||||
| Ramp programs | Up to 10 | Up to 1 | N/A | ||||||||
| Net Weight (kg/lb) | 55/122/5 | 55/122/5 | 55/122/5 | ||||||||
| Compliance | CE/ROHS/WEEE | CE/ROHS/WEEE | CE/ROHS/WEEE | ||||||||
| Languages | English, German, French, Spanish, Italian, Chinese | English, German, French, Spanish, Italian | English, German, French | ||||||||
| Bath Model | Volume | 115/60 | 230/50 | 100/50-60 | 115/60 | 230/50 | 100/50-60 | 115/60 | 230/50 | 100/50-60 | ||
| A40 | 12 Liters | 157-5408-HC | 157-5401-HC | 157-5406-HC | 156-5408-HC | 156-5401-HC | 156-5406-HC | 154-5408-HC | 154-5401-HC | 154-5406-HC | ||
ULT 80ZT | ULT 80 | ||
| Temperature Stability (°C) | -80 to +80 | -80 to +10 | |
| Cooling Capacity (click to learn more) | 60Hz: 250W 50Hz: 200W | 60Hz: 250W 50Hz: 200W | |
| Maximum Flow Rate (lpm at 0 mbar/psi) | 50Hz & 60Hz: 10 lpm | 50Hz & 60Hz: 10 lpm | |
| Maximum Pressure (mbar/psi at ) lpm) | 60 Hz: 359 mbar (5.2 psi) | 50Hz: 329 mbar (4.77 psi) | |
| Communication Options | |||
| USB port | -- | -- | |
| Remote Sensor Port | ✓ | ✓ | |
| Ramp programs | -- | -- | |
| Net Weight (kg/lb) | 152.4/336 | 152.4/336 | |
| Compliance | CE/RoHS/WEEE | CE/RoHS/WEEE | |
| Languages | English | English | |
| Bath Model |Volume | 220/50 | 230/60 | 220/50 |
| ULT80 | 15 Liters | 180106201601 | 180104201601 | 180104201600 |
ULT 95 | ||
| Temperature Stability (°C) | -90 to -30 | |
| Cooling Capacity (click to learn more) | 60Hz: 350W 50Hz: 280W | |
| Maximum Flow Rate (lpm at 0 mbar/psi) | 60Hz: 16 lpm 50Hz: 12.4 lpm | |
| Maximum Pressure (mbar/psi at ) lpm) | 60Hz: 927 mbar (13.44 psi) 50Hz: 628 mbar (9.1 psi) | |
| Communication Options | ||
| USB port | -- | |
| Remote Sensor Port | ✓ | |
| Ramp programs | -- | |
| Net Weight (kg/lb) | 168/370 | |
| Compliance | CE/RoHS/WEEE | |
| Languages | English | |
| Bath Model |Volume | 220/50 | 230/60 |
| ULT95 | 15 Liters | 179106241600 | 179104241600 |
Advanced | Standard | |||||
| Temperature Stability (°C) | -10 to 100 | -10 to 100 | ||||
| Cooling Capacity (click to learn more) | 240W | 240W | ||||
| Maximum Flow Rate (lpm at 0 mbar/psi) | 20 | 17 | ||||
| Maximum Pressure (mbar/psi at ) lpm) | 475/6.89 | 300/4.35 | ||||
| Communication Options | ||||||
| USB port | Optional | Optional | ||||
| Remote Sensor Port | ✓ | -- | ||||
| Ramp programs | Up to 1 | -- | ||||
| Net Weight (kg/lb) | 27.5/60.6 | 27.5/60.6 | ||||
| Compliance | CE/ROHS/WEEE?UL | CE/ROHS/WEEE | ||||
| Languages | English, German, French, Spanish, Italian | English, German, French, | ||||
| Bath Model | Volume | 115/60 | 230/50 | 100/50-60 | 115/60 | 230/50 | 100/50-60 |
| A10 | 6 Liters | 1555108 | 1555101 | 1555106 | 1525108 | 1525101 | 1525106 |
| A10 | 6 Liters with USB Port | 1565108 | 1565101 | 1565106 | 1535108 | 1535101 | 1535106 |
| A10B | 30 Liters | 1554108 | 1554101 | 1554106 | 1524108 | 1524101 | 1524106 |
| A10B | 30 Liters with USB Port | 1564108 | 1564101 | 1564106 | 1534108 | 1534101 | 1534106 |
Premium | Advanced | Standard | |||||||
| Temperature Range (°C) | -25 to 200 | -25 to 150 or 200 USB provides 200° range | -25 to 100 or 150 USB provides 150° range | ||||||
| Cooling Capacity (click to learn more) | 500W | 500W | 500W | ||||||
| Maximum Flow Rate (lpm at 0 mbar/psi) | 24 | 20 | 17 | ||||||
| Maximum Pressure (mbar/ psi at lpm) | 560/8.12 | 475/6.89 | 300/4.35 | ||||||
| Communication Options | |||||||||
| USB port | ✓ | Optional | Optional | ||||||
| Remote Sensor Port | ✓ | ✓ | -- | ||||||
| Ramp programs | Up to 10 | Up to 1 | -- | ||||||
| Net Weight (kg/lb) | 36.1/79.5 | 36.1/79.5 | 36.1/79.5 | ||||||
| Compliance | CE/ROHS/WEEE | CE/ROHS/WEEE/UL | CE/ROHS/WEEE | ||||||
| Languages | English, German, French,Spanish, Italian Chinese, Japanese | English, German, French, Spanish, Italian | English, German, French | ||||||
| Bath Model| Volume | 115/60 | 230/50 | 100/ 50-60 | 115/60 | 230/50 | 100/ 50-60 | 115/60 | 230/50 | 100/ 50-60 |
| A25|12 Liters | --- | --- | --- | 1555258 | 1555251 | 1555256 | 1525258 | 1525251 | 1525256 |
| A25|12 Liters with USB Port | 1575258 | 1575251 | 1575256 | 1565258 | 1565251 | 1565256 | 1535258 | 1535251 | 1535256 |
| A25 |21 Liters | --- | --- | --- | 1554258 | 1554251 | 1554256 | 1524258 | 1524251 | 1524256 |
| A25B|21 Liters with USB Port | --- | --- | --- | 1564258 | 1564251 | 1564256 | 1534258 | 1534251 | 1534256 |
| CE/ROHS/WEEE, UL |
| CE/ROHS/WEEE, UL |
| English, German, French, Spanish, Italian Chinese, Japanese |
PC200 | AC200 | SC150L | |||||||||
| Temperature Stability (°C) | -45 to 200 | -45 to 200 | -28 to 200 | ||||||||
| Cooling Capacity (click to learn more) | 900W | 900W | 900W | ||||||||
| Maximum Flow Rate (lpm at 0 mbar/psi) | 24 | 24 | 24 | ||||||||
| Maximum Pressure (mbar/psi at ) lpm) | 560/8.12 | 560/8.12 | 560/8.12 | ||||||||
| Communication Options | |||||||||||
| USB port | ✓ | ✓ | ✓ | ||||||||
| Remote Sensor Port | ✓ | ✓ | |||||||||
| Ramp programs | Up to 10 | Up to 1 | N/A | ||||||||
| Net Weight (kg/lb) | 55/122/5 | 55/122/5 | 55/122/5 | ||||||||
| Compliance | CE/ROHS/WEEE | CE/ROHS/WEEE | CE/ROHS/WEEE | ||||||||
| Languages | English, German, French, Spanish, Italian, Chinese | English, German, French, Spanish, Italian | English, German, French | ||||||||
| Bath Model | Volume | 115/60 | 230/50 | 100/50-60 | 115/60 | 230/50 | 100/50-60 | 115/60 | 230/50 | 100/50-60 | ||
| A40 | 12 Liters | 157-5408-HC | 157-5401-HC | 157-5406-HC | 156-5408-HC | 156-5401-HC | 156-5406-HC | 154-5408-HC | 154-5401-HC | 154-5406-HC | ||
ULT 80ZT | ULT 80 | ||
| Temperature Stability (°C) | -80 to +80 | -80 to +10 | |
| Cooling Capacity (click to learn more) | 60Hz: 250W 50Hz: 200W | 60Hz: 250W 50Hz: 200W | |
| Maximum Flow Rate (lpm at 0 mbar/psi) | 50Hz & 60Hz: 10 lpm | 50Hz & 60Hz: 10 lpm | |
| Maximum Pressure (mbar/psi at ) lpm) | 60 Hz: 359 mbar (5.2 psi) | 50Hz: 329 mbar (4.77 psi) | |
| Communication Options | |||
| USB port | -- | -- | |
| Remote Sensor Port | ✓ | ✓ | |
| Ramp programs | -- | -- | |
| Net Weight (kg/lb) | 152.4/336 | 152.4/336 | |
| Compliance | CE/RoHS/WEEE | CE/RoHS/WEEE | |
| Languages | English | English | |
| Bath Model |Volume | 220/50 | 230/60 | 220/50 |
| ULT80 | 15 Liters | 180106201601 | 180104201601 | 180104201600 |
ULT 95 | ||
| Temperature Stability (°C) | -90 to -30 | |
| Cooling Capacity (click to learn more) | 60Hz: 350W 50Hz: 280W | |
| Maximum Flow Rate (lpm at 0 mbar/psi) | 60Hz: 16 lpm 50Hz: 12.4 lpm | |
| Maximum Pressure (mbar/psi at ) lpm) | 60Hz: 927 mbar (13.44 psi) 50Hz: 628 mbar (9.1 psi) | |
| Communication Options | ||
| USB port | -- | |
| Remote Sensor Port | ✓ | |
| Ramp programs | -- | |
| Net Weight (kg/lb) | 168/370 | |
| Compliance | CE/RoHS/WEEE | |
| Languages | English | |
| Bath Model |Volume | 220/50 | 230/60 |
| ULT95 | 15 Liters | 179106241600 | 179104241600 |
Cooling capacity is a specification that defines how much heat energy can be removed from an application at a given temperature.
Specifications obtained at sea level using water (above 5°C to 90°C) or a fluid with a specific heat of 2.3 kJ/kg-K or 0.55 Btu/lb-F (less than 5°C) as the recirculating fluid at a 20°C ambient condition, at nominal operating voltage. Other fluids, process temperatures, ambient temperatures, altitude or operating voltage will affect performance. Specifications are for reference only and are subject to change.
Our Arctic and Glacier refrigerated bath circulators are versatile - used in labs and production settings all over the world. As we come across common applications, we learn more about what our customers really need to know. A few common applications we've learned a thing or two about include:
Rotary Evaporators: 
When speed and accuracy are critical
Learn more ›
But since these baths are used in nearly any research or production setting, we can tell you - there are some common questions amongst all labs. So we've created some easy ways to digest seemingly complex stuff - so you can get back to your work, fast. Some quick smart notes worth taking a glance:
Whether your temperature control unit (TCU) is a water bath, refrigerated/ heated bath circulator, cooling/heating recirculating chiller or recirculating chiller, a fluid will have to be selected. If you are operating between approximately 5 ºC and 95 ºC, then water seems the likely choice. You might find yourself close to a source of tap water, filtered water, distilled water, reverse osmosis (RO) water, or deionized (DI) water. However, just using the nearest convenient source may not be the right choice. While these water types all have their uses, not all are suitable for your TCU needs. Regardless of how it has been treated, all water starts out with the same basic properties, advantages and disadvantages.
No. A large bath reservoir is not an indication of increased cooling capacity or better temperature stability. Cooling capacity is based on setpoint temperature and compressor size. Temperature stability is affected by two things: the consistency of the heat load from your application, and how precisely the heating and/or cooling is controlled by the bath circulator.
The best selection for you would be a bath circulator with the smallest possible bath reservoir that still meets your setpoint temperature and cooling capacity requirements. This will lower the load when changing from one setpoint to another, improving your time to temperature.
A recirculating chiller is used to supply a steady flow of temperature-controlled fluid to an application. The fluid temperature coming from the chiller could be used to cool, maintain or even increase the temperature of the application. Five criteria must be defined in order to select the correct refrigeration, circulation, and control systems for your recirculating chiller.
Selecting energy efficient components does help with overall energy efficiency. However, the most efficient refrigerated bath circulators control temperature differently compared to standard refrigerated bath circulators. Through a special energy savings mode, they are able to further reduce the power usage under low demand situations without reducing performance during high demand operations.
First calculate the annual water usage and the cost of purchasing and disposing of that water. Then calculate the cost of purchasing and operating a chiller or circulator. Depending on what you pay for water, sewer and electricity in your community, you may be surprised how little a chiller or circulator actually costs. In fact, you may discover that using a chiller or circulator may save you money and pay for itself over time.
The VersaCool's headless, coil-less design gives more space for samples—and for everything else on your workbench. Bluetooth and USB connections allow remote operation, and the global voltage feature automatically detects the right voltage and frequency.
We provide service solutions to the complete portfolio of Thermo Scientific temperature control products. Don’t have a Thermo Scientific temperature control system? Don’t worry! We support most other manufacturers’ equipment as well. Contact us ›