Search
Host cell proteins (HCPs) are low-level, process-related protein impurities in drug products derived from the host organism during biotherapeutic manufacturing. During expression of a recombinant protein drug, host cell systems can express many endogenous proteins. Purification of the drug from these HCP contaminants can be challenging, with low-level contamination remaining after purification. Host cell proteins (HCPs) accompanied with recombinant biotherapeutics can significantly affect drug efficacy and cause immunogenicity. Detection and quantification of residual HCPs as potential process-related impurities is critical for biopharmaceutical companies in accordance with regulatory agency guidelines (ICH Q6B).
| Orbitrap Exploris 480 Mass Spectrometer | Orbitrap Exploris 240 Mass Spectrometer | |
|---|---|---|
![]() | ![]() | |
| Advantages | Extraordinary simplified within a compact footprint where next level exploration delivers quantifiable accuracy and the selectivity required to comprehensively identify and quantify host cell protein contaminants. | Realize complete BioPharma characterization and identify, quantify and confirm more host cell proteins rapidly and with confidence. |
| Resolving power | 480,000 at m/z 195 | Up to 240,000 at m/z 200 |
| Scan speed | Up to 40Hz | 22Hz |
| Mass range | 40 to 6,000 m/z (up to 8,000 m/z with the BioPharma option) | 4 to 8,000 m/z |
| Dynamic range | >5000:1 | >5000:1 |
| Mass accuracy | Internal <1 ppm RMS; External: <3 ppm RMS | Internal <1 ppm RMS; External: <3 ppm RMS |
Use Thermo Scientific Proteome Discoverer software to access multiple database search algorithms (SEQUEST, ZCore, Mascot, etc.) to confidently identify production related protein impurities down to low PPM levels and validate protein IDs with false discovery rate (FDR) determination.
Use store and manage libraries for all database searching requirements, including custom libraries based on your cell lines.
Learn how you can achieve confident, sensitive and reproducible AAVHCP identification and sub-ppm quantification using a novel HRAM platform with increased throughput.
The Thermo Scientific AccelerOme automated sample preparation platform, with factory-supplied reagents and kits, intuitive software and a unique experimental design experience transforms proteomics sample preparation.
The LC-MS are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our Products have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in inaccurate results and, more seriously, health and safety risks. They are designated for Research Use Only Applications.
General Laboratory Equipment – Not For Diagnostic Procedures. © 2026 Thermo Fisher Scientific Inc. All rights reserved. Thermo Scientific™ products are designated as General Laboratory Equipment and are not intended for use in diagnostic procedures or for in vitro diagnostic purposes. These products are intended for general laboratory use only and have not been tested or validated for diagnostic applications; use for such purposes may result in inaccurate results and may pose health and safety risks. They are designed for research, academic, and industrial applications and for general-purpose analytical use, including high-resolution and accurate-mass measurements, without reference to specific clinical conditions, analytes, or diagnostic decisions.
The Thermo Scientific Orbitrap Exploris and Excedion series mass spectrometers are Orbitrap instruments with an atmospheric pressure ionization (API) source for liquid chromatography (LC) mass spectrometry (MS) high-throughput applications. The instruments are designed to be placed on a bench in the laboratory and maintain mass accuracy across chromatographic peaks, supporting confident identification and quantitation using high-resolution accurate-mass detection.