Search
Exosomes are small (30–150 nm) vesicles containing sophisticated RNA and protein cargo, that are constantly secreted by all cells in vitro and in vivo. Exosomes are changing research due to the intriguing functions within the human body, including intercellular communication and signaling. These extracellular vesicles are growing, both in understanding their biological functions and using them towards practical applications such as development of non-invasive diagnostics and advanced therapeutics.
Dynabeads offers an array of exosome isolation products and tools for characterization and analysis of their RNA and protein content, as well as in vitro and in vivo tracing to help you further the understanding of these fascinating entities. Visit our storefront for other applications using Dynabeads Magnetic Beads.
Ultracentrifugation protocols can be tedious and time-consuming. Now it is possible to isolate intact exosomes from cell culture media or any body fluid using the flexible and scalable Total Exosome Isolation reagents. These products and accompanying protocols are ideal for a wide range of experiments, including processing low sample volumes and handling multiple samples.
Total exosomes enriched from cell culture (by using the Total Exosome Isolation reagents or ultracentrifugation) can be further purified into specific subpopulations by immunomagnetic capture. Use the Dynabeads-based CD9, CD63, CD81, or EpCam-specific reagent, or combine the Streptavidin reagent with your choice of a biotinylated antibody to purify any population based on a surface antigen.
Analysis of exosomes is very challenging due to their small size of 30-150 nm. There are a few methods, though, which can be used for intact exosome analysis.
Exosomes have been shown to transport a range of molecules between different cell types. Their cargo includes various proteins, mRNA (fragments and full length), rRNA, tRNA, miRNA, and ncRNA.
Gibco exosome-depleted FBS has the highest level of exosome depletion compared to homebrewed material and competitor FBS while retaining the vital nutrients necessary for cell culture. The work of Gibco exosome-depleted FBS has cut inefficiency that comes with ultracentrifugation. Some of the product highlights include:
Standardization of extracellular vesicles isolation and characterization has been a highly requested need of the International Society of Extracellular Vesicles (ISEV) for several years. A highly reproducible method for pull-down and analysis was developed using the KingFisher Flex Purification System in combination with Dynabeads magnetic beads.
See how Dynabeads Exosome products are optimized on the KingFisher
Ultracentrifugation protocols can be tedious and time-consuming. Now it is possible to isolate intact exosomes from cell culture media or any body fluid using the flexible and scalable Total Exosome Isolation reagents. These products and accompanying protocols are ideal for a wide range of experiments, including processing low sample volumes and handling multiple samples.
Total exosomes enriched from cell culture (by using the Total Exosome Isolation reagents or ultracentrifugation) can be further purified into specific subpopulations by immunomagnetic capture. Use the Dynabeads-based CD9, CD63, CD81, or EpCam-specific reagent, or combine the Streptavidin reagent with your choice of a biotinylated antibody to purify any population based on a surface antigen.
Analysis of exosomes is very challenging due to their small size of 30-150 nm. There are a few methods, though, which can be used for intact exosome analysis.
Exosomes have been shown to transport a range of molecules between different cell types. Their cargo includes various proteins, mRNA (fragments and full length), rRNA, tRNA, miRNA, and ncRNA.
Gibco exosome-depleted FBS has the highest level of exosome depletion compared to homebrewed material and competitor FBS while retaining the vital nutrients necessary for cell culture. The work of Gibco exosome-depleted FBS has cut inefficiency that comes with ultracentrifugation. Some of the product highlights include:
Standardization of extracellular vesicles isolation and characterization has been a highly requested need of the International Society of Extracellular Vesicles (ISEV) for several years. A highly reproducible method for pull-down and analysis was developed using the KingFisher Flex Purification System in combination with Dynabeads magnetic beads.
See how Dynabeads Exosome products are optimized on the KingFisher
We asked 10 prominent scientists to share their thoughts on exosomes research.