Total Exosome Isolation Kit (from plasma)
Invitrogen™

Total Exosome Isolation Kit (from plasma)

トータルエキソソーム単離試薬(血漿用)は、血漿用サンプルからインタクトなエキソソームを迅速かつ効率的に濃縮します。このキットは、トータルエキソソーム単離試薬(血漿用)およびプロテイナーゼK(20 mg/mL詳細を見る
製品番号(カタログ番号)数量
44844506 mL
製品番号(カタログ番号) 4484450
価格(JPY)
65,900
Each
数量:
6 mL
トータルエキソソーム単離試薬(血漿用)は、血漿用サンプルからインタクトなエキソソームを迅速かつ効率的に濃縮します。このキットは、トータルエキソソーム単離試薬(血漿用)およびプロテイナーゼK(20 mg/mL)の2つから成ります。トータルエキソソーム単離試薬(血漿用)は、サンプルサイズに合わせてスケールアップ、スケールダウンが可能なプロトコルで、血漿サンプルからインタクトなエキソソームを簡単かつ確実に濃縮します。プロテアーゼKは、任意で行う血漿の前処理用に別個に提供されます。プロテアーゼKを使用して血漿タンパク質をすべて除去することで、最高純度のエキソソームが得られます。

•あらゆる種類の下流アプリケーションで使用できるインタクトなエキソソームの回収率を最大化
• シンプルかつ信頼性の高いプロトコルで簡単にエキソソームを分離
• 時間のかかる超遠心分離は不要
• 最大限の柔軟性—サンプルサイズに合わせてスケールアップまたはスケールダウンが可能

トータルエキソソーム単離試薬は、水分子を結合させることにより、小胞など溶解性の低い成分を溶液外に押し出し、短時間の低速遠心による回収を可能にします。プロテイナーゼKで10分間処理した後、試薬を血漿に加え、溶液を2–8℃で30分間インキュベートします。沈殿したエキソソームを室温で10,000 x g、5分間の標準的な遠心分離で回収します。次いでペレットをPBSまたは同様のバッファーに再懸濁すれば、エキソソームを下流の解析またはアフィニティー法による次段階の精製で使用できるようになります。その後、トータルエキソソームRNAタンパク質単離キット(カタログ番号4478545)を使用してトータルRNAとタンパク質を精製できます。インタクトなエキソソームを経路、機能、輸送などの生物学的研究に使用することもできます。

下流の質量分析では、分析前にゲル電気泳動による単離エキソソーム懸濁液の精製を行うことを推奨します。
研究用にのみ使用できます。診断用には使用いただけません。
仕様
最終産物タイプエキソソーム
フォーマット液体
数量6 mL
製品タイプTotal Exosome Isolation Kit
Unit SizeEach
組成および保存条件
• 6 mLのトータルエキソソーム沈殿試薬(血漿用)。2~8℃で保存してください。
• トータルエキソソーム単離(血漿用)(20 mg/mL)に使用する1.25 mLのプロテイナーゼK。-5~-30℃で保存。

よくあるご質問(FAQ)

What is the best way to store my exosomes?

For the short-term, exosomes can be stored at 4 degrees C for up to 1 week. For the long-term, exosomes can be stored at -20 degrees C or -80 degrees C. When storing exosomes for the long term, it is important to consider whether they will need to be thawed more than once for the target application. If multiple applications (and thus multiple thaws) will be used for analysis, then we recommend aliquoting the exosome resuspensions into multiple tubes so that each tube will only undergo one freeze/thaw cycle. We have found that multiple freeze thaw cycles can cause damage to the exosomes and reduce their numbers.

There are two protocol options for exosome isolation from plasma samples, which one should I choose?

Unlike serum, plasma contains numerous clotting factors and some additional proteins that can make it difficult to work with. We‘ve provided two protocol options, one with proteinase K (PK) and one without, in order to ease this difficulty. The protocol using PK is most useful when the end goal is analysis of the RNA or protein cargo contained inside the exosomes. It can also be used to isolate exosomes for use in other downstream applications, but it is most useful for RNA and protein analysis. The protocol without PK also isolates good quality exosomes, just not quite as pure as the PK protocol. The “no PK” protocol is more useful for isolating exosomes that will be used for surface protein analysis or electron microscopy identification.

My Westerns do not seem to work after exosome isolation. Can you help?

There are several possible reasons why Western blotting analysis is challenging:

1. Not enough sample volume added. Exosomes can contain a fairly low amount of protein cargo, so for an initial experiment we recommend adding as much of the sample as possible.
2. Antibody concentration should be titrated. Also, they should ideally be used fresh and need to be stored properly.
3. Depending on the exosomal surface marker, certain gel conditions might be more optimal for the target antibody (e.g., reducing/nonreducing and denaturing/nondenaturing). We suggest checking with the manufacturer and exosome community about which Western blotting conditions are recommended for the specific marker you are targeting and the specific antibody you are using.
4. General Western techniques. Westerns can be tricky so we recommend the use of a positive control for initial testing to make sure the entire workflow is functioning as it should. Any protein or antibody can be used as long as they meet the conditions you need (e.g., denaturing vs. non-denaturing). In addition, when picking the protein, try to steer clear of those that are present at very high or very low concentrations in your sample to prevent overloading the blot or total absence of signal.

How much RNA can be recovered from the exosomes?

This can vary depending on the sample type, volume of sample, isolation method, and exosome content/concentration. Listed below are some examples:

1) When exosomes are isolated from 30 mL of HeLa cell culture medium using the Total Exosome Isolation Reagent, it is possible to recover approximately 8 ng exosomal RNA.
2) For exosomes recovered from 4 mL serum, approximately 2 ng exosomal RNA can be obtained.

In both cases, these amounts of RNA are sufficient for RNA library prep for Ion PGM or Ion Proton sequencing. For real-time PCR analysis, substantially smaller amounts of RNA are needed and much lower sample volumes can be used. For example, RNA recovered from 3 µL serum or 30 µL medium is enough for one qRT-PCR reaction.

I'm using the Total Exosome RNA & Protein Isolation Kit. When ethanol is added to buffer 2/3, the solution turns turbid. Does this affect the efficiency of RNA recovery?

No, the described effect does not have a negative impact on the RNA recovery.