Search
Our gas chromatography (GC) solutions are designed to be customized so that they can fit any laboratory workflow. From Thermo Scientific iConnect injector and detector modules to our performance-leading Thermo Scientific mass spectrometers, we offer the largest portfolio of GC solutions that enable you to innovate and customize your gas chromatography applications.
Discover superior workflow performance with our extensive GC solutions
Our state-of-the-art GC systems are modular for unlimited combinations of iConnect injector and detector modules, which can be switched out by the user in minutes, and can be used for stand-alone chromatography or with mass spectrometry.
Mass spectrometry instruments and solutions for routine, high-throughput, research and development, environmental, pharma/biopharma, forensic toxicology, and food and beverage applications.
iConnect injector modules that swap out in just 2 minutes to offer maximum instrument uptime with off-line maintenance capability. Our GC injectors include split/splitless and programmable temperature vaporizing injectors with optional backflush capabilities and cold-on-column injectors.
iConnect detector modules that swap in just two minutes and enable maximum flexibility to meet evolving analytical requirements. Our GC detectors include thermal conductivity, flame ionization, pulsed discharge, nitrogen phosphorus, and electron capture detectors.
Different GC autosamplers are available to fit any requirement for liquid, headspace, and solid phase microextraction (SPME) injections; automated sample preparation; as well as dilutions, additions, and derivatization functions.
Featuring high temperature stability, low bleed, and long lifetimes, our proven GC columns range from polar to non-polar to specialty GC columns for a multitude of GC or GC-MS applications.
Find gas chromatography filters, connectors, tubing, ovens, reagents and standards, and injection port accessories. We also offer GC sample handling products such as derivatization kits, vials, and closures.
Improve GC or GC-MS analysis with our derivatization reagents and siliconizing fluids for various laboratory applications including environmental, food and beverage, forensic toxicology, and pharma/biopharma.
Our GC solutions include gas chromatographs, injectors, detectors, columns, autosamplers, and software to enable you to achieve success with your gas chromatography analyses.
Modern GC Autosamplers allow for unattended liquid or headspace sample injection into the GC column, increasing the sample throughput and delivering better injection repeatability compared to manual operation. Robotic autosamplers like the Thermo Scientific TriPlus RSH SMART enable a higher level of automation and versatility, offering a range of sampling techniques spanning liquid, static and dynamic headspace, and SPME, as well as sample preparation capability like dilutions, additions, and derivatization functions.
Our iConnect injectors offer maximum flexibility and performance to meet all types of routine and research analytical needs. The unique modular design allows injector modules to be swapped in less than two minutes, maximizing instrument uptime during maintenance offering a cost-saving back up in the lab while facilitating troubleshooting.
Our iConnect detectors redefine flexibility and usability in GC and GC-MS analysis. This unique capability can also enable continuous operation or dramatically reduce maintenance downtime—just replace dirty units with clean backups and be back to running samples again in a few minutes.
Featuring high temperature stability, low bleed, and long lifetimes, our proven GC columns range from polar to non-polar to specialty GC columns for a multitude of GC or GC-MS applications.
Experience an unprecedented level of quantitative and qualitative GC-MS performance easily and routinely with Thermo Scientific Orbitrap GC-MS systems. Go beyond what’s possible in your lab and redefine the frontiers of GC-MS analysis
Add the specificity and confidence of mass spectral data to a wide range of GC applications and methodologies with a Thermo Scientific single quadrupole mass spectrometer. This robust, easy-to-use system ensures laboratory productivity.
Use the gold standard for sensitive, specific quantitation of target compounds. This Thermo Scientific Triple Quadrupole GC-MS/MS system combines superb sensitivity and selectivity with outstanding productivity and reliability.
Get the performance of high-sensitivity, high-resolution magnetic sector mass spectrometry in a simple, easy-to-use package with a Thermo Scientific DFS Magnetic Sector GC-HRMS system.
Discover why our Chromeleon Data Systems offer is the all-in-one solution for chromatography instrument control and integration with other systems.
Modern GC Autosamplers allow for unattended liquid or headspace sample injection into the GC column, increasing the sample throughput and delivering better injection repeatability compared to manual operation. Robotic autosamplers like the Thermo Scientific TriPlus RSH SMART enable a higher level of automation and versatility, offering a range of sampling techniques spanning liquid, static and dynamic headspace, and SPME, as well as sample preparation capability like dilutions, additions, and derivatization functions.
Our iConnect injectors offer maximum flexibility and performance to meet all types of routine and research analytical needs. The unique modular design allows injector modules to be swapped in less than two minutes, maximizing instrument uptime during maintenance offering a cost-saving back up in the lab while facilitating troubleshooting.
Our iConnect detectors redefine flexibility and usability in GC and GC-MS analysis. This unique capability can also enable continuous operation or dramatically reduce maintenance downtime—just replace dirty units with clean backups and be back to running samples again in a few minutes.
Featuring high temperature stability, low bleed, and long lifetimes, our proven GC columns range from polar to non-polar to specialty GC columns for a multitude of GC or GC-MS applications.
Experience an unprecedented level of quantitative and qualitative GC-MS performance easily and routinely with Thermo Scientific Orbitrap GC-MS systems. Go beyond what’s possible in your lab and redefine the frontiers of GC-MS analysis
Add the specificity and confidence of mass spectral data to a wide range of GC applications and methodologies with a Thermo Scientific single quadrupole mass spectrometer. This robust, easy-to-use system ensures laboratory productivity.
Use the gold standard for sensitive, specific quantitation of target compounds. This Thermo Scientific Triple Quadrupole GC-MS/MS system combines superb sensitivity and selectivity with outstanding productivity and reliability.
Get the performance of high-sensitivity, high-resolution magnetic sector mass spectrometry in a simple, easy-to-use package with a Thermo Scientific DFS Magnetic Sector GC-HRMS system.
Discover why our Chromeleon Data Systems offer is the all-in-one solution for chromatography instrument control and integration with other systems.
A partnership with Thermo Fisher Scientific gives you the collective power of technology, methods and workflows to serve a wider range of industries and applications – ensuring you and the communities you serve are completely confident in the results.
The world needs science to move faster. Now more than ever. Which is why when more scientists have access to the latest scientific instrumentation, we all benefit. That is the sole purpose of our Lab-Forward Trade-In Program. For a limited time, trade in your chromatography and mass spectrometry instruments and save!
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.