iConnect™ Thermal Conductivity Detector (TCD) for TRACE™ 1300 and 1600 Series GC
iConnect™ Thermal Conductivity Detector (TCD) for TRACE™ 1300 and 1600 Series GC
iConnect™ Thermal Conductivity Detector (TCD) for TRACE™ 1300 and 1600 Series GC
Thermo Scientific™

iConnect™ Thermal Conductivity Detector (TCD) for TRACE™ 1300 and 1600 Series GC

Get exceptional thermal stability and extremely fast response with the Thermo Scientific™ iConnect™ Thermal Conductivity Detector (TCD) for TRACE™ 1300 and 1600 Series GC systems.

Have Questions?
Catalog NumberDetection Mode
19070040Thermal Conductivity
Catalog number 19070040
Price (USD)
6,350.00
Each
Detection Mode:
Thermal Conductivity
Price (USD)
6,350.00
Each

Get exceptional thermal stability and extremely fast response across a wide variety of capillary and packed column applications with the Thermo Scientific™ iConnect™ Thermal Conductivity Detector (TCD) for TRACE™ 1300 and 1600 Series GC systems. The micro-volume TCD is ideal as a general-purpose detector for industrial, petrochemical, and environmental determination of unknown samples.

FOR US ONLINE SALES ONLY

Thermo Scientific “instant connect” (iConnect) injectors and detectors redefine flexibility and usability in GC and GC-MS analyses. In two minutes, without special training or tools, you can swap out these modules and reconfigure your instrument for a specific workflow or method. This unique capability can also enable continuous operation or dramatically reduce maintenance downtime — just replace dirty units with clean backups and be back to running samples again in a few minutes.

 

Fast Response, Outstanding Sensitivity

Nondestructive to samples, the iConnect TCD provides exceptional sensitivity for an extremely wide range of GC applications.

  • Compatible with 1/8 in. and 1/16 in. packed columns; optimized for capillary columns
  • Maximum Temperature: 400°C in steps of 0.1°C
  • MDL: Less than 400 pg tridecane/mL with He carrier, or less than 20 pg tridecane/s with a total flow through the cell of 3 mL/min
  • Linear Dynamic Range: 105

 

Options for Expansion

For budget-conscious labs, iConnect modules offer the flexibility to start with a single-channel instrument, and expand to multi-injector or multi-detector configurations to accommodate new applications or increased throughput requirements.

Specifications
DescriptioniConnect Thermal Conductivity Detector (TCD) module
Detection ModeThermal Conductivity
Dynamic Range105
For Use With (Equipment)TRACE 1300 and 1600 Series Gas Chromatographs
QuantityEach
Detector TypeTCD
Product TypeiConnect Thermal Conductivity Detector
Temperature RangeUp to 400°C in steps of 0.1°C
Unit SizeEach