Search
Search

Entrez Gene ID: | 10153 |
Gene Name: | CCAAT/enhancer binding protein zeta |
Gene Aliases: |
CBF, CBF2, HSP-CBF, NOC1 |
Location: |
Chr.2:37201629-37231597 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPZ | NM_005760.2 | 2 | 468 | NP_005751.2 |
| AK312322.1 | 2 | 467 | ||
| BC034475.2 | 2 | 472 | AAH34475.1 | |
| M37197.1 | 2 | 449 | AAA51924.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2684 | Chr.2:37203356 - 37236290 on Build GRCh38 | Insertion |
|
Set Membership: |
|