Search
Search

Entrez Gene ID: | 1063 |
Gene Name: | centromere protein F |
Gene Aliases: |
CENF, CILD31, PRO1779, STROMS, hcp-1 |
Location: |
Chr.1:214603195-214664571 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPF | NM_016343.4 | 12 | 2578 | NP_057427.3 |
| XM_011509082.4 | 12 | 2578 | XP_011507384.1 | |
| XM_017000086.3 | 12 | 6011 | XP_016855575.1 | |
| BU155312.1 | ||||
| U30872.1 | 12 | 2482 | AAA82935.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv574n100 | Chr.1:214590435 - 214840032 on Build GRCh38 | Gain |
|
| nsv4061003 | Chr.1:214615605 - 214643867 on Build GRCh38 | Deletion |
|
Set Membership: |
|