Search
Search

Entrez Gene ID: | 407738 |
Gene Name: | TAFA chemokine like family member 1 |
Gene Aliases: |
FAM19A1, TAFA-1 |
Location: |
Chr.3:67991544-68545621 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAFA1 | NM_001252216.2 | NP_001239145.1 | ||
| NM_001438030.1 | ||||
| NM_001438647.1 | ||||
| NM_001438648.1 | ||||
| NM_001438649.1 | ||||
| NM_001438650.1 | ||||
| NM_213609.4 | NP_998774.2 | |||
| XM_017006386.2 | XP_016861875.1 | |||
| XM_017006387.3 | XP_016861876.1 | |||
| XM_017006388.2 | XP_016861877.1 | |||
| XM_017006389.2 | XP_016861878.1 | |||
| XM_017006390.2 | XP_016861879.1 | |||
| AK289668.1 | BAF82357.1 | |||
| AK290446.1 | BAF83135.1 | |||
| AL713702.1 | CAD28501.1 | |||
| AY325114.1 | AAP92406.1 | |||
| BC025746.1 | AAH25746.1 | |||
| DA485098.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003158 | Chr.3:66999779 - 68395539 on Build GRCh38 | Loss |
|
| nsv6365632 | Chr.3:68115412 - 68177920 on Build GRCh38 | Duplication |
|
| esv2752014 | Chr.3:68042729 - 68367839 on Build GRCh38 | Gain |
|
| nsv6366784 | Chr.3:66994301 - 68395700 on Build GRCh38 | Deletion |
|
| nsv4074520 | Chr.3:67792291 - 68263320 on Build GRCh38 | Duplication |
|
| nsv6366126 | Chr.3:67809067 - 68935602 on Build GRCh38 | Duplication |
|
Set Membership: |
|