Search
Search

Entrez Gene ID: | 11154 |
Gene Name: | adaptor related protein complex 4 sigma 1 subunit |
Gene Aliases: |
AP47B, CLA20, CLAPS4, CPSQ6, SPG52 |
Location: |
Chr.14:31025106-31096450 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4S1 | NM_001128126.2 | NP_001121598.1 | ||
| NM_001254728.1 | NP_001241657.1 | |||
| NM_001254729.1 | NP_001241658.1 | |||
| AB030654.1 | BAA82970.1 | |||
| AF155159.1 | AAD43329.1 | |||
| AK304115.1 | ||||
| AK316517.1 | ||||
| BF671561.1 | ||||
| CR457100.1 | CAG33381.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832763 | Chr.14:31032391 - 31210815 on Build GRCh38 | Loss |
|
| nsv1049063 | Chr.14:31004315 - 31153535 on Build GRCh38 | Gain |
|
| esv3584215 | Chr.14:31075625 - 31202337 on Build GRCh38 | Gain |
|
| esv3892495 | Chr.14:30985830 - 31116083 on Build GRCh38 | Gain |
|
| esv33189 | Chr.14:30331017 - 31442539 on Build GRCh38 | Loss |
|
| nsv1050054 | Chr.14:31032497 - 31264436 on Build GRCh38 | Gain |
|
| nsv1043033 | Chr.14:31069778 - 31217490 on Build GRCh38 | Gain |
|
Set Membership: |
|