Search
Search

Entrez Gene ID: | 8844 |
Gene Name: | kinase suppressor of ras 1 |
Gene Aliases: |
KSR, RSU2 |
Location: |
Chr.17:27456448-27626435 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758683 | Chr.17:27551461 - 27850093 on Build GRCh38 | Gain |
|
| esv2422193 | Chr.17:27479927 - 27623674 on Build GRCh38 | Duplication |
|
Set Membership: |
|