Search
Search

Entrez Gene ID: | 10149 |
Gene Name: | adhesion G protein-coupled receptor G2 |
Gene Aliases: |
EDDM6, GPR64, HE6, TM7LN2 |
Location: |
Chr.X:18989307-19122956 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRG2 | NM_001079858.2 | NP_001073327.1 | ||
| NM_001079859.2 | NP_001073328.1 | |||
| NM_001079860.2 | NP_001073329.1 | |||
| NM_001184833.1 | NP_001171762.1 | |||
| NM_001184834.1 | NP_001171763.1 | |||
| NM_001184835.1 | NP_001171764.1 | |||
| NM_001184836.1 | NP_001171765.1 | |||
| NM_001184837.1 | NP_001171766.1 | |||
| NM_005756.3 | NP_005747.2 | |||
| XM_006724455.3 | XP_006724518.1 | |||
| XM_011545434.1 | XP_011543736.1 | |||
| AF538954.1 | AAN33056.1 | |||
| AF539455.1 | AAN33064.1 | |||
| AF539456.1 | AAN33065.1 | |||
| AK291012.1 | ||||
| AY143364.1 | AAN38971.1 | |||
| AY143365.1 | AAN38972.1 | |||
| AY143366.1 | AAN38973.1 | |||
| AY143367.1 | AAN38974.1 | |||
| AY148343.1 | AAN75702.1 | |||
| BC099901.1 | AAH99901.1 | |||
| X81892.1 | CAA57479.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33517 | Chr.X:18661048 - 19128069 on Build GRCh38 | Loss |
|
| esv3576793 | Chr.X:18780019 - 19298577 on Build GRCh38 | Gain |
|
| esv2752317 | Chr.X:18691267 - 19292303 on Build GRCh38 | Gain |
|
| esv2660427 | Chr.X:19108990 - 19117738 on Build GRCh38 | Deletion |
|
Set Membership: |
|