Search
Search

Entrez Gene ID: | 79783 |
Gene Name: | succinyl-CoA:glutarate-CoA transferase |
Gene Aliases: |
C7orf10, DERP13, GA3, ORF19 |
Location: |
Chr.7:40135005-41038816 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032278 | Chr.7:40057772 - 40356701 on Build GRCh38 | Gain |
|
| nsv4133003 | Chr.7:40181736 - 40362377 on Build GRCh38 | Deletion |
|
| esv3891101 | Chr.7:40263685 - 40320658 on Build GRCh38 | Loss |
|
| dgv11290n54 | Chr.7:40153936 - 40320658 on Build GRCh38 | Gain |
|
| nsv6609716 | Chr.7:40251762 - 40319261 on Build GRCh38 | Deletion |
|
| nsv4140988 | Chr.7:40262712 - 40501511 on Build GRCh38 | Deletion |
|
Set Membership: |
|