Search
Search

Entrez Gene ID: | 5820 |
Gene Name: | Pvt1 oncogene |
Gene Aliases: |
LINC00079, MIR1204HG, NCRNA00079, TP53LC09, onco-lncRNA-100 |
Location: |
Chr.8:127794524-128101256 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4154221 | Chr.8:127936249 - 127947705 on Build GRCh38 | Deletion |
|
| nsv5490037 | Chr.8:127933362 - 127951132 on Build GRCh38 | Deletion |
|
| nsv4435392 | Chr.8:127936059 - 127947705 on Build GRCh38 | Deletion |
|
| esv3572989 | Chr.8:127935979 - 127945629 on Build GRCh38 | Loss |
|
| esv3584895 | Chr.8:127937888 - 127945252 on Build GRCh38 | Loss |
|