Search
Search

Entrez Gene ID: | 11214 |
Gene Name: | A-kinase anchoring protein 13 |
Gene Aliases: |
AKAP-13, AKAP-Lbc, ARHGEF13, BRX, HA-3, Ht31, LBC, PRKA13, PROTO-LB, PROTO-LBC, c-lbc, p47 |
Location: |
Chr.15:85380616-85749358 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKAP13 | NM_001270546.1 | NP_001257475.1 | ||
| NM_006738.5 | NP_006729.4 | |||
| NM_007200.4 | NP_009131.2 | |||
| AB055890.1 | BAB62913.1 | |||
| AF126008.1 | AAD21311.1 | |||
| AF387101.1 | AAL40923.1 | |||
| AF406992.1 | AAL11723.1 | |||
| AK026186.1 | ||||
| AK125331.1 | ||||
| AK307618.1 | ||||
| AK309805.1 | ||||
| AK315885.1 | ||||
| M90360.1 | AAA58670.1 |
Set Membership: |
|