Search
Search

Entrez Gene ID: | 9701 |
Gene Name: | protein phosphatase 6 regulatory subunit 2 |
Gene Aliases: |
KIAA0685, PP6R2, SAP190, SAPS2 |
Location: |
Chr.22:50330774-50445090 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834238 | Chr.22:50402690 - 50556965 on Build GRCh38 | Loss |
|
| nsv3668 | Chr.22:50391793 - 50436883 on Build GRCh38 | Deletion |
|
| nsv6535590 | Chr.22:48991693 - 50757224 on Build GRCh38 | Duplication |
|
Set Membership: |
|