Search
Search

Entrez Gene ID: | 57494 |
Gene Name: | ribosomal modification protein rimK like family member B |
Gene Aliases: |
FAM80B, NAAGS, NAAGS-I |
Location: |
Chr.12:8668638-8783091 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2342n54 | Chr.12:8697651 - 8698961 on Build GRCh38 | Gain |
|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| esv22028 | Chr.12:8697564 - 8699297 on Build GRCh38 | Loss |
|
| dgv2341n54 | Chr.12:8697591 - 8698822 on Build GRCh38 | Loss |
|
| dgv2343n54 | Chr.12:8697754 - 8699250 on Build GRCh38 | Loss |
|
| nsv557353 | Chr.12:8697934 - 8698670 on Build GRCh38 | Loss |
|
Set Membership: |
|