Search
Search

Entrez Gene ID: | 4155 |
Gene Name: | myelin basic protein |
Gene Aliases: |
- |
Location: |
Chr.18:76978833-77133708 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBP | NM_001025100.1 | NP_001020271.1 | ||
| NM_001025101.1 | NP_001020272.1 | |||
| XM_011526009.2 | XP_011524311.1 | |||
| XM_017025778.1 | XP_016881267.1 | |||
| XM_017025780.1 | XP_016881269.1 | |||
| AB208986.1 | BAD92223.1 | |||
| BC065248.1 | AAH65248.1 | |||
| L18865.1 | AAA72010.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2717388 | Chr.18:76288671 - 78160297 on Build GRCh38 | Deletion |
|
| esv3643201 | Chr.18:77032848 - 77365699 on Build GRCh38 | Loss |
|
| nsv1063632 | Chr.18:76317786 - 77170410 on Build GRCh38 | Gain |
|
Set Membership: |
|