Search
Search

Entrez Gene ID: | 79817 |
Gene Name: | MOB kinase activator 3B |
Gene Aliases: |
C9orf35, MOB1D, MOBKL2B |
Location: |
Chr.9:27325209-27529814 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB3B | NM_024761.5 | NP_079037.3 | ||
| XM_047423891.1 | XP_047279847.1 | |||
| XM_047423892.1 | XP_047279848.1 | |||
| XM_047423893.1 | XP_047279849.1 | |||
| XM_047423894.1 | XP_047279850.1 | |||
| XM_047423895.1 | XP_047279851.1 | |||
| AJ580636.1 | CAE45268.1 | |||
| AK023266.1 | BAB14497.1 | |||
| AL832572.2 | CAD89934.1 | |||
| BC033027.1 | AAH33027.1 | |||
| CR457307.1 | CAG33588.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4178699 | Chr.9:26680968 - 27607029 on Build GRCh38 | Duplication |
|
| nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain |
|
| nsv4191010 | Chr.9:27442548 - 27452521 on Build GRCh38 | Deletion |
|
| nsv6009951 | Chr.9:27442549 - 27452522 on Build GRCh38 | Deletion |
|
| nsv6422512 | Chr.9:27283380 - 28841224 on Build GRCh38 | Deletion |
|
Set Membership: |
|