Search
Search

Entrez Gene ID: | 4140 |
Gene Name: | microtubule affinity regulating kinase 3 |
Gene Aliases: |
CTAK1, KP78, PAR1A, Par-1a, VIPB |
Location: |
Chr.14:103385415-103503831 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
Set Membership: |
|