Search
Search

Entrez Gene ID: | 54563 |
Gene Name: | nucleoporin 210 |
Gene Aliases: |
9830001L10, AI836801, Pom210, gp190, gp210 |
Location: |
Chr.6:91013067-91117003 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup210 | NM_018815.2 | 28 | 3772 | NP_061285.2 |
| AF113751.1 | 28 | 3740 | AAF21969.1 | |
| AK036378.1 | 12 | 1369 | ||
| AK090317.1 | 6 | 555 | ||
| AK173056.1 | 28 | 3726 | BAD32334.1 | |
| BC052468.1 | 28 | 3633 | AAH52468.1 |
Set Membership: |
|