Search
Search

Entrez Gene ID: | 71607 |
Gene Name: | sorting nexin 20 |
Gene Aliases: |
9130017C17Rik |
Location: |
Chr.8:88626561-88636133 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snx20 | NM_027840.3 | NP_082116.1 | ||
| AF395844.1 | AAK73125.1 | |||
| AK018632.1 | BAB31317.1 | |||
| AK042068.1 | BAC31150.1 | |||
| AK157032.1 | BAE33939.1 | |||
| AK157884.1 | ||||
| BC039809.1 | AAH39809.1 |
Set Membership: |
|