Search
Search

Entrez Gene ID: | 66855 |
Gene Name: | transcription factor 25 (basic helix-loop-helix) |
Gene Aliases: |
1100001J13Rik, 1810041K11Rik, D8Ertd325e, Nulp1, mKIAA1049 |
Location: |
Chr.8:123373635-123404174 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcf25 | NM_001037877.3 | NP_001032966.1 | ||
| NM_001037878.3 | NP_001032967.1 | |||
| NM_001286362.1 | NP_001273291.1 | |||
| NM_001286363.1 | NP_001273292.1 | |||
| NM_025804.3 | NP_080080.2 | |||
| AK003180.1 | ||||
| AK013754.1 | BAB28983.2 | |||
| AK045397.1 | BAC32344.1 | |||
| AK050274.1 | ||||
| AK075728.1 | ||||
| AK076151.1 | BAC36220.1 | |||
| AK088360.1 | BAC40301.1 | |||
| AK088602.1 | 1 | 203 | ||
| AK089964.1 | BAC41018.1 | |||
| AK122430.1 | BAC65712.1 | |||
| AK148082.1 | BAE28334.1 | |||
| AK150918.1 | BAE29956.1 | |||
| AK150942.1 | BAE29977.1 | |||
| AK151143.1 | BAE30150.1 | |||
| AK151558.1 | ||||
| AK152178.1 | BAE31009.1 | |||
| AK152494.1 | BAE31265.1 | |||
| AK161627.1 | BAE36500.1 | |||
| AK167837.1 | BAE39858.1 | |||
| AK168167.1 | BAE40128.1 | |||
| BC025071.1 | AAH25071.1 | |||
| BC046768.1 | AAH46768.2 | |||
| EU624323.1 | ||||
| U94988.1 | AAG27133.1 |