Search
Search

Entrez Gene ID: | 19935 |
Gene Name: | mitochondrial ribosomal protein L23 |
Gene Aliases: |
L23mrp, Rpl23, Rpl23l |
Location: |
Chr.7:142533030-142540747 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl23 | NM_011288.1 | NP_035418.1 | ||
| AB049646.1 | BAB40851.1 | |||
| AK086805.1 | BAC39746.1 | |||
| BC038626.1 | AAH38626.1 | |||
| BC081460.1 | AAH81460.1 | |||
| U84902.1 | AAC53392.1 |
Set Membership: |
|