Search
Search

Entrez Gene ID: | 84274 |
Gene Name: | coenzyme Q5, methyltransferase |
Gene Aliases: |
- |
Location: |
Chr.12:120503279-120529171 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COQ5 | NM_032314.3 | NP_115690.3 | ||
| XM_006719639.2 | XP_006719702.1 | |||
| AK057777.1 | BAB71567.1 | |||
| AK222610.1 | BAD96330.1 | |||
| AK293656.1 | ||||
| BC004916.2 | AAH04916.2 | |||
| BC107874.1 | AAI07875.1 | |||
| BX647562.1 | CAI46073.1 | |||
| EU700459.1 |
Set Membership: |
|