Search
Search

Entrez Gene ID: | 2495 |
Gene Name: | ferritin heavy chain 1 |
Gene Aliases: |
FHC, FTH, FTHL6, HFE5, PIG15, PLIF |
Location: |
Chr.11:61964285-61967660 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FTH1 | NM_002032.2 | 4 | 998 | NP_002023.2 |
| BM971420.1 | 1 | 248 | ||
| L20941.1 | 4 | 971 | AAA35833.1 | |
| M97164.1 | 4 | 892 | AAA35832.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv975936 | Chr.11:61964459 - 61965666 on Build GRCh38 | Duplication |
|
Set Membership: |
|