Search
Search

Entrez Gene ID: | 6315 |
Gene Name: | ATXN8 opposite strand lncRNA |
Gene Aliases: |
KLHL1AS, NCRNA00003, SCA8 |
Location: |
Chr.13:70107421-70171738 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATXN8OS | NR_002717.3 | |||
| NR_185834.1 | ||||
| NR_185835.1 | ||||
| NR_185836.1 | ||||
| NR_185837.1 | ||||
| NR_185838.1 | ||||
| NR_185839.1 | ||||
| NR_185840.1 | ||||
| NR_185841.1 | ||||
| NR_185842.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051815 | Chr.13:69996822 - 70161932 on Build GRCh38 | Gain |
|
| nsv6484026 | Chr.13:69999075 - 71028657 on Build GRCh38 | Duplication |
|
| nsv1053979 | Chr.13:70079770 - 70208426 on Build GRCh38 | Loss |
|
| nsv6480609 | Chr.13:70033567 - 70135972 on Build GRCh38 | Deletion |
|
| nsv562220 | Chr.13:70066513 - 70131876 on Build GRCh38 | Loss |
|
| nsv6494601 | Chr.13:70079282 - 70219604 on Build GRCh38 | Deletion |
|