Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu482828_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-653-5p
          Stem-loop Accession Number: MI0003674
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 93482760 - 93482855 [-] on Build GRCh38
          Mature miRNA Sequence: GUGUUGAAACAAUCUCUACUG
          Species: Human, Mouse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu482828_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000435, mir-653

          Mature miRNA Sequence:

          GUGUUGAAACAAUCUCUACUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-653-5p MIMAT0003328
          Mouse mmu-miR-653-5p MIMAT0004943
          Pan troglodytes ptr-miR-653 MIMAT0008303
          Pongo pygmaeus ppy-miR-653 MIMAT0016086
          Rhesus monkey mml-miR-653-5p MIMAT0006494

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-653
          Stem-loop Accession # MI0003674
          Stem-loop Sequence
          UUCAUUCCUUCAGUGUUGAAACAAUCUCUACUGAACCAGCUUCAAACAAGUUCACUGGAGUUUGUUUCAAUAUUGCAAGAAUGAUAAGAUGGAAGC
          Chromosome Location Chr. 7 - 93482760 - 93482855 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003328
          miRBase ID: hsa-miR-653-5p
          Mature miRNA Sequence: GUGUUGAAACAAUCUCUACUG
          Chromosome Location: Chr. 7 - 93482760 - 93482855 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-653
          Stem-loop Accession # MI0005557
          Stem-loop Sequence
          CAUUCUUUCAGUGUUGAAACAAUCUCUACUGAACCAAGCUCCAAAGCGAGUUCACUGGAGUUUGUUUCAGUAUUGCAGGAGUGCU
          Chromosome Location Chr. 6 - 3721301 - 3721385 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004943
          miRBase ID: mmu-miR-653-5p
          Mature miRNA Sequence: GUGUUGAAACAAUCUCUACUG
          Chromosome Location: Chr. 6 - 3721301 - 3721385 [-] on Build GRCm38

          Pan troglodytes

          Stem-loop ID ptr-mir-653
          Stem-loop Accession # MI0008840
          Stem-loop Sequence
          UUCAUUCCUUCAGUGUUGAAACAAUCUCUACUGAACCAGCUUCAAACAAGUUCACUGGAGUUUGUUUCAAUAUUGCAAGAAUGAUAAGAUGGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008303
          miRBase ID: ptr-miR-653
          Mature miRNA Sequence: GUGUUGAAACAAUCUCUACUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-653
          Stem-loop Accession # MI0015128
          Stem-loop Sequence
          UUCAUUCCUUCAGUGUUGAAACAAUCUCUACUGAACCAGCUUCAAACAAGUUCACUGGAGUUUGUUUCAAUAUUGCAAGAAUGAUAAGAUGGAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016086
          miRBase ID: ppy-miR-653
          Mature miRNA Sequence: GUGUUGAAACAAUCUCUACUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-653
          Stem-loop Accession # MI0007897
          Stem-loop Sequence
          UUCAUUCCUUCAGUGUUGAAACAAUCUCUACUGAACCAGCUUCAAACAAAUUCACUGGAGUUUGUUUCAAUAUUGCAAGAAUGAUAAGAUGGAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006494
          miRBase ID: mml-miR-653-5p
          Mature miRNA Sequence: GUGUUGAAACAAUCUCUACUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304860_pri
          TaqMan™ Pri-miRNA Assay : Mm03308693_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12482
          Ambion® Pre-miR™ miRNA Precursor : PM12482
          TaqMan™ MicroRNA Assay : 002292
          mirVana® miRNA inhibitor : MH12482
          mirVana® miRNA mimic : MC12482
          TaqMan™ Advanced miRNA Assay : 479134_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline